Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 16560271-16560799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMO3
Entrez Gene ID
55885
Gene Name
LIM domain only 3
Gene Aliases
RBTN3, RBTNL2, RHOM3, Rhom-3
UniGene
Hs.504908
Gene Chromosome Location
Chr.12: 16548372-16610233 on Build GRCh38
Transcripts
NM_001001395.2 NM_001243609.1 NM_001243610.1 NM_001243611.1 NM_001243612.1 NM_001243613.1 NM_018640.4 NR_045012.1 NR_045013.1 NR_045014.1 XM_006719110.3 XM_006719111.2 XM_011520762.1 XM_011520763.1 XM_017019643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6701859 c.294C>T p.Y98Y Chr.12: 16560451-16560451 on Build GRCh38
223528 c.283G>T p.D95Y Chr.12: 16560462-16560462 on Build GRCh38
4540288 c.215G>A p.G72D Chr.12: 16560530-16560530 on Build GRCh38
2003433 c.237C>T p.A79A Chr.12: 16560508-16560508 on Build GRCh38
6701861 c.327C>T p.Y109Y Chr.12: 16560451-16560451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973618169 Chr.12: 16560296-16560296 on Build GRCh38
rs3214385 Chr.12: 16560554-16560555 on Build GRCh38
rs570981354 Chr.12: 16560304-16560304 on Build GRCh38
rs745377837 Chr.12: 16560430-16560430 on Build GRCh38
rs535443745 Chr.12: 16560601-16560601 on Build GRCh38
rs756604706 Chr.12: 16560372-16560372 on Build GRCh38
rs372187914 Chr.12: 16560546-16560546 on Build GRCh38
rs538056260 Chr.12: 16560556-16560556 on Build GRCh38
rs963502422 Chr.12: 16560695-16560695 on Build GRCh38
rs764584553 Chr.12: 16560552-16560553 on Build GRCh38
rs775350108 Chr.12: 16560442-16560442 on Build GRCh38
rs752851249 Chr.12: 16560595-16560595 on Build GRCh38
rs568698857 Chr.12: 16560594-16560594 on Build GRCh38
rs754371324 Chr.12: 16560772-16560772 on Build GRCh38
rs1012667841 Chr.12: 16560607-16560607 on Build GRCh38
rs370435885 Chr.12: 16560737-16560737 on Build GRCh38
rs375115316 Chr.12: 16560692-16560692 on Build GRCh38
rs201967277 Chr.12: 16560578-16560578 on Build GRCh38
rs971094513 Chr.12: 16560628-16560628 on Build GRCh38
rs908085602 Chr.12: 16560354-16560354 on Build GRCh38
rs995075314 Chr.12: 16560705-16560705 on Build GRCh38
rs762735860 Chr.12: 16560458-16560458 on Build GRCh38
rs879289893 Chr.12: 16560347-16560347 on Build GRCh38
rs576139884 Chr.12: 16560746-16560746 on Build GRCh38
rs763567672 Chr.12: 16560363-16560369 on Build GRCh38
rs201376237 Chr.12: 16560371-16560371 on Build GRCh38
rs755449815 Chr.12: 16560393-16560393 on Build GRCh38
rs932050919 Chr.12: 16560508-16560508 on Build GRCh38
rs761974230 Chr.12: 16560487-16560487 on Build GRCh38
rs767665466 Chr.12: 16560493-16560493 on Build GRCh38
rs149014326 Chr.12: 16560514-16560514 on Build GRCh38
rs754368865 Chr.12: 16560523-16560523 on Build GRCh38
rs920709632 Chr.12: 16560475-16560475 on Build GRCh38
rs774536860 Chr.12: 16560479-16560479 on Build GRCh38
rs572132454 Chr.12: 16560673-16560673 on Build GRCh38
rs766406866 Chr.12: 16560764-16560764 on Build GRCh38
rs757101550 Chr.12: 16560556-16560557 on Build GRCh38
rs757297928 Chr.12: 16560368-16560368 on Build GRCh38
rs780725342 Chr.12: 16560552-16560552 on Build GRCh38
rs750449182 Chr.12: 16560507-16560507 on Build GRCh38
rs765029443 Chr.12: 16560740-16560741 on Build GRCh38
rs199844873 Chr.12: 16560567-16560567 on Build GRCh38
rs79952246 Chr.12: 16560554-16560554 on Build GRCh38
rs1015189680 Chr.12: 16560670-16560670 on Build GRCh38
rs777559832 Chr.12: 16560403-16560403 on Build GRCh38
rs950730013 Chr.12: 16560333-16560333 on Build GRCh38
rs749606460 Chr.12: 16560390-16560390 on Build GRCh38
rs191066630 Chr.12: 16560709-16560709 on Build GRCh38
rs397839947 Chr.12: 16560566-16560567 on Build GRCh38
rs1023957542 Chr.12: 16560305-16560305 on Build GRCh38
rs1032830733 Chr.12: 16560598-16560598 on Build GRCh38
rs760804328 Chr.12: 16560758-16560758 on Build GRCh38
rs35463115 Chr.12: 16560554-16560554 on Build GRCh38
rs780429015 Chr.12: 16560387-16560387 on Build GRCh38
rs770538916 Chr.12: 16560408-16560408 on Build GRCh38
rs756926903 Chr.12: 16560545-16560545 on Build GRCh38
rs369974150 Chr.12: 16560432-16560432 on Build GRCh38
rs1044905540 Chr.12: 16560453-16560453 on Build GRCh38
rs1051837387 Chr.12: 16560587-16560587 on Build GRCh38
rs763918054 Chr.12: 16560470-16560470 on Build GRCh38
rs761602150 Chr.12: 16560739-16560739 on Build GRCh38
rs796606859 Chr.12: 16560554-16560555 on Build GRCh38
rs775470192 Chr.12: 16560571-16560571 on Build GRCh38
rs200911029 Chr.12: 16560568-16560568 on Build GRCh38
rs971425296 Chr.12: 16560341-16560341 on Build GRCh38
rs754451798 Chr.12: 16560567-16560568 on Build GRCh38
rs920762010 Chr.12: 16560704-16560704 on Build GRCh38
rs749273263 Chr.12: 16560575-16560575 on Build GRCh38
rs573095189 Chr.12: 16560553-16560554 on Build GRCh38
rs2302690 Chr.12: 16560611-16560611 on Build GRCh38
rs986187953 Chr.12: 16560773-16560773 on Build GRCh38
rs199951012 Chr.12: 16560555-16560556 on Build GRCh38
rs879096400 Chr.12: 16560602-16560602 on Build GRCh38
rs756161447 Chr.12: 16560511-16560511 on Build GRCh38
rs769258227 Chr.12: 16560558-16560558 on Build GRCh38
rs773361894 Chr.12: 16560754-16560754 on Build GRCh38
rs777365034 Chr.12: 16560658-16560658 on Build GRCh38
rs748541526 Chr.12: 16560538-16560538 on Build GRCh38
rs746697540 Chr.12: 16560404-16560404 on Build GRCh38
rs77050222 Chr.12: 16560712-16560712 on Build GRCh38
rs755359866 Chr.12: 16560535-16560535 on Build GRCh38
rs973567147 Chr.12: 16560679-16560679 on Build GRCh38
rs375839550 Chr.12: 16560424-16560424 on Build GRCh38

View Full Product Details