Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 66310469-66311020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HELB
Entrez Gene ID
92797
Gene Name
DNA helicase B
Gene Aliases
DHB, hDHB
UniGene
Hs.505941
Gene Chromosome Location
Chr.12: 66302545-66343643 on Build GRCh38
Transcripts
NM_033647.4 NR_135080.1 NR_135081.1 XM_005269234.2 XM_017020230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4689960 c.1585G>A p.D529N Chr.12: 66310513-66310513 on Build GRCh38
1991917 c.1566G>C p.E522D Chr.12: 66310494-66310494 on Build GRCh38
1166897 c.1616C>G p.P539R Chr.12: 66310544-66310544 on Build GRCh38
1363690 c.1666T>C p.Y556H Chr.12: 66310594-66310594 on Build GRCh38
4920713 c.1588A>T p.K530* Chr.12: 66310516-66310516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772248234 Chr.12: 66310595-66310595 on Build GRCh38
rs984410036 Chr.12: 66310741-66310741 on Build GRCh38
rs911754775 Chr.12: 66310913-66310913 on Build GRCh38
rs147532405 Chr.12: 66310874-66310874 on Build GRCh38
rs143282026 Chr.12: 66310871-66310871 on Build GRCh38
rs747088876 Chr.12: 66310597-66310597 on Build GRCh38
rs535561282 Chr.12: 66310818-66310818 on Build GRCh38
rs986358974 Chr.12: 66310908-66310908 on Build GRCh38
rs7973358 Chr.12: 66310811-66310811 on Build GRCh38
rs757101432 Chr.12: 66310653-66310653 on Build GRCh38
rs911793361 Chr.12: 66310769-66310769 on Build GRCh38
rs181522992 Chr.12: 66310907-66310907 on Build GRCh38
rs767319374 Chr.12: 66310524-66310524 on Build GRCh38
rs967249059 Chr.12: 66310659-66310659 on Build GRCh38
rs972583172 Chr.12: 66310952-66310952 on Build GRCh38
rs916551269 Chr.12: 66310703-66310703 on Build GRCh38
rs1035109023 Chr.12: 66310669-66310669 on Build GRCh38
rs199708480 Chr.12: 66310619-66310619 on Build GRCh38
rs1036857264 Chr.12: 66310766-66310766 on Build GRCh38
rs763463148 Chr.12: 66310639-66310639 on Build GRCh38
rs185642951 Chr.12: 66310748-66310748 on Build GRCh38
rs753650085 Chr.12: 66310562-66310562 on Build GRCh38
rs531304418 Chr.12: 66310650-66310650 on Build GRCh38
rs534035119 Chr.12: 66310894-66310894 on Build GRCh38
rs999685056 Chr.12: 66310856-66310856 on Build GRCh38
rs777847734 Chr.12: 66310548-66310548 on Build GRCh38
rs1010297991 Chr.12: 66310604-66310604 on Build GRCh38
rs960770691 Chr.12: 66310674-66310674 on Build GRCh38
rs913826431 Chr.12: 66310944-66310945 on Build GRCh38
rs889418599 Chr.12: 66310824-66310824 on Build GRCh38
rs11176139 Chr.12: 66310917-66310917 on Build GRCh38
rs571041296 Chr.12: 66310727-66310727 on Build GRCh38
rs933739501 Chr.12: 66310965-66310965 on Build GRCh38
rs922404031 Chr.12: 66310956-66310956 on Build GRCh38
rs148111807 Chr.12: 66310541-66310541 on Build GRCh38
rs1043077554 Chr.12: 66310977-66310977 on Build GRCh38
rs112885700 Chr.12: 66310654-66310654 on Build GRCh38
rs533599392 Chr.12: 66310747-66310747 on Build GRCh38
rs769492316 Chr.12: 66310591-66310591 on Build GRCh38
rs551310435 Chr.12: 66310694-66310694 on Build GRCh38
rs375686074 Chr.12: 66310931-66310931 on Build GRCh38
rs948684020 Chr.12: 66310724-66310724 on Build GRCh38
rs752235269 Chr.12: 66310531-66310531 on Build GRCh38
rs765184274 Chr.12: 66310860-66310860 on Build GRCh38
rs762177690 Chr.12: 66310623-66310623 on Build GRCh38
rs900949535 Chr.12: 66310775-66310775 on Build GRCh38
rs201414904 Chr.12: 66310511-66310511 on Build GRCh38
rs1002106110 Chr.12: 66310845-66310845 on Build GRCh38
rs577856299 Chr.12: 66310882-66310882 on Build GRCh38
rs775067181 Chr.12: 66310713-66310713 on Build GRCh38
rs546873547 Chr.12: 66310753-66310753 on Build GRCh38
rs778948204 Chr.12: 66310658-66310658 on Build GRCh38
rs184414193 Chr.12: 66310989-66310989 on Build GRCh38
rs1043087404 Chr.12: 66310823-66310823 on Build GRCh38
rs773421189 Chr.12: 66310637-66310637 on Build GRCh38
rs780301116 Chr.12: 66310596-66310596 on Build GRCh38
rs978152074 Chr.12: 66310663-66310663 on Build GRCh38
rs371118804 Chr.12: 66310647-66310647 on Build GRCh38
rs754546915 Chr.12: 66310518-66310518 on Build GRCh38
rs1027404939 Chr.12: 66310895-66310895 on Build GRCh38
rs968827401 Chr.12: 66310628-66310628 on Build GRCh38
rs761021635 Chr.12: 66310548-66310549 on Build GRCh38
rs768441813 Chr.12: 66310801-66310801 on Build GRCh38
rs374925671 Chr.12: 66310649-66310649 on Build GRCh38
rs141898186 Chr.12: 66310621-66310621 on Build GRCh38
rs189199692 Chr.12: 66310828-66310828 on Build GRCh38
rs200580401 Chr.12: 66310496-66310496 on Build GRCh38
rs762390862 Chr.12: 66310500-66310500 on Build GRCh38
rs770368804 Chr.12: 66310634-66310634 on Build GRCh38
rs909719482 Chr.12: 66310742-66310742 on Build GRCh38
rs150033325 Chr.12: 66310940-66310940 on Build GRCh38
rs944671252 Chr.12: 66310773-66310773 on Build GRCh38
rs757231594 Chr.12: 66310567-66310567 on Build GRCh38
rs573835088 Chr.12: 66310916-66310916 on Build GRCh38
rs990810408 Chr.12: 66310680-66310680 on Build GRCh38
rs946010612 Chr.12: 66310953-66310953 on Build GRCh38
rs141956990 Chr.12: 66310496-66310496 on Build GRCh38
rs765093964 Chr.12: 66310606-66310606 on Build GRCh38
rs1014486077 Chr.12: 66310865-66310865 on Build GRCh38
rs537213940 Chr.12: 66310864-66310864 on Build GRCh38
rs767268025 Chr.12: 66310643-66310643 on Build GRCh38
rs139061595 Chr.12: 66310846-66310846 on Build GRCh38
rs753405877 Chr.12: 66310652-66310652 on Build GRCh38
rs528392944 Chr.12: 66310578-66310578 on Build GRCh38
rs200233536 Chr.12: 66310646-66310646 on Build GRCh38
rs984806490 Chr.12: 66310921-66310921 on Build GRCh38
rs879667943 Chr.12: 66310848-66310848 on Build GRCh38
rs548544614 Chr.12: 66310679-66310679 on Build GRCh38
rs750821718 Chr.12: 66310512-66310512 on Build GRCh38
rs1015198450 Chr.12: 66310870-66310870 on Build GRCh38

View Full Product Details