Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 92141838-92142380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTG1
Entrez Gene ID
694
Gene Name
BTG anti-proliferation factor 1
Gene Aliases
APRO2
UniGene
Hs.255935
Gene Chromosome Location
Chr.12: 92140278-92145897 on Build GRCh38
Transcripts
NM_001731.2 NR_046160.2 NR_132339.1 NR_132340.1 NR_132341.1

Gene Symbol
LINC01619
Entrez Gene ID
256021
Gene Name
long intergenic non-protein coding RNA 1619
Gene Aliases
C12orf79
UniGene
Hs.651357
Gene Chromosome Location
Chr.12: 91984976-92142831 on Build GRCh38
Transcripts
NM_001731.2 NR_046160.2 NR_132339.1 NR_132340.1 NR_132341.1

SNP Detail

SNP ID Chromosome Location
rs146332177 Chr.12: 92142031-92142031 on Build GRCh38
rs909763334 Chr.12: 92141946-92141946 on Build GRCh38
rs561726012 Chr.12: 92142230-92142230 on Build GRCh38
rs914208007 Chr.12: 92142111-92142111 on Build GRCh38
rs572696774 Chr.12: 92142318-92142318 on Build GRCh38
rs751975020 Chr.12: 92142057-92142057 on Build GRCh38
rs1049978062 Chr.12: 92142116-92142116 on Build GRCh38
rs964886692 Chr.12: 92142046-92142046 on Build GRCh38
rs879910506 Chr.12: 92141900-92141900 on Build GRCh38
rs766628353 Chr.12: 92141957-92141957 on Build GRCh38
rs892023513 Chr.12: 92142197-92142197 on Build GRCh38
rs933734558 Chr.12: 92142085-92142085 on Build GRCh38
rs1006174783 Chr.12: 92142339-92142340 on Build GRCh38
rs567007332 Chr.12: 92142333-92142333 on Build GRCh38
rs79497339 Chr.12: 92141910-92141911 on Build GRCh38
rs1034887638 Chr.12: 92142319-92142319 on Build GRCh38
rs923058378 Chr.12: 92142048-92142048 on Build GRCh38
rs927302212 Chr.12: 92141891-92141891 on Build GRCh38
rs930672881 Chr.12: 92141918-92141918 on Build GRCh38
rs559935771 Chr.12: 92142307-92142307 on Build GRCh38
rs988499313 Chr.12: 92142092-92142092 on Build GRCh38
rs1056577970 Chr.12: 92141923-92141923 on Build GRCh38
rs545783079 Chr.12: 92141915-92141915 on Build GRCh38
rs781639865 Chr.12: 92142325-92142326 on Build GRCh38
rs763230441 Chr.12: 92141919-92141919 on Build GRCh38
rs187402457 Chr.12: 92141965-92141965 on Build GRCh38
rs896585361 Chr.12: 92142302-92142302 on Build GRCh38
rs753265664 Chr.12: 92142227-92142227 on Build GRCh38
rs532384660 Chr.12: 92142345-92142345 on Build GRCh38
rs76386052 Chr.12: 92141911-92141911 on Build GRCh38
rs889458322 Chr.12: 92142123-92142123 on Build GRCh38
rs549862250 Chr.12: 92142317-92142317 on Build GRCh38
rs976637387 Chr.12: 92142047-92142047 on Build GRCh38
rs905730482 Chr.12: 92141912-92141912 on Build GRCh38
rs1024461342 Chr.12: 92142263-92142263 on Build GRCh38
rs1006393849 Chr.12: 92141978-92141978 on Build GRCh38
rs999826140 Chr.12: 92142125-92142125 on Build GRCh38
rs546210127 Chr.12: 92142336-92142336 on Build GRCh38
rs980056908 Chr.12: 92142131-92142131 on Build GRCh38
rs1054132582 Chr.12: 92142175-92142175 on Build GRCh38
rs993523560 Chr.12: 92141936-92141936 on Build GRCh38
rs887811625 Chr.12: 92142323-92142323 on Build GRCh38
rs1057336755 Chr.12: 92142250-92142250 on Build GRCh38
rs530933846 Chr.12: 92141971-92141972 on Build GRCh38
rs983998964 Chr.12: 92141932-92141932 on Build GRCh38
rs542057890 Chr.12: 92142154-92142154 on Build GRCh38
rs541796990 Chr.12: 92142139-92142139 on Build GRCh38
rs751865743 Chr.12: 92141921-92141924 on Build GRCh38
rs997713016 Chr.12: 92142352-92142352 on Build GRCh38
rs768019194 Chr.12: 92142069-92142069 on Build GRCh38
rs576936964 Chr.12: 92141899-92141899 on Build GRCh38
rs191972874 Chr.12: 92142253-92142253 on Build GRCh38
rs773356842 Chr.12: 92141913-92141913 on Build GRCh38
rs900737649 Chr.12: 92142348-92142348 on Build GRCh38

View Full Product Details