Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 26215505-26216012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF6
Entrez Gene ID
6049
Gene Name
ring finger protein 6
Gene Aliases
-
UniGene
Hs.136885
Gene Chromosome Location
Chr.13: 26132115-26222559 on Build GRCh38
Transcripts
NM_005977.3 NM_183043.2 NM_183044.2 NM_183045.1 XM_005266485.2 XM_005266486.2 XM_011535177.2 XM_011535178.1 XM_017020685.1 XM_017020686.1 XM_017020687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6768030 c.373C>T p.R125* Chr.13: 26215509-26215509 on Build GRCh38
2069779 c.374G>A p.R125Q Chr.13: 26215508-26215508 on Build GRCh38
72464 c.370A>G p.T124A Chr.13: 26215512-26215512 on Build GRCh38
7138822 c.354G>A p.R118R Chr.13: 26215528-26215528 on Build GRCh38
6768041 c.307G>T p.E103* Chr.13: 26215575-26215575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527491725 Chr.13: 26215822-26215822 on Build GRCh38
rs149345283 Chr.13: 26215882-26215882 on Build GRCh38
rs773803704 Chr.13: 26215559-26215559 on Build GRCh38
rs927738343 Chr.13: 26215854-26215854 on Build GRCh38
rs577809435 Chr.13: 26215969-26215969 on Build GRCh38
rs1021375948 Chr.13: 26215541-26215541 on Build GRCh38
rs986352006 Chr.13: 26215761-26215761 on Build GRCh38
rs993439441 Chr.13: 26215714-26215714 on Build GRCh38
rs1016279379 Chr.13: 26215697-26215697 on Build GRCh38
rs529271179 Chr.13: 26215534-26215534 on Build GRCh38
rs544421359 Chr.13: 26215909-26215909 on Build GRCh38
rs766865470 Chr.13: 26215589-26215589 on Build GRCh38
rs574956916 Chr.13: 26215908-26215908 on Build GRCh38
rs570639878 Chr.13: 26215752-26215752 on Build GRCh38
rs765384693 Chr.13: 26215621-26215621 on Build GRCh38
rs888373295 Chr.13: 26215665-26215665 on Build GRCh38
rs745827769 Chr.13: 26215638-26215638 on Build GRCh38
rs371597920 Chr.13: 26215530-26215530 on Build GRCh38
rs569216154 Chr.13: 26215540-26215540 on Build GRCh38
rs1052363492 Chr.13: 26215976-26215976 on Build GRCh38
rs1046089163 Chr.13: 26215842-26215842 on Build GRCh38
rs121434522 Chr.13: 26215577-26215577 on Build GRCh38
rs780923575 Chr.13: 26215636-26215636 on Build GRCh38
rs1054974163 Chr.13: 26215875-26215875 on Build GRCh38
rs368385230 Chr.13: 26215615-26215615 on Build GRCh38
rs752671145 Chr.13: 26215622-26215622 on Build GRCh38
rs937719098 Chr.13: 26215864-26215864 on Build GRCh38
rs549462514 Chr.13: 26215536-26215536 on Build GRCh38
rs138213113 Chr.13: 26215829-26215829 on Build GRCh38
rs779887506 Chr.13: 26215644-26215644 on Build GRCh38
rs1005847149 Chr.13: 26215677-26215677 on Build GRCh38
rs141660942 Chr.13: 26215529-26215529 on Build GRCh38
rs754099680 Chr.13: 26215594-26215594 on Build GRCh38
rs561609077 Chr.13: 26215753-26215753 on Build GRCh38
rs922650650 Chr.13: 26215952-26215952 on Build GRCh38
rs539780247 Chr.13: 26215803-26215803 on Build GRCh38
rs530742227 Chr.13: 26215707-26215707 on Build GRCh38
rs760883219 Chr.13: 26215581-26215581 on Build GRCh38
rs542990280 Chr.13: 26215716-26215716 on Build GRCh38
rs758534703 Chr.13: 26215634-26215634 on Build GRCh38
rs557956236 Chr.13: 26215938-26215938 on Build GRCh38
rs934007124 Chr.13: 26215974-26215975 on Build GRCh38
rs114008142 Chr.13: 26215808-26215808 on Build GRCh38
rs370729289 Chr.13: 26215830-26215830 on Build GRCh38

View Full Product Details