Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39177749-39178248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNN
Entrez Gene ID
5411
Gene Name
pinin, desmosome associated protein
Gene Aliases
DRS, DRSP, SDK3, memA
UniGene
Hs.409965
Gene Chromosome Location
Chr.14: 39175183-39183218 on Build GRCh38
Transcripts
NM_002687.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
304535 c.467T>A p.F156Y Chr.14: 39177885-39177885 on Build GRCh38
6751388 c.426C>T p.N142N Chr.14: 39177844-39177844 on Build GRCh38
3419790 c.475G>T p.E159* Chr.14: 39177893-39177893 on Build GRCh38
6751397 c.428G>A p.R143Q Chr.14: 39177846-39177846 on Build GRCh38
7430404 c.461A>G p.Q154R Chr.14: 39177879-39177879 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756188429 Chr.14: 39177963-39177963 on Build GRCh38
rs1032860731 Chr.14: 39178201-39178201 on Build GRCh38
rs574483083 Chr.14: 39177970-39177970 on Build GRCh38
rs10148712 Chr.14: 39178202-39178202 on Build GRCh38
rs1036603386 Chr.14: 39178106-39178106 on Build GRCh38
rs547679060 Chr.14: 39178098-39178098 on Build GRCh38
rs981973466 Chr.14: 39178022-39178022 on Build GRCh38
rs1138657 Chr.14: 39177862-39177862 on Build GRCh38
rs997455784 Chr.14: 39178105-39178105 on Build GRCh38
rs200540809 Chr.14: 39177944-39177944 on Build GRCh38
rs1030365219 Chr.14: 39177947-39177947 on Build GRCh38
rs567657580 Chr.14: 39178118-39178118 on Build GRCh38
rs898118105 Chr.14: 39178111-39178111 on Build GRCh38
rs1016057161 Chr.14: 39178145-39178145 on Build GRCh38
rs530001416 Chr.14: 39178172-39178172 on Build GRCh38
rs774729452 Chr.14: 39177866-39177866 on Build GRCh38
rs766286129 Chr.14: 39177946-39177946 on Build GRCh38
rs753744697 Chr.14: 39177802-39177802 on Build GRCh38
rs750363513 Chr.14: 39177809-39177809 on Build GRCh38
rs778629112 Chr.14: 39178168-39178168 on Build GRCh38
rs771229020 Chr.14: 39177848-39177848 on Build GRCh38
rs371604285 Chr.14: 39177832-39177832 on Build GRCh38
rs781286725 Chr.14: 39177817-39177817 on Build GRCh38
rs889292593 Chr.14: 39178166-39178166 on Build GRCh38
rs897980804 Chr.14: 39177906-39177906 on Build GRCh38
rs888113135 Chr.14: 39177821-39177821 on Build GRCh38
rs893382270 Chr.14: 39178010-39178010 on Build GRCh38
rs184778740 Chr.14: 39178188-39178188 on Build GRCh38
rs1065608 Chr.14: 39177880-39177880 on Build GRCh38
rs1061586 Chr.14: 39177871-39177871 on Build GRCh38
rs763642358 Chr.14: 39177921-39177921 on Build GRCh38
rs1138656 Chr.14: 39177847-39177847 on Build GRCh38
rs759396856 Chr.14: 39177832-39177833 on Build GRCh38
rs953503350 Chr.14: 39177983-39177983 on Build GRCh38
rs939699340 Chr.14: 39178076-39178076 on Build GRCh38
rs570142086 Chr.14: 39177775-39177775 on Build GRCh38
rs3064996 Chr.14: 39177953-39177957 on Build GRCh38
rs775174968 Chr.14: 39177962-39177963 on Build GRCh38
rs958293987 Chr.14: 39177986-39177986 on Build GRCh38
rs145557069 Chr.14: 39178144-39178145 on Build GRCh38
rs68193573 Chr.14: 39177984-39177984 on Build GRCh38
rs551588084 Chr.14: 39177885-39177885 on Build GRCh38
rs368955851 Chr.14: 39177830-39177830 on Build GRCh38
rs763100329 Chr.14: 39177897-39177897 on Build GRCh38
rs369957940 Chr.14: 39177935-39177935 on Build GRCh38
rs777788910 Chr.14: 39177965-39177965 on Build GRCh38
rs750873658 Chr.14: 39178063-39178063 on Build GRCh38
rs764968215 Chr.14: 39177927-39177927 on Build GRCh38
rs776488810 Chr.14: 39177949-39177950 on Build GRCh38
rs202226032 Chr.14: 39177977-39177977 on Build GRCh38
rs2753605 Chr.14: 39177783-39177783 on Build GRCh38
rs1046160160 Chr.14: 39177782-39177782 on Build GRCh38
rs749444445 Chr.14: 39177846-39177846 on Build GRCh38
rs913842267 Chr.14: 39178011-39178011 on Build GRCh38
rs527905085 Chr.14: 39178052-39178052 on Build GRCh38
rs570023098 Chr.14: 39178189-39178189 on Build GRCh38
rs866971890 Chr.14: 39177985-39177985 on Build GRCh38
rs545245821 Chr.14: 39178003-39178003 on Build GRCh38
rs879566949 Chr.14: 39178157-39178157 on Build GRCh38
rs563198404 Chr.14: 39177990-39177990 on Build GRCh38
rs780685815 Chr.14: 39178059-39178059 on Build GRCh38
rs201450370 Chr.14: 39177807-39177807 on Build GRCh38
rs199829484 Chr.14: 39177799-39177799 on Build GRCh38
rs756487572 Chr.14: 39178071-39178071 on Build GRCh38
rs762449656 Chr.14: 39178021-39178021 on Build GRCh38
rs981767203 Chr.14: 39178205-39178205 on Build GRCh38
rs761489994 Chr.14: 39177925-39177925 on Build GRCh38
rs768133302 Chr.14: 39178029-39178029 on Build GRCh38
rs971977753 Chr.14: 39178162-39178162 on Build GRCh38
rs1046155269 Chr.14: 39177976-39177977 on Build GRCh38
rs757301393 Chr.14: 39177928-39177928 on Build GRCh38
rs565118151 Chr.14: 39178018-39178018 on Build GRCh38
rs963633365 Chr.14: 39178145-39178145 on Build GRCh38
rs1051842699 Chr.14: 39178124-39178124 on Build GRCh38
rs5808025 Chr.14: 39177950-39177954 on Build GRCh38
rs765018173 Chr.14: 39177808-39177808 on Build GRCh38
rs532239634 Chr.14: 39177993-39177993 on Build GRCh38
rs371973976 Chr.14: 39177811-39177811 on Build GRCh38
rs937736695 Chr.14: 39178058-39178058 on Build GRCh38
rs953453961 Chr.14: 39178141-39178141 on Build GRCh38
rs949128329 Chr.14: 39177973-39177973 on Build GRCh38
rs750214059 Chr.14: 39177932-39177932 on Build GRCh38
rs777981655 Chr.14: 39177845-39177845 on Build GRCh38
rs775207139 Chr.14: 39177889-39177889 on Build GRCh38

View Full Product Details