Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64748918-64749424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPTB
Entrez Gene ID
6710
Gene Name
spectrin beta, erythrocytic
Gene Aliases
EL3, HS2, HSPTB1, SPH2
UniGene
Hs.417303
Gene Chromosome Location
Chr.14: 64746283-64879892 on Build GRCh38
Transcripts
NM_001024858.2 XM_005268023.4 XM_017021612.1 XM_017021613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6076272 c.6922C>A p.P2308T Chr.14: 64749371-64749371 on Build GRCh38
7174680 c.6917C>T p.S2306F Chr.14: 64749376-64749376 on Build GRCh38
4827893 c.6891G>A p.A2297A Chr.14: 64749402-64749402 on Build GRCh38
4388570 c.6928G>A p.A2310T Chr.14: 64749365-64749365 on Build GRCh38
401491 c.6892C>T p.Q2298* Chr.14: 64749401-64749401 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs953975604 Chr.14: 64749181-64749181 on Build GRCh38
rs962678299 Chr.14: 64749186-64749186 on Build GRCh38
rs115474163 Chr.14: 64749257-64749257 on Build GRCh38
rs967332391 Chr.14: 64749224-64749224 on Build GRCh38
rs756408647 Chr.14: 64749277-64749277 on Build GRCh38
rs563633753 Chr.14: 64749107-64749107 on Build GRCh38
rs746861808 Chr.14: 64749305-64749305 on Build GRCh38
rs532527939 Chr.14: 64749146-64749146 on Build GRCh38
rs764314389 Chr.14: 64749357-64749357 on Build GRCh38
rs978173546 Chr.14: 64749052-64749052 on Build GRCh38
rs901983270 Chr.14: 64749175-64749182 on Build GRCh38
rs201133464 Chr.14: 64749286-64749286 on Build GRCh38
rs768011952 Chr.14: 64749271-64749271 on Build GRCh38
rs918347434 Chr.14: 64749220-64749220 on Build GRCh38
rs1017313849 Chr.14: 64749154-64749154 on Build GRCh38
rs1031447749 Chr.14: 64749058-64749058 on Build GRCh38
rs769300381 Chr.14: 64749312-64749312 on Build GRCh38
rs955564434 Chr.14: 64749212-64749212 on Build GRCh38
rs757486021 Chr.14: 64749372-64749372 on Build GRCh38
rs768297594 Chr.14: 64749308-64749308 on Build GRCh38
rs990018882 Chr.14: 64749150-64749150 on Build GRCh38
rs918300914 Chr.14: 64749342-64749342 on Build GRCh38
rs948286328 Chr.14: 64749240-64749240 on Build GRCh38
rs1036824427 Chr.14: 64749102-64749102 on Build GRCh38
rs147254132 Chr.14: 64749335-64749335 on Build GRCh38
rs999562428 Chr.14: 64749139-64749139 on Build GRCh38
rs1028343175 Chr.14: 64749190-64749190 on Build GRCh38
rs985139023 Chr.14: 64749086-64749086 on Build GRCh38
rs182353062 Chr.14: 64749013-64749013 on Build GRCh38
rs1024915785 Chr.14: 64749129-64749129 on Build GRCh38
rs747325010 Chr.14: 64749309-64749311 on Build GRCh38
rs957039290 Chr.14: 64749383-64749383 on Build GRCh38
rs893904967 Chr.14: 64749274-64749274 on Build GRCh38
rs758529551 Chr.14: 64749377-64749377 on Build GRCh38
rs943722034 Chr.14: 64749217-64749217 on Build GRCh38
rs780072609 Chr.14: 64749390-64749390 on Build GRCh38
rs377266593 Chr.14: 64749351-64749351 on Build GRCh38
rs745803591 Chr.14: 64749291-64749291 on Build GRCh38
rs748362034 Chr.14: 64749356-64749367 on Build GRCh38
rs753168397 Chr.14: 64749272-64749272 on Build GRCh38
rs550173997 Chr.14: 64749358-64749358 on Build GRCh38
rs552641164 Chr.14: 64749215-64749215 on Build GRCh38
rs1031079064 Chr.14: 64749144-64749144 on Build GRCh38
rs970797220 Chr.14: 64749000-64749000 on Build GRCh38
rs559973555 Chr.14: 64749231-64749231 on Build GRCh38
rs370207013 Chr.14: 64749315-64749315 on Build GRCh38
rs780775844 Chr.14: 64749263-64749264 on Build GRCh38
rs776687653 Chr.14: 64749342-64749344 on Build GRCh38
rs753963756 Chr.14: 64749283-64749283 on Build GRCh38
rs931508539 Chr.14: 64749105-64749105 on Build GRCh38
rs57421986 Chr.14: 64749402-64749402 on Build GRCh38
rs750587649 Chr.14: 64749375-64749375 on Build GRCh38
rs1021577593 Chr.14: 64749338-64749338 on Build GRCh38
rs925702440 Chr.14: 64749069-64749069 on Build GRCh38
rs772202684 Chr.14: 64749297-64749297 on Build GRCh38
rs536702051 Chr.14: 64749356-64749356 on Build GRCh38
rs367706006 Chr.14: 64749234-64749235 on Build GRCh38
rs912261667 Chr.14: 64749096-64749096 on Build GRCh38
rs928221807 Chr.14: 64749394-64749394 on Build GRCh38
rs761418574 Chr.14: 64749260-64749260 on Build GRCh38
rs1012251210 Chr.14: 64748962-64748962 on Build GRCh38
rs746873092 Chr.14: 64749396-64749396 on Build GRCh38
rs367617790 Chr.14: 64749374-64749374 on Build GRCh38
rs935772441 Chr.14: 64749077-64749077 on Build GRCh38
rs546919926 Chr.14: 64749115-64749115 on Build GRCh38
rs987091860 Chr.14: 64749042-64749042 on Build GRCh38
rs769116949 Chr.14: 64749339-64749341 on Build GRCh38
rs55839068 Chr.14: 64748951-64748954 on Build GRCh38
rs769616625 Chr.14: 64749141-64749141 on Build GRCh38
rs985398481 Chr.14: 64749196-64749200 on Build GRCh38
rs149576500 Chr.14: 64749049-64749049 on Build GRCh38
rs1014084019 Chr.14: 64749108-64749108 on Build GRCh38
rs1041739178 Chr.14: 64749064-64749064 on Build GRCh38
rs761315155 Chr.14: 64749311-64749311 on Build GRCh38
rs757432912 Chr.14: 64749285-64749285 on Build GRCh38
rs897321656 Chr.14: 64749104-64749109 on Build GRCh38
rs369148076 Chr.14: 64749282-64749282 on Build GRCh38
rs779842361 Chr.14: 64749301-64749301 on Build GRCh38
rs773243167 Chr.14: 64749143-64749143 on Build GRCh38
rs768065442 Chr.14: 64749346-64749346 on Build GRCh38
rs895106619 Chr.14: 64748960-64748960 on Build GRCh38
rs914175934 Chr.14: 64749221-64749221 on Build GRCh38
rs914124623 Chr.14: 64749045-64749045 on Build GRCh38
rs921466819 Chr.14: 64748990-64748990 on Build GRCh38
rs1055552966 Chr.14: 64749082-64749082 on Build GRCh38
rs990464698 Chr.14: 64749065-64749065 on Build GRCh38
rs570395215 Chr.14: 64749368-64749368 on Build GRCh38
rs776370834 Chr.14: 64749310-64749310 on Build GRCh38
rs543839169 Chr.14: 64749097-64749097 on Build GRCh38
rs935637087 Chr.14: 64749249-64749249 on Build GRCh38
rs996925338 Chr.14: 64749189-64749189 on Build GRCh38
rs776434582 Chr.14: 64749084-64749085 on Build GRCh38
rs979701631 Chr.14: 64749228-64749228 on Build GRCh38
rs991130228 Chr.14: 64749387-64749387 on Build GRCh38
rs374895714 Chr.14: 64749288-64749288 on Build GRCh38
rs780786493 Chr.14: 64749403-64749403 on Build GRCh38
rs772778745 Chr.14: 64749313-64749313 on Build GRCh38
rs762351158 Chr.14: 64749241-64749241 on Build GRCh38
rs552287579 Chr.14: 64748954-64748954 on Build GRCh38
rs906567571 Chr.14: 64749131-64749131 on Build GRCh38
rs868339479 Chr.14: 64748983-64748983 on Build GRCh38
rs759972947 Chr.14: 64749270-64749270 on Build GRCh38
rs867725744 Chr.14: 64749365-64749365 on Build GRCh38
rs1004218224 Chr.14: 64749145-64749145 on Build GRCh38
rs1024989357 Chr.14: 64748992-64748992 on Build GRCh38
rs867043971 Chr.14: 64749230-64749230 on Build GRCh38
rs895329845 Chr.14: 64749236-64749236 on Build GRCh38
rs374987034 Chr.14: 64749261-64749261 on Build GRCh38
rs558266782 Chr.14: 64749219-64749220 on Build GRCh38
rs752077415 Chr.14: 64749269-64749269 on Build GRCh38
rs887002876 Chr.14: 64749128-64749128 on Build GRCh38
rs916752152 Chr.14: 64749369-64749369 on Build GRCh38

View Full Product Details