Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64753455-64753960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPTB
Entrez Gene ID
6710
Gene Name
spectrin beta, erythrocytic
Gene Aliases
EL3, HS2, HSPTB1, SPH2
UniGene
Hs.417303
Gene Chromosome Location
Chr.14: 64746283-64879892 on Build GRCh38
Transcripts
NM_001024858.2 XM_005268023.4 XM_017021612.1 XM_017021613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6318830 c.6561C>A p.R2187R Chr.14: 64753578-64753578 on Build GRCh38
5607631 c.6497C>T p.P2166L Chr.14: 64753642-64753642 on Build GRCh38
4148179 c.6509C>G p.P2170R Chr.14: 64753630-64753630 on Build GRCh38
3793792 c.6516G>A p.P2172P Chr.14: 64753623-64753623 on Build GRCh38
4148180 c.6507G>T p.L2169L Chr.14: 64753632-64753632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544875952 Chr.14: 64753875-64753875 on Build GRCh38
rs368147978 Chr.14: 64753843-64753843 on Build GRCh38
rs764403195 Chr.14: 64753549-64753549 on Build GRCh38
rs776175032 Chr.14: 64753503-64753503 on Build GRCh38
rs971672545 Chr.14: 64753602-64753602 on Build GRCh38
rs200556500 Chr.14: 64753747-64753747 on Build GRCh38
rs916548631 Chr.14: 64753895-64753895 on Build GRCh38
rs558224489 Chr.14: 64753554-64753554 on Build GRCh38
rs777203819 Chr.14: 64753736-64753736 on Build GRCh38
rs778713147 Chr.14: 64753846-64753846 on Build GRCh38
rs890288381 Chr.14: 64753675-64753675 on Build GRCh38
rs756733142 Chr.14: 64753787-64753787 on Build GRCh38
rs757383953 Chr.14: 64753561-64753561 on Build GRCh38
rs1019799716 Chr.14: 64753785-64753785 on Build GRCh38
rs776233124 Chr.14: 64753584-64753584 on Build GRCh38
rs143820600 Chr.14: 64753656-64753656 on Build GRCh38
rs754726157 Chr.14: 64753630-64753630 on Build GRCh38
rs765947895 Chr.14: 64753825-64753825 on Build GRCh38
rs371440636 Chr.14: 64753777-64753777 on Build GRCh38
rs139236988 Chr.14: 64753624-64753624 on Build GRCh38
rs983009166 Chr.14: 64753830-64753830 on Build GRCh38
rs978895532 Chr.14: 64753615-64753615 on Build GRCh38
rs146688698 Chr.14: 64753621-64753621 on Build GRCh38
rs571733505 Chr.14: 64753778-64753778 on Build GRCh38
rs1012311534 Chr.14: 64753565-64753565 on Build GRCh38
rs190118021 Chr.14: 64753740-64753740 on Build GRCh38
rs756353822 Chr.14: 64753548-64753548 on Build GRCh38
rs545218935 Chr.14: 64753918-64753918 on Build GRCh38
rs770438205 Chr.14: 64753698-64753698 on Build GRCh38
rs187385876 Chr.14: 64753629-64753629 on Build GRCh38
rs548137633 Chr.14: 64753689-64753689 on Build GRCh38
rs984373626 Chr.14: 64753876-64753876 on Build GRCh38
rs973447142 Chr.14: 64753871-64753871 on Build GRCh38
rs141953610 Chr.14: 64753922-64753922 on Build GRCh38
rs769241247 Chr.14: 64753599-64753599 on Build GRCh38
rs144452926 Chr.14: 64753567-64753567 on Build GRCh38
rs747462256 Chr.14: 64753714-64753714 on Build GRCh38
rs761083364 Chr.14: 64753612-64753612 on Build GRCh38
rs759888913 Chr.14: 64753604-64753604 on Build GRCh38
rs768060224 Chr.14: 64753605-64753605 on Build GRCh38
rs373600922 Chr.14: 64753657-64753657 on Build GRCh38
rs200751290 Chr.14: 64753746-64753746 on Build GRCh38
rs767426834 Chr.14: 64753768-64753768 on Build GRCh38
rs769148718 Chr.14: 64753719-64753719 on Build GRCh38
rs190896895 Chr.14: 64753528-64753528 on Build GRCh38
rs745368836 Chr.14: 64753791-64753791 on Build GRCh38
rs187773838 Chr.14: 64753744-64753744 on Build GRCh38
rs575946264 Chr.14: 64753820-64753820 on Build GRCh38
rs900347871 Chr.14: 64753479-64753479 on Build GRCh38
rs746806448 Chr.14: 64753579-64753579 on Build GRCh38
rs769095162 Chr.14: 64753649-64753649 on Build GRCh38
rs780451289 Chr.14: 64753701-64753701 on Build GRCh38
rs779648071 Chr.14: 64753573-64753573 on Build GRCh38
rs747738552 Chr.14: 64753635-64753635 on Build GRCh38
rs149033754 Chr.14: 64753770-64753770 on Build GRCh38
rs146516991 Chr.14: 64753928-64753928 on Build GRCh38
rs748814015 Chr.14: 64753641-64753641 on Build GRCh38
rs567906349 Chr.14: 64753481-64753481 on Build GRCh38
rs775232504 Chr.14: 64753494-64753494 on Build GRCh38
rs536822649 Chr.14: 64753489-64753489 on Build GRCh38
rs1037475281 Chr.14: 64753651-64753651 on Build GRCh38
rs777007887 Chr.14: 64753653-64753653 on Build GRCh38
rs145280081 Chr.14: 64753740-64753741 on Build GRCh38
rs745701182 Chr.14: 64753487-64753487 on Build GRCh38
rs375625321 Chr.14: 64753692-64753692 on Build GRCh38
rs187548635 Chr.14: 64753666-64753666 on Build GRCh38
rs755875675 Chr.14: 64753776-64753776 on Build GRCh38
rs553245049 Chr.14: 64753620-64753620 on Build GRCh38
rs775319859 Chr.14: 64753854-64753854 on Build GRCh38
rs112462309 Chr.14: 64753640-64753640 on Build GRCh38
rs769948840 Chr.14: 64753742-64753742 on Build GRCh38
rs148132963 Chr.14: 64753905-64753905 on Build GRCh38
rs748553498 Chr.14: 64753797-64753797 on Build GRCh38
rs201270587 Chr.14: 64753502-64753502 on Build GRCh38
rs774322325 Chr.14: 64753548-64753550 on Build GRCh38
rs535592697 Chr.14: 64753693-64753693 on Build GRCh38
rs778420646 Chr.14: 64753790-64753790 on Build GRCh38
rs781754468 Chr.14: 64753796-64753796 on Build GRCh38
rs775369407 Chr.14: 64753838-64753838 on Build GRCh38
rs904173609 Chr.14: 64753885-64753885 on Build GRCh38
rs754077015 Chr.14: 64753616-64753616 on Build GRCh38
rs375291013 Chr.14: 64753534-64753534 on Build GRCh38
rs770146235 Chr.14: 64753798-64753798 on Build GRCh38
rs1052921955 Chr.14: 64753935-64753935 on Build GRCh38
rs759442638 Chr.14: 64753578-64753578 on Build GRCh38
rs764972508 Chr.14: 64753748-64753749 on Build GRCh38
rs774798643 Chr.14: 64753522-64753522 on Build GRCh38
rs774849809 Chr.14: 64753601-64753601 on Build GRCh38
rs778974940 Chr.14: 64753486-64753486 on Build GRCh38
rs2295523 Chr.14: 64753864-64753864 on Build GRCh38
rs576450384 Chr.14: 64753792-64753792 on Build GRCh38
rs200555048 Chr.14: 64753495-64753495 on Build GRCh38
rs866418806 Chr.14: 64753828-64753828 on Build GRCh38
rs771050794 Chr.14: 64753811-64753811 on Build GRCh38
rs182745580 Chr.14: 64753743-64753743 on Build GRCh38
rs779026292 Chr.14: 64753562-64753562 on Build GRCh38
rs747685779 Chr.14: 64753597-64753597 on Build GRCh38
rs376845836 Chr.14: 64753748-64753748 on Build GRCh38
rs368734255 Chr.14: 64753783-64753783 on Build GRCh38
rs775779339 Chr.14: 64753606-64753606 on Build GRCh38
rs750154106 Chr.14: 64753783-64753785 on Build GRCh38
rs543215760 Chr.14: 64753873-64753873 on Build GRCh38
rs576022847 Chr.14: 64753505-64753505 on Build GRCh38
rs754023731 Chr.14: 64753550-64753550 on Build GRCh38
rs749457089 Chr.14: 64753808-64753808 on Build GRCh38
rs377565646 Chr.14: 64753665-64753665 on Build GRCh38
rs143163263 Chr.14: 64753782-64753782 on Build GRCh38
rs191850273 Chr.14: 64753636-64753636 on Build GRCh38
rs760607339 Chr.14: 64753632-64753632 on Build GRCh38
rs780017738 Chr.14: 64753625-64753625 on Build GRCh38
rs780895917 Chr.14: 64753687-64753687 on Build GRCh38
rs750379513 Chr.14: 64753623-64753623 on Build GRCh38
rs144051169 Chr.14: 64753643-64753643 on Build GRCh38
rs375816870 Chr.14: 64753655-64753655 on Build GRCh38
rs199938084 Chr.14: 64753580-64753580 on Build GRCh38
rs766909882 Chr.14: 64753510-64753510 on Build GRCh38
rs555587693 Chr.14: 64753805-64753806 on Build GRCh38
rs754778995 Chr.14: 64753672-64753672 on Build GRCh38
rs746584891 Chr.14: 64753497-64753497 on Build GRCh38
rs373540763 Chr.14: 64753614-64753614 on Build GRCh38
rs992923811 Chr.14: 64753763-64753763 on Build GRCh38
rs768488051 Chr.14: 64753684-64753684 on Build GRCh38
rs780844613 Chr.14: 64753633-64753633 on Build GRCh38
rs201552144 Chr.14: 64753801-64753801 on Build GRCh38
rs374329599 Chr.14: 64753529-64753529 on Build GRCh38
rs776417574 Chr.14: 64753736-64753753 on Build GRCh38
rs1022797315 Chr.14: 64753575-64753575 on Build GRCh38

View Full Product Details