Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96391729-96392256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK7
Entrez Gene ID
122481
Gene Name
adenylate kinase 7
Gene Aliases
AK 7
UniGene
Hs.667462
Gene Chromosome Location
Chr.14: 96392111-96489427 on Build GRCh38
Transcripts
NM_152327.3 XM_006720021.2 XM_006720022.2 XM_011536403.2 XM_017020956.1 XM_017020957.1 XM_017020958.1 XM_017020959.1 XM_017020960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5670327 c.65T>G p.I22R Chr.14: 96392219-96392219 on Build GRCh38
6597359 c.71T>C p.L24P Chr.14: 96392225-96392225 on Build GRCh38
459085 c.76G>T p.D26Y Chr.14: 96392230-96392230 on Build GRCh38
5549430 c.5C>A p.A2D Chr.14: 96392159-96392159 on Build GRCh38
4595728 c.52C>G p.Q18E Chr.14: 96392206-96392206 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs918853079 Chr.14: 96391952-96391952 on Build GRCh38
rs775195771 Chr.14: 96392132-96392132 on Build GRCh38
rs144597609 Chr.14: 96392200-96392200 on Build GRCh38
rs12891542 Chr.14: 96392235-96392235 on Build GRCh38
rs561626913 Chr.14: 96391949-96391949 on Build GRCh38
rs774547959 Chr.14: 96392197-96392197 on Build GRCh38
rs12891202 Chr.14: 96392086-96392086 on Build GRCh38
rs369760162 Chr.14: 96392121-96392121 on Build GRCh38
rs867228692 Chr.14: 96391983-96391983 on Build GRCh38
rs909590097 Chr.14: 96391943-96391943 on Build GRCh38
rs559142105 Chr.14: 96392120-96392120 on Build GRCh38
rs1036079215 Chr.14: 96391842-96391842 on Build GRCh38
rs756331532 Chr.14: 96392112-96392112 on Build GRCh38
rs983152136 Chr.14: 96391973-96391973 on Build GRCh38
rs1012022371 Chr.14: 96392062-96392062 on Build GRCh38
rs781570202 Chr.14: 96392110-96392110 on Build GRCh38
rs955654043 Chr.14: 96392161-96392163 on Build GRCh38
rs773853288 Chr.14: 96392233-96392233 on Build GRCh38
rs766558630 Chr.14: 96392168-96392168 on Build GRCh38
rs769399562 Chr.14: 96392131-96392131 on Build GRCh38
rs779070716 Chr.14: 96392184-96392184 on Build GRCh38
rs746971909 Chr.14: 96392122-96392122 on Build GRCh38
rs966503818 Chr.14: 96391817-96391817 on Build GRCh38
rs1018867749 Chr.14: 96391775-96391775 on Build GRCh38
rs750725074 Chr.14: 96392111-96392111 on Build GRCh38
rs754059777 Chr.14: 96392182-96392182 on Build GRCh38
rs1034897062 Chr.14: 96392043-96392043 on Build GRCh38
rs1054759498 Chr.14: 96392053-96392053 on Build GRCh38
rs770941736 Chr.14: 96392126-96392126 on Build GRCh38
rs111419100 Chr.14: 96392205-96392205 on Build GRCh38
rs899981290 Chr.14: 96392194-96392194 on Build GRCh38
rs111918026 Chr.14: 96391997-96391997 on Build GRCh38
rs902435625 Chr.14: 96392006-96392006 on Build GRCh38
rs867470948 Chr.14: 96391766-96391766 on Build GRCh38
rs1042304131 Chr.14: 96392003-96392003 on Build GRCh38
rs937249158 Chr.14: 96392052-96392052 on Build GRCh38
rs959161860 Chr.14: 96391902-96391902 on Build GRCh38
rs1054843457 Chr.14: 96391977-96391977 on Build GRCh38
rs893168574 Chr.14: 96391986-96391986 on Build GRCh38
rs1010850358 Chr.14: 96391783-96391783 on Build GRCh38
rs544887722 Chr.14: 96392145-96392145 on Build GRCh38
rs563271490 Chr.14: 96392161-96392161 on Build GRCh38
rs540819426 Chr.14: 96391972-96391972 on Build GRCh38
rs951650584 Chr.14: 96392229-96392229 on Build GRCh38
rs1035145129 Chr.14: 96391866-96391866 on Build GRCh38
rs1008347357 Chr.14: 96391765-96391765 on Build GRCh38
rs757267586 Chr.14: 96392193-96392193 on Build GRCh38
rs949786623 Chr.14: 96392012-96392012 on Build GRCh38
rs564346655 Chr.14: 96392019-96392019 on Build GRCh38
rs1020784354 Chr.14: 96391802-96391802 on Build GRCh38
rs780314773 Chr.14: 96392117-96392117 on Build GRCh38
rs775049122 Chr.14: 96392208-96392208 on Build GRCh38
rs749003834 Chr.14: 96392210-96392210 on Build GRCh38
rs370931258 Chr.14: 96392156-96392156 on Build GRCh38
rs934121563 Chr.14: 96391969-96391969 on Build GRCh38
rs372110819 Chr.14: 96392042-96392042 on Build GRCh38
rs533020457 Chr.14: 96392130-96392130 on Build GRCh38
rs748181906 Chr.14: 96392192-96392192 on Build GRCh38
rs573688301 Chr.14: 96391951-96391951 on Build GRCh38
rs111944358 Chr.14: 96391813-96391813 on Build GRCh38
rs768148434 Chr.14: 96392227-96392227 on Build GRCh38
rs756454123 Chr.14: 96392166-96392166 on Build GRCh38
rs893082113 Chr.14: 96392081-96392081 on Build GRCh38
rs893711325 Chr.14: 96391780-96391780 on Build GRCh38
rs981237937 Chr.14: 96392027-96392027 on Build GRCh38
rs981958651 Chr.14: 96391855-96391855 on Build GRCh38
rs964299814 Chr.14: 96391795-96391795 on Build GRCh38
rs907624501 Chr.14: 96391978-96391978 on Build GRCh38
rs907065752 Chr.14: 96392071-96392071 on Build GRCh38
rs974749028 Chr.14: 96391942-96391942 on Build GRCh38
rs911417725 Chr.14: 96392231-96392231 on Build GRCh38
rs12891217 Chr.14: 96392152-96392152 on Build GRCh38
rs927130012 Chr.14: 96392028-96392028 on Build GRCh38
rs1018421193 Chr.14: 96392190-96392190 on Build GRCh38
rs962326034 Chr.14: 96392088-96392088 on Build GRCh38
rs1010094755 Chr.14: 96392075-96392094 on Build GRCh38
rs1010253068 Chr.14: 96391987-96391987 on Build GRCh38
rs762318097 Chr.14: 96392155-96392155 on Build GRCh38
rs749382043 Chr.14: 96392118-96392118 on Build GRCh38
rs941598449 Chr.14: 96391947-96391947 on Build GRCh38
rs776582512 Chr.14: 96392129-96392129 on Build GRCh38
rs368458386 Chr.14: 96392142-96392142 on Build GRCh38

View Full Product Details