Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3516330-3516729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPV3
Entrez Gene ID
162514
Gene Name
transient receptor potential cation channel subfamily V member 3
Gene Aliases
FNEPPK2, OLMS, VRL3
UniGene
Hs.446255
Gene Chromosome Location
Chr.17: 3510502-3557995 on Build GRCh38
Transcripts
NM_001258205.1 NM_145068.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6195273 c.2197C>A p.R733R Chr.17: 3516458-3516458 on Build GRCh38
6195272 c.2197C>A p.R733R Chr.17: 3516458-3516458 on Build GRCh38
5055392 c.2089G>T p.A697S Chr.17: 3516566-3516566 on Build GRCh38
6261996 c.2086-5C>A p.? Chr.17: 3516574-3516574 on Build GRCh38
6261995 c.2086-5C>A p.? Chr.17: 3516574-3516574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539339155 Chr.17: 3516469-3516469 on Build GRCh38
rs1014918662 Chr.17: 3516571-3516571 on Build GRCh38
rs980938833 Chr.17: 3516388-3516388 on Build GRCh38
rs751891074 Chr.17: 3516503-3516503 on Build GRCh38
rs11654533 Chr.17: 3516470-3516470 on Build GRCh38
rs756150733 Chr.17: 3516508-3516508 on Build GRCh38
rs369069738 Chr.17: 3516558-3516558 on Build GRCh38
rs200251594 Chr.17: 3516574-3516574 on Build GRCh38
rs8081785 Chr.17: 3516395-3516395 on Build GRCh38
rs781688821 Chr.17: 3516506-3516506 on Build GRCh38
rs767621452 Chr.17: 3516447-3516447 on Build GRCh38
rs956437857 Chr.17: 3516364-3516364 on Build GRCh38
rs558778917 Chr.17: 3516509-3516509 on Build GRCh38
rs879613617 Chr.17: 3516395-3516396 on Build GRCh38
rs919318859 Chr.17: 3516432-3516432 on Build GRCh38
rs757451377 Chr.17: 3516654-3516654 on Build GRCh38
rs977526304 Chr.17: 3516677-3516677 on Build GRCh38
rs771270573 Chr.17: 3516533-3516533 on Build GRCh38
rs759455415 Chr.17: 3516683-3516683 on Build GRCh38
rs911618938 Chr.17: 3516369-3516369 on Build GRCh38
rs749435514 Chr.17: 3516528-3516528 on Build GRCh38
rs375812083 Chr.17: 3516482-3516482 on Build GRCh38
rs1039519595 Chr.17: 3516513-3516513 on Build GRCh38
rs1056594542 Chr.17: 3516431-3516431 on Build GRCh38
rs1033930562 Chr.17: 3516701-3516701 on Build GRCh38
rs141628122 Chr.17: 3516591-3516591 on Build GRCh38
rs749523445 Chr.17: 3516439-3516439 on Build GRCh38
rs764073922 Chr.17: 3516457-3516457 on Build GRCh38
rs935631285 Chr.17: 3516684-3516684 on Build GRCh38
rs150932901 Chr.17: 3516491-3516491 on Build GRCh38
rs748237281 Chr.17: 3516418-3516418 on Build GRCh38
rs759968033 Chr.17: 3516431-3516431 on Build GRCh38
rs779314465 Chr.17: 3516540-3516540 on Build GRCh38
rs769829281 Chr.17: 3516575-3516575 on Build GRCh38
rs773442081 Chr.17: 3516434-3516434 on Build GRCh38
rs769784660 Chr.17: 3516426-3516426 on Build GRCh38
rs542492740 Chr.17: 3516692-3516692 on Build GRCh38
rs772707052 Chr.17: 3516582-3516582 on Build GRCh38
rs371447802 Chr.17: 3516542-3516542 on Build GRCh38
rs767776070 Chr.17: 3516609-3516609 on Build GRCh38
rs1003976845 Chr.17: 3516557-3516557 on Build GRCh38
rs968266392 Chr.17: 3516387-3516387 on Build GRCh38
rs755506245 Chr.17: 3516406-3516406 on Build GRCh38
rs370497774 Chr.17: 3516599-3516599 on Build GRCh38
rs8082339 Chr.17: 3516396-3516396 on Build GRCh38
rs574856332 Chr.17: 3516662-3516662 on Build GRCh38
rs1009253519 Chr.17: 3516637-3516637 on Build GRCh38
rs762489873 Chr.17: 3516590-3516590 on Build GRCh38
rs915799633 Chr.17: 3516700-3516700 on Build GRCh38
rs368817028 Chr.17: 3516504-3516504 on Build GRCh38
rs375957310 Chr.17: 3516483-3516483 on Build GRCh38
rs759903820 Chr.17: 3516602-3516602 on Build GRCh38
rs911656172 Chr.17: 3516640-3516640 on Build GRCh38
rs201696477 Chr.17: 3516606-3516606 on Build GRCh38
rs986293454 Chr.17: 3516368-3516368 on Build GRCh38
rs771932965 Chr.17: 3516440-3516440 on Build GRCh38
rs906594232 Chr.17: 3516547-3516547 on Build GRCh38
rs986294538 Chr.17: 3516631-3516631 on Build GRCh38
rs948577530 Chr.17: 3516703-3516703 on Build GRCh38
rs78767235 Chr.17: 3516641-3516641 on Build GRCh38
rs781588929 Chr.17: 3516413-3516413 on Build GRCh38
rs386794632 Chr.17: 3516395-3516396 on Build GRCh38
rs776459642 Chr.17: 3516458-3516458 on Build GRCh38
rs79268479 Chr.17: 3516367-3516367 on Build GRCh38
rs566915761 Chr.17: 3516452-3516452 on Build GRCh38

View Full Product Details