Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7262799-7263309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN7
Entrez Gene ID
1366
Gene Name
claudin 7
Gene Aliases
CEPTRL2, CLDN-7, CPETRL2, Hs.84359, claudin-1
UniGene
Hs.513915
Gene Chromosome Location
Chr.17: 7259903-7263193 on Build GRCh38
Transcripts
NM_001185022.1 NM_001185023.1 NM_001307.5

SNP Detail

SNP ID Chromosome Location
rs1015007659 Chr.17: 7262860-7262860 on Build GRCh38
rs931932390 Chr.17: 7263091-7263091 on Build GRCh38
rs1040257920 Chr.17: 7263056-7263056 on Build GRCh38
rs780571096 Chr.17: 7262908-7262908 on Build GRCh38
rs944317709 Chr.17: 7263148-7263163 on Build GRCh38
rs919405496 Chr.17: 7263106-7263106 on Build GRCh38
rs192262608 Chr.17: 7262943-7262943 on Build GRCh38
rs561870245 Chr.17: 7262962-7262962 on Build GRCh38
rs1002503196 Chr.17: 7263072-7263072 on Build GRCh38
rs748400960 Chr.17: 7263219-7263219 on Build GRCh38
rs1006218809 Chr.17: 7263147-7263147 on Build GRCh38
rs887788196 Chr.17: 7263141-7263141 on Build GRCh38
rs545899911 Chr.17: 7262842-7262842 on Build GRCh38
rs1050824230 Chr.17: 7263127-7263127 on Build GRCh38
rs981047204 Chr.17: 7263238-7263238 on Build GRCh38
rs781781183 Chr.17: 7263120-7263120 on Build GRCh38
rs796914082 Chr.17: 7262856-7262856 on Build GRCh38
rs913017973 Chr.17: 7263044-7263044 on Build GRCh38
rs374458234 Chr.17: 7263051-7263051 on Build GRCh38
rs1014955078 Chr.17: 7263184-7263184 on Build GRCh38
rs35710187 Chr.17: 7263132-7263132 on Build GRCh38
rs142756120 Chr.17: 7262846-7262846 on Build GRCh38
rs758838454 Chr.17: 7262883-7262883 on Build GRCh38
rs143304921 Chr.17: 7262975-7262975 on Build GRCh38
rs753298999 Chr.17: 7262834-7262834 on Build GRCh38
rs1053232858 Chr.17: 7263223-7263223 on Build GRCh38
rs1029014350 Chr.17: 7262841-7262841 on Build GRCh38
rs556193383 Chr.17: 7262873-7262873 on Build GRCh38
rs989893881 Chr.17: 7263040-7263040 on Build GRCh38
rs970009012 Chr.17: 7263225-7263225 on Build GRCh38
rs754281884 Chr.17: 7263155-7263155 on Build GRCh38
rs540915520 Chr.17: 7263125-7263125 on Build GRCh38
rs945852888 Chr.17: 7263054-7263054 on Build GRCh38
rs998657472 Chr.17: 7262905-7262905 on Build GRCh38
rs947254744 Chr.17: 7263283-7263283 on Build GRCh38
rs980145963 Chr.17: 7262956-7262956 on Build GRCh38
rs901933810 Chr.17: 7262904-7262904 on Build GRCh38
rs976082096 Chr.17: 7263192-7263192 on Build GRCh38
rs1035363930 Chr.17: 7263083-7263083 on Build GRCh38
rs747325905 Chr.17: 7262955-7262955 on Build GRCh38
rs1011891722 Chr.17: 7262986-7262986 on Build GRCh38
rs137931761 Chr.17: 7262935-7262936 on Build GRCh38
rs897938120 Chr.17: 7263188-7263188 on Build GRCh38
rs985422068 Chr.17: 7263176-7263176 on Build GRCh38
rs957050892 Chr.17: 7263263-7263263 on Build GRCh38
rs972543154 Chr.17: 7263098-7263098 on Build GRCh38
rs962183181 Chr.17: 7262874-7262874 on Build GRCh38
rs923302818 Chr.17: 7263214-7263214 on Build GRCh38
rs371045446 Chr.17: 7262900-7262900 on Build GRCh38
rs781453939 Chr.17: 7263138-7263138 on Build GRCh38
rs1013682568 Chr.17: 7263197-7263197 on Build GRCh38
rs910536331 Chr.17: 7262858-7262858 on Build GRCh38
rs935709228 Chr.17: 7263020-7263020 on Build GRCh38
rs1033044889 Chr.17: 7263245-7263245 on Build GRCh38
rs910819119 Chr.17: 7263181-7263181 on Build GRCh38
rs901736550 Chr.17: 7263087-7263087 on Build GRCh38
rs775894437 Chr.17: 7262917-7262917 on Build GRCh38
rs576961998 Chr.17: 7262830-7262830 on Build GRCh38
rs969684358 Chr.17: 7262942-7262942 on Build GRCh38

View Full Product Details