Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27879547-27880052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THEMIS2
Entrez Gene ID
9473
Gene Name
thymocyte selection associated family member 2
Gene Aliases
C1orf38, ICB-1
UniGene
Hs.10649
Gene Chromosome Location
Chr.1: 27872531-27886685 on Build GRCh38
Transcripts
NM_001039477.2 NM_001105556.2 NM_001286113.1 NM_001286115.1 NM_004848.3 XM_005246041.3 XM_006711050.1 XM_011542445.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6801208 c.569T>C p.L190P Chr.1: 27879977-27879977 on Build GRCh38
5569167 c.375T>C p.T125T Chr.1: 27879783-27879783 on Build GRCh38
139276 c.419G>A p.G140E Chr.1: 27879827-27879827 on Build GRCh38
4497438 c.496C>T p.P166S Chr.1: 27879904-27879904 on Build GRCh38
6875927 c.527G>A p.R176Q Chr.1: 27879935-27879935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867054094 Chr.1: 27879755-27879755 on Build GRCh38
rs201225150 Chr.1: 27879929-27879929 on Build GRCh38
rs200209595 Chr.1: 27879773-27879773 on Build GRCh38
rs199929444 Chr.1: 27879695-27879696 on Build GRCh38
rs764238122 Chr.1: 27879865-27879865 on Build GRCh38
rs763173212 Chr.1: 27879731-27879731 on Build GRCh38
rs200496526 Chr.1: 27879850-27879850 on Build GRCh38
rs751514629 Chr.1: 27879989-27879989 on Build GRCh38
rs372894304 Chr.1: 27879746-27879746 on Build GRCh38
rs1015985965 Chr.1: 27879588-27879588 on Build GRCh38
rs757941847 Chr.1: 27879903-27879903 on Build GRCh38
rs774829838 Chr.1: 27879842-27879842 on Build GRCh38
rs774398082 Chr.1: 27879747-27879747 on Build GRCh38
rs868330041 Chr.1: 27879945-27879945 on Build GRCh38
rs1004181083 Chr.1: 27879580-27879580 on Build GRCh38
rs779512916 Chr.1: 27879665-27879665 on Build GRCh38
rs750799391 Chr.1: 27879599-27879599 on Build GRCh38
rs755841148 Chr.1: 27880002-27880002 on Build GRCh38
rs879034401 Chr.1: 27879645-27879645 on Build GRCh38
rs757693342 Chr.1: 27880008-27880008 on Build GRCh38
rs774159361 Chr.1: 27879773-27879773 on Build GRCh38
rs776137764 Chr.1: 27879696-27879696 on Build GRCh38
rs78644103 Chr.1: 27879837-27879837 on Build GRCh38
rs780646435 Chr.1: 27879822-27879822 on Build GRCh38
rs746675739 Chr.1: 27879675-27879675 on Build GRCh38
rs770357358 Chr.1: 27879699-27879699 on Build GRCh38
rs768698160 Chr.1: 27879935-27879935 on Build GRCh38
rs920520443 Chr.1: 27879625-27879625 on Build GRCh38
rs768112330 Chr.1: 27879688-27879688 on Build GRCh38
rs751391949 Chr.1: 27879886-27879886 on Build GRCh38
rs201129055 Chr.1: 27879832-27879832 on Build GRCh38
rs201674442 Chr.1: 27879955-27879955 on Build GRCh38
rs372068799 Chr.1: 27879617-27879617 on Build GRCh38
rs895406030 Chr.1: 27879734-27879734 on Build GRCh38
rs766403774 Chr.1: 27879984-27879984 on Build GRCh38
rs571279225 Chr.1: 27879958-27879958 on Build GRCh38
rs202053560 Chr.1: 27879986-27879986 on Build GRCh38
rs777119411 Chr.1: 27880003-27880003 on Build GRCh38
rs767683149 Chr.1: 27879761-27879761 on Build GRCh38
rs150226809 Chr.1: 27879831-27879831 on Build GRCh38
rs753781345 Chr.1: 27879640-27879640 on Build GRCh38
rs778739366 Chr.1: 27879657-27879657 on Build GRCh38
rs369425095 Chr.1: 27879749-27879749 on Build GRCh38
rs35953961 Chr.1: 27879587-27879587 on Build GRCh38
rs762102564 Chr.1: 27879760-27879760 on Build GRCh38
rs151326930 Chr.1: 27879597-27879597 on Build GRCh38
rs757255562 Chr.1: 27879769-27879769 on Build GRCh38
rs747718205 Chr.1: 27879826-27879826 on Build GRCh38
rs771740449 Chr.1: 27879663-27879663 on Build GRCh38
rs41284294 Chr.1: 27879739-27879739 on Build GRCh38
rs766061596 Chr.1: 27879881-27879881 on Build GRCh38
rs746291214 Chr.1: 27879940-27879940 on Build GRCh38
rs758381094 Chr.1: 27879880-27879880 on Build GRCh38
rs143326302 Chr.1: 27879706-27879706 on Build GRCh38
rs983978064 Chr.1: 27879927-27879927 on Build GRCh38
rs747806049 Chr.1: 27879907-27879907 on Build GRCh38
rs145503231 Chr.1: 27879697-27879697 on Build GRCh38
rs755541788 Chr.1: 27879921-27879921 on Build GRCh38
rs777367255 Chr.1: 27879923-27879923 on Build GRCh38
rs1056608063 Chr.1: 27879717-27879717 on Build GRCh38
rs766571282 Chr.1: 27879607-27879607 on Build GRCh38
rs377762719 Chr.1: 27880007-27880007 on Build GRCh38
rs894711082 Chr.1: 27879775-27879775 on Build GRCh38
rs867811365 Chr.1: 27879905-27879905 on Build GRCh38
rs532896556 Chr.1: 27879948-27879948 on Build GRCh38
rs139829655 Chr.1: 27880022-27880022 on Build GRCh38
rs550315014 Chr.1: 27880021-27880021 on Build GRCh38
rs559721374 Chr.1: 27879852-27879852 on Build GRCh38
rs944417245 Chr.1: 27879691-27879691 on Build GRCh38
rs758061218 Chr.1: 27879793-27879793 on Build GRCh38
rs1045846619 Chr.1: 27879825-27879825 on Build GRCh38
rs781454382 Chr.1: 27879639-27879639 on Build GRCh38
rs149302250 Chr.1: 27879833-27879833 on Build GRCh38
rs763974146 Chr.1: 27879653-27879653 on Build GRCh38
rs146409624 Chr.1: 27879658-27879658 on Build GRCh38
rs985951269 Chr.1: 27879668-27879668 on Build GRCh38
rs752300534 Chr.1: 27879998-27879998 on Build GRCh38
rs759861782 Chr.1: 27879748-27879748 on Build GRCh38
rs779927473 Chr.1: 27879798-27879798 on Build GRCh38
rs376428161 Chr.1: 27879808-27879808 on Build GRCh38
rs759263906 Chr.1: 27879990-27879990 on Build GRCh38
rs760777328 Chr.1: 27879861-27879861 on Build GRCh38
rs776819505 Chr.1: 27879978-27879978 on Build GRCh38
rs750284195 Chr.1: 27879871-27879871 on Build GRCh38
rs138336447 Chr.1: 27879784-27879784 on Build GRCh38
rs780769284 Chr.1: 27879904-27879904 on Build GRCh38
rs186451576 Chr.1: 27879606-27879606 on Build GRCh38
rs754670690 Chr.1: 27879818-27879818 on Build GRCh38
rs573092977 Chr.1: 27879577-27879577 on Build GRCh38
rs752865176 Chr.1: 27879762-27879762 on Build GRCh38
rs757308881 Chr.1: 27879649-27879649 on Build GRCh38
rs767334851 Chr.1: 27879995-27879995 on Build GRCh38
rs201157658 Chr.1: 27879956-27879956 on Build GRCh38
rs375175632 Chr.1: 27879623-27879623 on Build GRCh38
rs749737835 Chr.1: 27879926-27879926 on Build GRCh38
rs762763832 Chr.1: 27879979-27879979 on Build GRCh38
rs375892526 Chr.1: 27879888-27879888 on Build GRCh38
rs771153130 Chr.1: 27879841-27879841 on Build GRCh38

View Full Product Details