Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 31365514-31366020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FABP3
Entrez Gene ID
2170
Gene Name
fatty acid binding protein 3
Gene Aliases
FABP11, H-FABP, M-FABP, MDGI, O-FABP
UniGene
Hs.576372
Gene Chromosome Location
Chr.1: 31359611-31373618 on Build GRCh38
Transcripts
NM_001320996.1 NM_004102.4 XM_011541007.2 XM_006710681.3 XM_011541567.2

Gene Symbol
ZCCHC17
Entrez Gene ID
51538
Gene Name
zinc finger CCHC-type containing 17
Gene Aliases
HSPC251, PS1D, pNO40
UniGene
Hs.524094
Gene Chromosome Location
Chr.1: 31296982-31365764 on Build GRCh38
Transcripts
NM_001320996.1 NM_004102.4 XM_011541007.2 XM_006710681.3 XM_011541567.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5550882 c.373T>C p.C125R Chr.1: 31365915-31365915 on Build GRCh38
4031354 c.361G>A p.G121S Chr.1: 31365927-31365927 on Build GRCh38
4682273 c.375C>T p.C125C Chr.1: 31365913-31365913 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs577433423 Chr.1: 31365594-31365594 on Build GRCh38
rs542648280 Chr.1: 31365729-31365729 on Build GRCh38
rs200751117 Chr.1: 31365927-31365927 on Build GRCh38
rs553283923 Chr.1: 31365628-31365628 on Build GRCh38
rs368661026 Chr.1: 31365833-31365833 on Build GRCh38
rs931649685 Chr.1: 31365636-31365636 on Build GRCh38
rs376904316 Chr.1: 31365913-31365913 on Build GRCh38
rs533593819 Chr.1: 31365814-31365814 on Build GRCh38
rs747004314 Chr.1: 31365987-31365987 on Build GRCh38
rs779101907 Chr.1: 31365982-31365982 on Build GRCh38
rs201298570 Chr.1: 31365920-31365920 on Build GRCh38
rs1045393777 Chr.1: 31365789-31365789 on Build GRCh38
rs746180937 Chr.1: 31365542-31365542 on Build GRCh38
rs748434286 Chr.1: 31365890-31365890 on Build GRCh38
rs138959172 Chr.1: 31365918-31365918 on Build GRCh38
rs564709104 Chr.1: 31365716-31365716 on Build GRCh38
rs766128256 Chr.1: 31365963-31365964 on Build GRCh38
rs940491237 Chr.1: 31365725-31365725 on Build GRCh38
rs572732210 Chr.1: 31365755-31365755 on Build GRCh38
rs974032329 Chr.1: 31365798-31365799 on Build GRCh38
rs529266933 Chr.1: 31365907-31365907 on Build GRCh38
rs764806410 Chr.1: 31365926-31365926 on Build GRCh38
rs957482855 Chr.1: 31365849-31365849 on Build GRCh38
rs5773353 Chr.1: 31365705-31365706 on Build GRCh38
rs752102311 Chr.1: 31365939-31365939 on Build GRCh38
rs1013462709 Chr.1: 31365785-31365785 on Build GRCh38
rs534569223 Chr.1: 31365965-31365965 on Build GRCh38
rs903363514 Chr.1: 31365585-31365585 on Build GRCh38
rs377335936 Chr.1: 31365600-31365600 on Build GRCh38
rs749331943 Chr.1: 31365905-31365905 on Build GRCh38
rs770963031 Chr.1: 31365971-31365971 on Build GRCh38
rs374828766 Chr.1: 31365832-31365832 on Build GRCh38
rs775307287 Chr.1: 31365909-31365909 on Build GRCh38
rs141085536 Chr.1: 31365778-31365778 on Build GRCh38
rs367796823 Chr.1: 31365872-31365872 on Build GRCh38
rs951271877 Chr.1: 31365828-31365828 on Build GRCh38
rs183264861 Chr.1: 31365772-31365772 on Build GRCh38
rs150723706 Chr.1: 31365928-31365928 on Build GRCh38
rs966291248 Chr.1: 31365603-31365603 on Build GRCh38
rs947667163 Chr.1: 31365572-31365572 on Build GRCh38
rs1048168 Chr.1: 31365901-31365901 on Build GRCh38
rs953055346 Chr.1: 31365966-31365966 on Build GRCh38
rs534605042 Chr.1: 31365552-31365552 on Build GRCh38
rs778545299 Chr.1: 31365797-31365797 on Build GRCh38
rs483352865 Chr.1: 31365893-31365893 on Build GRCh38
rs374764850 Chr.1: 31365964-31365964 on Build GRCh38
rs371423628 Chr.1: 31365837-31365837 on Build GRCh38
rs764017250 Chr.1: 31365917-31365917 on Build GRCh38
rs557051059 Chr.1: 31365625-31365625 on Build GRCh38
rs765891427 Chr.1: 31365930-31365930 on Build GRCh38
rs776191827 Chr.1: 31365858-31365860 on Build GRCh38
rs549991256 Chr.1: 31365819-31365819 on Build GRCh38
rs999667791 Chr.1: 31365602-31365602 on Build GRCh38
rs12064676 Chr.1: 31365667-31365667 on Build GRCh38
rs976759838 Chr.1: 31365610-31365610 on Build GRCh38
rs483352875 Chr.1: 31365952-31365953 on Build GRCh38
rs397979796 Chr.1: 31365722-31365723 on Build GRCh38
rs769892963 Chr.1: 31365897-31365897 on Build GRCh38
rs564785601 Chr.1: 31365704-31365705 on Build GRCh38
rs761233222 Chr.1: 31365894-31365895 on Build GRCh38
rs764811590 Chr.1: 31365930-31365930 on Build GRCh38
rs781506707 Chr.1: 31365883-31365883 on Build GRCh38
rs374809975 Chr.1: 31365848-31365848 on Build GRCh38
rs431905488 Chr.1: 31365958-31365958 on Build GRCh38
rs924754516 Chr.1: 31365941-31365941 on Build GRCh38
rs189871114 Chr.1: 31365574-31365574 on Build GRCh38
rs371992036 Chr.1: 31365874-31365874 on Build GRCh38
rs562241959 Chr.1: 31365733-31365733 on Build GRCh38
rs202201820 Chr.1: 31365843-31365843 on Build GRCh38
rs755473631 Chr.1: 31365957-31365957 on Build GRCh38
rs768731381 Chr.1: 31365990-31365990 on Build GRCh38
rs879224212 Chr.1: 31365705-31365705 on Build GRCh38
rs762695038 Chr.1: 31365954-31365955 on Build GRCh38
rs12064630 Chr.1: 31365593-31365593 on Build GRCh38
rs890789325 Chr.1: 31365719-31365720 on Build GRCh38
rs908985604 Chr.1: 31365723-31365723 on Build GRCh38

View Full Product Details