Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28733843-28734251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEK8
Entrez Gene ID
284086
Gene Name
NIMA related kinase 8
Gene Aliases
JCK, NEK12A, NPHP9, RHPD2
UniGene
Hs.448468
Gene Chromosome Location
Chr.17: 28727899-28743457 on Build GRCh38
Transcripts
NM_178170.2 XM_011524638.2 XM_011524640.2 XM_017024499.1 XM_017024500.1 XM_017024501.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6724546 c.134G>A p.R45Q Chr.17: 28734069-28734069 on Build GRCh38
4065045 c.224C>T p.A75V Chr.17: 28734159-28734159 on Build GRCh38
5192519 c.130G>C p.E44Q Chr.17: 28734065-28734065 on Build GRCh38
3937305 c.201C>T p.Y67Y Chr.17: 28734136-28734136 on Build GRCh38
5923229 c.253+6C>T p.? Chr.17: 28734194-28734194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780350636 Chr.17: 28733969-28733969 on Build GRCh38
rs909308407 Chr.17: 28734112-28734112 on Build GRCh38
rs757098308 Chr.17: 28734061-28734061 on Build GRCh38
rs768501708 Chr.17: 28734088-28734088 on Build GRCh38
rs774737716 Chr.17: 28733977-28733977 on Build GRCh38
rs748430513 Chr.17: 28733992-28733992 on Build GRCh38
rs763887913 Chr.17: 28733937-28733937 on Build GRCh38
rs760073182 Chr.17: 28734154-28734155 on Build GRCh38
rs75328009 Chr.17: 28734002-28734002 on Build GRCh38
rs750671320 Chr.17: 28734069-28734069 on Build GRCh38
rs181429557 Chr.17: 28733875-28733875 on Build GRCh38
rs983686212 Chr.17: 28734207-28734207 on Build GRCh38
rs745461608 Chr.17: 28734174-28734174 on Build GRCh38
rs774144851 Chr.17: 28734199-28734199 on Build GRCh38
rs749351161 Chr.17: 28734116-28734116 on Build GRCh38
rs769058088 Chr.17: 28733973-28733973 on Build GRCh38
rs771147281 Chr.17: 28734118-28734119 on Build GRCh38
rs1011256360 Chr.17: 28733920-28733920 on Build GRCh38
rs942079921 Chr.17: 28734164-28734164 on Build GRCh38
rs1043614621 Chr.17: 28733880-28733880 on Build GRCh38
rs759248580 Chr.17: 28734023-28734023 on Build GRCh38
rs201825819 Chr.17: 28733952-28733952 on Build GRCh38
rs755125104 Chr.17: 28734122-28734122 on Build GRCh38
rs781536247 Chr.17: 28733961-28733961 on Build GRCh38
rs764658381 Chr.17: 28734029-28734029 on Build GRCh38
rs147141279 Chr.17: 28734047-28734047 on Build GRCh38
rs200648580 Chr.17: 28733962-28733962 on Build GRCh38
rs199984152 Chr.17: 28734169-28734169 on Build GRCh38
rs202217310 Chr.17: 28734066-28734066 on Build GRCh38
rs751337514 Chr.17: 28733942-28733942 on Build GRCh38
rs763972758 Chr.17: 28734057-28734057 on Build GRCh38
rs758269662 Chr.17: 28733933-28733933 on Build GRCh38
rs756411828 Chr.17: 28734082-28734082 on Build GRCh38
rs780124748 Chr.17: 28734091-28734091 on Build GRCh38
rs775028329 Chr.17: 28734188-28734188 on Build GRCh38
rs774871392 Chr.17: 28734031-28734031 on Build GRCh38
rs774868062 Chr.17: 28734145-28734145 on Build GRCh38
rs768264404 Chr.17: 28734193-28734193 on Build GRCh38
rs776675685 Chr.17: 28734170-28734170 on Build GRCh38
rs749672981 Chr.17: 28734021-28734022 on Build GRCh38
rs957062166 Chr.17: 28733911-28733911 on Build GRCh38
rs749737984 Chr.17: 28733972-28733972 on Build GRCh38
rs776251322 Chr.17: 28734003-28734003 on Build GRCh38
rs372807519 Chr.17: 28734060-28734060 on Build GRCh38
rs769455568 Chr.17: 28734183-28734183 on Build GRCh38
rs779265033 Chr.17: 28734133-28734133 on Build GRCh38
rs967718734 Chr.17: 28733944-28733944 on Build GRCh38
rs572442707 Chr.17: 28733925-28733925 on Build GRCh38
rs772419251 Chr.17: 28734149-28734149 on Build GRCh38
rs371542903 Chr.17: 28734136-28734136 on Build GRCh38
rs762343001 Chr.17: 28734192-28734192 on Build GRCh38

View Full Product Details