Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44667828-44668325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEIOC
Entrez Gene ID
284071
Gene Name
meiosis specific with coiled-coil domain
Gene Aliases
C17orf104
UniGene
Hs.349758
Gene Chromosome Location
Chr.17: 44656394-44677086 on Build GRCh38
Transcripts
NM_001145080.2 XM_005257236.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5426093 c.2234A>G p.K745R Chr.17: 44668145-44668145 on Build GRCh38
4130146 c.2151T>C p.H717H Chr.17: 44668062-44668062 on Build GRCh38
7400321 c.2010G>C p.M670I Chr.17: 44667921-44667921 on Build GRCh38
436707 c.2157C>G p.Y719* Chr.17: 44668068-44668068 on Build GRCh38
1283741 c.1967G>A p.R656H Chr.17: 44667878-44667878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762035999 Chr.17: 44668060-44668060 on Build GRCh38
rs772318625 Chr.17: 44668158-44668158 on Build GRCh38
rs769200629 Chr.17: 44668244-44668244 on Build GRCh38
rs113617409 Chr.17: 44668087-44668087 on Build GRCh38
rs114536199 Chr.17: 44668298-44668298 on Build GRCh38
rs547945790 Chr.17: 44667938-44667938 on Build GRCh38
rs763940507 Chr.17: 44667878-44667878 on Build GRCh38
rs369272462 Chr.17: 44667931-44667931 on Build GRCh38
rs148051553 Chr.17: 44667886-44667886 on Build GRCh38
rs140839045 Chr.17: 44668151-44668151 on Build GRCh38
rs779651214 Chr.17: 44667892-44667892 on Build GRCh38
rs746206047 Chr.17: 44668147-44668147 on Build GRCh38
rs367563139 Chr.17: 44668267-44668267 on Build GRCh38
rs752932139 Chr.17: 44667876-44667876 on Build GRCh38
rs780030755 Chr.17: 44668103-44668103 on Build GRCh38
rs754496535 Chr.17: 44668210-44668210 on Build GRCh38
rs764196036 Chr.17: 44667949-44667949 on Build GRCh38
rs747791345 Chr.17: 44668018-44668018 on Build GRCh38
rs765303195 Chr.17: 44667940-44667940 on Build GRCh38
rs61745789 Chr.17: 44668045-44668045 on Build GRCh38
rs768630704 Chr.17: 44668155-44668155 on Build GRCh38
rs778431102 Chr.17: 44668100-44668100 on Build GRCh38
rs1002931902 Chr.17: 44667930-44667930 on Build GRCh38
rs758547214 Chr.17: 44667877-44667877 on Build GRCh38
rs781466044 Chr.17: 44668139-44668139 on Build GRCh38
rs750397396 Chr.17: 44667979-44667979 on Build GRCh38
rs747174247 Chr.17: 44668268-44668268 on Build GRCh38
rs146753311 Chr.17: 44667946-44667946 on Build GRCh38
rs571717712 Chr.17: 44667870-44667870 on Build GRCh38
rs755707054 Chr.17: 44667891-44667891 on Build GRCh38
rs372918454 Chr.17: 44668163-44668163 on Build GRCh38
rs376006001 Chr.17: 44667887-44667887 on Build GRCh38
rs373156959 Chr.17: 44667862-44667862 on Build GRCh38
rs866393237 Chr.17: 44668116-44668116 on Build GRCh38
rs985795933 Chr.17: 44668029-44668029 on Build GRCh38
rs751410737 Chr.17: 44667958-44667958 on Build GRCh38
rs770269656 Chr.17: 44668035-44668035 on Build GRCh38
rs770322687 Chr.17: 44668149-44668149 on Build GRCh38
rs774457557 Chr.17: 44668046-44668046 on Build GRCh38
rs748786553 Chr.17: 44667990-44667990 on Build GRCh38
rs754718819 Chr.17: 44668002-44668002 on Build GRCh38
rs765256069 Chr.17: 44667871-44667871 on Build GRCh38
rs760578992 Chr.17: 44668161-44668161 on Build GRCh38
rs779626194 Chr.17: 44668254-44668259 on Build GRCh38
rs1027862272 Chr.17: 44668178-44668178 on Build GRCh38
rs370990129 Chr.17: 44668082-44668082 on Build GRCh38
rs547768132 Chr.17: 44667908-44667908 on Build GRCh38
rs917220983 Chr.17: 44668237-44668237 on Build GRCh38
rs753791741 Chr.17: 44668086-44668086 on Build GRCh38
rs533630811 Chr.17: 44667852-44667852 on Build GRCh38
rs767219883 Chr.17: 44668071-44668071 on Build GRCh38
rs143171146 Chr.17: 44668181-44668181 on Build GRCh38
rs36040376 Chr.17: 44668243-44668244 on Build GRCh38
rs184219664 Chr.17: 44667857-44667857 on Build GRCh38
rs867171473 Chr.17: 44668117-44668117 on Build GRCh38
rs778910581 Chr.17: 44668249-44668249 on Build GRCh38
rs938661314 Chr.17: 44668180-44668180 on Build GRCh38
rs375642697 Chr.17: 44668150-44668150 on Build GRCh38
rs141779273 Chr.17: 44667898-44667898 on Build GRCh38
rs1010010912 Chr.17: 44668047-44668047 on Build GRCh38
rs868028456 Chr.17: 44668211-44668211 on Build GRCh38
rs758032857 Chr.17: 44668196-44668196 on Build GRCh38
rs373486210 Chr.17: 44667980-44667980 on Build GRCh38
rs747308552 Chr.17: 44667917-44667917 on Build GRCh38
rs1029317726 Chr.17: 44668062-44668062 on Build GRCh38
rs764724595 Chr.17: 44668188-44668188 on Build GRCh38
rs201039364 Chr.17: 44668271-44668271 on Build GRCh38
rs777368661 Chr.17: 44668208-44668208 on Build GRCh38
rs773181104 Chr.17: 44668160-44668160 on Build GRCh38
rs746439013 Chr.17: 44668031-44668031 on Build GRCh38
rs776566271 Chr.17: 44668274-44668274 on Build GRCh38
rs147259416 Chr.17: 44668128-44668128 on Build GRCh38
rs756492936 Chr.17: 44668239-44668239 on Build GRCh38
rs749762084 Chr.17: 44668242-44668242 on Build GRCh38
rs368705145 Chr.17: 44668114-44668114 on Build GRCh38
rs143842546 Chr.17: 44668091-44668091 on Build GRCh38
rs751294861 Chr.17: 44667883-44667883 on Build GRCh38
rs1021909557 Chr.17: 44667993-44667993 on Build GRCh38
rs1017298512 Chr.17: 44668290-44668290 on Build GRCh38
rs752234531 Chr.17: 44668194-44668194 on Build GRCh38
rs374249223 Chr.17: 44668083-44668083 on Build GRCh38
rs144776407 Chr.17: 44668204-44668204 on Build GRCh38
rs774421535 Chr.17: 44668157-44668157 on Build GRCh38
rs1044390417 Chr.17: 44668153-44668153 on Build GRCh38
rs78536036 Chr.17: 44668017-44668017 on Build GRCh38
rs748229425 Chr.17: 44668252-44668252 on Build GRCh38
rs771216681 Chr.17: 44668023-44668023 on Build GRCh38
rs771548518 Chr.17: 44668238-44668249 on Build GRCh38
rs554750912 Chr.17: 44668026-44668026 on Build GRCh38
rs767307622 Chr.17: 44667895-44667895 on Build GRCh38
rs376480149 Chr.17: 44668039-44668039 on Build GRCh38
rs767292987 Chr.17: 44667977-44667977 on Build GRCh38
rs772005564 Chr.17: 44668253-44668253 on Build GRCh38
rs777029690 Chr.17: 44667936-44667936 on Build GRCh38
rs775540950 Chr.17: 44667943-44667943 on Build GRCh38
rs766389167 Chr.17: 44668084-44668084 on Build GRCh38
rs773675107 Chr.17: 44667915-44667915 on Build GRCh38
rs779769750 Chr.17: 44667987-44667987 on Build GRCh38
rs751169103 Chr.17: 44668238-44668238 on Build GRCh38
rs907307566 Chr.17: 44667906-44667906 on Build GRCh38
rs753844969 Chr.17: 44668177-44668177 on Build GRCh38

View Full Product Details