Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32684785-32685314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP4
Entrez Gene ID
5928
Gene Name
RB binding protein 4, chromatin remodeling factor
Gene Aliases
NURF55, RBAP48, lin-53
UniGene
Hs.16003
Gene Chromosome Location
Chr.1: 32651148-32686211 on Build GRCh38
Transcripts
NM_001135255.1 NM_001135256.1 NM_005610.2 NM_001161708.1 NM_030786.2

Gene Symbol
SYNC
Entrez Gene ID
81493
Gene Name
syncoilin, intermediate filament protein
Gene Aliases
SYNC1, SYNCOILIN
UniGene
Hs.712631
Gene Chromosome Location
Chr.1: 32679906-32702760 on Build GRCh38
Transcripts
NM_001135255.1 NM_001135256.1 NM_005610.2 NM_001161708.1 NM_030786.2

SNP Detail

SNP ID Chromosome Location
rs866866353 Chr.1: 32684936-32684936 on Build GRCh38
rs34609133 Chr.1: 32685021-32685022 on Build GRCh38
rs551481602 Chr.1: 32684967-32684967 on Build GRCh38
rs1034611649 Chr.1: 32685011-32685011 on Build GRCh38
rs923198413 Chr.1: 32685280-32685280 on Build GRCh38
rs977352501 Chr.1: 32685273-32685273 on Build GRCh38
rs550316395 Chr.1: 32684942-32684942 on Build GRCh38
rs886697727 Chr.1: 32685239-32685239 on Build GRCh38
rs1034558223 Chr.1: 32685237-32685237 on Build GRCh38
rs879213088 Chr.1: 32684999-32684999 on Build GRCh38
rs748944155 Chr.1: 32684939-32684939 on Build GRCh38
rs958886890 Chr.1: 32685253-32685253 on Build GRCh38
rs1025905089 Chr.1: 32685170-32685170 on Build GRCh38
rs143792838 Chr.1: 32684946-32684946 on Build GRCh38
rs752062045 Chr.1: 32685234-32685234 on Build GRCh38
rs923887318 Chr.1: 32685067-32685067 on Build GRCh38
rs398500 Chr.1: 32684997-32684997 on Build GRCh38
rs933271972 Chr.1: 32685283-32685283 on Build GRCh38
rs941813879 Chr.1: 32685128-32685128 on Build GRCh38
rs890547852 Chr.1: 32684946-32684948 on Build GRCh38
rs760869840 Chr.1: 32685161-32685161 on Build GRCh38
rs888825197 Chr.1: 32684955-32684955 on Build GRCh38
rs923165944 Chr.1: 32685115-32685115 on Build GRCh38
rs552726162 Chr.1: 32685011-32685012 on Build GRCh38
rs772565656 Chr.1: 32685096-32685096 on Build GRCh38
rs888939991 Chr.1: 32685120-32685120 on Build GRCh38
rs981569457 Chr.1: 32685040-32685040 on Build GRCh38
rs897497159 Chr.1: 32685153-32685153 on Build GRCh38
rs61248295 Chr.1: 32685023-32685024 on Build GRCh38
rs189994202 Chr.1: 32684954-32684954 on Build GRCh38
rs1016376624 Chr.1: 32684972-32684972 on Build GRCh38
rs977753655 Chr.1: 32685101-32685101 on Build GRCh38
rs907421579 Chr.1: 32685175-32685175 on Build GRCh38
rs879428901 Chr.1: 32684996-32684997 on Build GRCh38
rs181180646 Chr.1: 32684995-32684995 on Build GRCh38
rs979402167 Chr.1: 32684890-32684890 on Build GRCh38
rs916393626 Chr.1: 32684871-32684871 on Build GRCh38
rs914578424 Chr.1: 32685065-32685065 on Build GRCh38
rs764601014 Chr.1: 32685049-32685049 on Build GRCh38
rs956790889 Chr.1: 32685106-32685106 on Build GRCh38
rs139842775 Chr.1: 32685039-32685039 on Build GRCh38
rs979673019 Chr.1: 32685064-32685064 on Build GRCh38
rs1041614994 Chr.1: 32684914-32684914 on Build GRCh38
rs944674311 Chr.1: 32685132-32685132 on Build GRCh38
rs71493586 Chr.1: 32684996-32684997 on Build GRCh38
rs1023328225 Chr.1: 32685044-32685044 on Build GRCh38
rs990618848 Chr.1: 32684860-32684860 on Build GRCh38
rs634787 Chr.1: 32684996-32684996 on Build GRCh38
rs634717 Chr.1: 32685042-32685042 on Build GRCh38
rs933157090 Chr.1: 32685116-32685116 on Build GRCh38
rs897486881 Chr.1: 32684976-32684976 on Build GRCh38
rs602580 Chr.1: 32685117-32685117 on Build GRCh38
rs990251018 Chr.1: 32685052-32685052 on Build GRCh38
rs774808362 Chr.1: 32685001-32685001 on Build GRCh38
rs769342411 Chr.1: 32684856-32684856 on Build GRCh38
rs934719524 Chr.1: 32684901-32684901 on Build GRCh38
rs923837068 Chr.1: 32684896-32684897 on Build GRCh38
rs146794054 Chr.1: 32685015-32685016 on Build GRCh38
rs146454154 Chr.1: 32684982-32684982 on Build GRCh38
rs911866804 Chr.1: 32685118-32685118 on Build GRCh38
rs946648735 Chr.1: 32684881-32684884 on Build GRCh38
rs702832 Chr.1: 32685264-32685264 on Build GRCh38
rs990666030 Chr.1: 32685255-32685255 on Build GRCh38
rs777625168 Chr.1: 32684932-32684932 on Build GRCh38
rs1021681913 Chr.1: 32685256-32685256 on Build GRCh38
rs986007889 Chr.1: 32685291-32685291 on Build GRCh38
rs538790747 Chr.1: 32685134-32685134 on Build GRCh38

View Full Product Details