Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14799915-14800370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR7C1
Entrez Gene ID
26664
Gene Name
olfactory receptor family 7 subfamily C member 1
Gene Aliases
CIT-HSP-146E8, HSTPCR86P, OR19-5, OR7C4, TPCR86
UniGene
Hs.466053
Gene Chromosome Location
Chr.19: 14797550-14835376 on Build GRCh38
Transcripts
NM_198944.1 XM_005259853.3 XM_011527915.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4831085 c.26C>A p.A9D Chr.19: 14800111-14800111 on Build GRCh38
6150308 c.11G>A p.G4E Chr.19: 14800126-14800126 on Build GRCh38
403735 c.222C>T p.T74T Chr.19: 14799915-14799915 on Build GRCh38
1129976 c.111C>G p.V37V Chr.19: 14800026-14800026 on Build GRCh38
7133058 c.206C>A p.A69D Chr.19: 14799931-14799931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76274197 Chr.19: 14800142-14800142 on Build GRCh38
rs574002002 Chr.19: 14800319-14800319 on Build GRCh38
rs778837991 Chr.19: 14800061-14800061 on Build GRCh38
rs1043459058 Chr.19: 14800004-14800004 on Build GRCh38
rs746929414 Chr.19: 14800188-14800188 on Build GRCh38
rs759744235 Chr.19: 14800115-14800115 on Build GRCh38
rs934481752 Chr.19: 14800268-14800268 on Build GRCh38
rs758160110 Chr.19: 14800178-14800178 on Build GRCh38
rs776783210 Chr.19: 14799975-14799975 on Build GRCh38
rs571688519 Chr.19: 14800245-14800245 on Build GRCh38
rs552070339 Chr.19: 14800017-14800017 on Build GRCh38
rs137947822 Chr.19: 14800065-14800065 on Build GRCh38
rs777722744 Chr.19: 14800080-14800080 on Build GRCh38
rs1043430143 Chr.19: 14800302-14800302 on Build GRCh38
rs373552926 Chr.19: 14800057-14800057 on Build GRCh38
rs754690552 Chr.19: 14800169-14800169 on Build GRCh38
rs769950304 Chr.19: 14800117-14800117 on Build GRCh38
rs753265154 Chr.19: 14800002-14800002 on Build GRCh38
rs200940777 Chr.19: 14800016-14800016 on Build GRCh38
rs765984373 Chr.19: 14800156-14800156 on Build GRCh38
rs921730331 Chr.19: 14800281-14800281 on Build GRCh38
rs778639947 Chr.19: 14800170-14800170 on Build GRCh38
rs779902407 Chr.19: 14800032-14800032 on Build GRCh38
rs950895400 Chr.19: 14800035-14800035 on Build GRCh38
rs754886551 Chr.19: 14800051-14800051 on Build GRCh38
rs775724333 Chr.19: 14800128-14800128 on Build GRCh38
rs374301247 Chr.19: 14799997-14799997 on Build GRCh38
rs760144085 Chr.19: 14800149-14800149 on Build GRCh38
rs763386345 Chr.19: 14800015-14800015 on Build GRCh38
rs368030126 Chr.19: 14800143-14800143 on Build GRCh38
rs995549101 Chr.19: 14800106-14800106 on Build GRCh38
rs753510548 Chr.19: 14800036-14800036 on Build GRCh38
rs923986613 Chr.19: 14800250-14800250 on Build GRCh38
rs776874098 Chr.19: 14800109-14800109 on Build GRCh38
rs149561973 Chr.19: 14800088-14800088 on Build GRCh38
rs371593335 Chr.19: 14800162-14800162 on Build GRCh38
rs561926489 Chr.19: 14800303-14800303 on Build GRCh38
rs763340853 Chr.19: 14800129-14800129 on Build GRCh38
rs557246039 Chr.19: 14800187-14800187 on Build GRCh38
rs8108750 Chr.19: 14800288-14800288 on Build GRCh38
rs145774543 Chr.19: 14799940-14799940 on Build GRCh38
rs901494006 Chr.19: 14799981-14799981 on Build GRCh38
rs771907297 Chr.19: 14800077-14800077 on Build GRCh38
rs775745262 Chr.19: 14800093-14800095 on Build GRCh38
rs903508113 Chr.19: 14800005-14800005 on Build GRCh38
rs777618006 Chr.19: 14800185-14800185 on Build GRCh38
rs773177105 Chr.19: 14799943-14799943 on Build GRCh38
rs934975152 Chr.19: 14800337-14800337 on Build GRCh38
rs117130085 Chr.19: 14799957-14799957 on Build GRCh38
rs573324632 Chr.19: 14800150-14800150 on Build GRCh38
rs949389599 Chr.19: 14800155-14800155 on Build GRCh38
rs766615730 Chr.19: 14799959-14799959 on Build GRCh38
rs892961363 Chr.19: 14800318-14800318 on Build GRCh38
rs772251459 Chr.19: 14799941-14799941 on Build GRCh38
rs770970993 Chr.19: 14800107-14800107 on Build GRCh38
rs925064551 Chr.19: 14800332-14800332 on Build GRCh38
rs138417629 Chr.19: 14800149-14800150 on Build GRCh38
rs535439693 Chr.19: 14799994-14799994 on Build GRCh38
rs755603828 Chr.19: 14800020-14800020 on Build GRCh38

View Full Product Details