Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14827980-14828482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR7A5
Entrez Gene ID
26659
Gene Name
olfactory receptor family 7 subfamily A member 5
Gene Aliases
HTPCR2
UniGene
Hs.137573
Gene Chromosome Location
Chr.19: 14825743-14835197 on Build GRCh38
Transcripts
NM_017506.1 XM_011527912.2 XM_011527913.2 XM_017026583.1 XM_005259853.3 XM_011527915.2

Gene Symbol
OR7C1
Entrez Gene ID
26664
Gene Name
olfactory receptor family 7 subfamily C member 1
Gene Aliases
CIT-HSP-146E8, HSTPCR86P, OR19-5, OR7C4, TPCR86
UniGene
Hs.466053
Gene Chromosome Location
Chr.19: 14797550-14835376 on Build GRCh38
Transcripts
NM_017506.1 XM_011527912.2 XM_011527913.2 XM_017026583.1 XM_005259853.3 XM_011527915.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4978904 c.31G>C p.E11Q Chr.19: 14828211-14828211 on Build GRCh38
1564654 c.224C>G p.S75C Chr.19: 14828018-14828018 on Build GRCh38
5388829 c.256C>T p.Q86* Chr.19: 14827986-14827986 on Build GRCh38
6896242 c.79C>T p.L27F Chr.19: 14828163-14828163 on Build GRCh38
5388830 c.165C>T p.L55L Chr.19: 14828077-14828077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115828008 Chr.19: 14828271-14828271 on Build GRCh38
rs771578284 Chr.19: 14828448-14828448 on Build GRCh38
rs771170959 Chr.19: 14828224-14828224 on Build GRCh38
rs781463480 Chr.19: 14828292-14828292 on Build GRCh38
rs768508692 Chr.19: 14828077-14828077 on Build GRCh38
rs201864574 Chr.19: 14828159-14828159 on Build GRCh38
rs749025863 Chr.19: 14828076-14828076 on Build GRCh38
rs756644152 Chr.19: 14828067-14828068 on Build GRCh38
rs775922417 Chr.19: 14828256-14828256 on Build GRCh38
rs756772915 Chr.19: 14828047-14828047 on Build GRCh38
rs746661340 Chr.19: 14828259-14828260 on Build GRCh38
rs141415832 Chr.19: 14828121-14828121 on Build GRCh38
rs757894280 Chr.19: 14828198-14828200 on Build GRCh38
rs370546644 Chr.19: 14828087-14828087 on Build GRCh38
rs765683311 Chr.19: 14828140-14828140 on Build GRCh38
rs754396526 Chr.19: 14828021-14828021 on Build GRCh38
rs752197178 Chr.19: 14828105-14828105 on Build GRCh38
rs866796294 Chr.19: 14828056-14828056 on Build GRCh38
rs571545351 Chr.19: 14828273-14828273 on Build GRCh38
rs922817074 Chr.19: 14828368-14828368 on Build GRCh38
rs147832964 Chr.19: 14828101-14828101 on Build GRCh38
rs1054326728 Chr.19: 14828093-14828093 on Build GRCh38
rs761066740 Chr.19: 14828015-14828015 on Build GRCh38
rs930010818 Chr.19: 14828411-14828411 on Build GRCh38
rs201380372 Chr.19: 14828285-14828285 on Build GRCh38
rs367579319 Chr.19: 14828157-14828157 on Build GRCh38
rs56328009 Chr.19: 14828427-14828427 on Build GRCh38
rs779119376 Chr.19: 14828380-14828380 on Build GRCh38
rs951017608 Chr.19: 14828181-14828181 on Build GRCh38
rs970794831 Chr.19: 14828045-14828045 on Build GRCh38
rs758464738 Chr.19: 14828212-14828212 on Build GRCh38
rs778755183 Chr.19: 14828196-14828196 on Build GRCh38
rs749781903 Chr.19: 14828290-14828290 on Build GRCh38
rs765067170 Chr.19: 14828284-14828284 on Build GRCh38
rs551120689 Chr.19: 14828362-14828362 on Build GRCh38
rs767879125 Chr.19: 14828024-14828025 on Build GRCh38
rs934617521 Chr.19: 14828356-14828356 on Build GRCh38
rs764628503 Chr.19: 14828263-14828263 on Build GRCh38
rs61731995 Chr.19: 14828048-14828048 on Build GRCh38
rs769062612 Chr.19: 14828112-14828112 on Build GRCh38
rs772880095 Chr.19: 14828265-14828265 on Build GRCh38
rs979213779 Chr.19: 14828298-14828298 on Build GRCh38
rs117611282 Chr.19: 14828434-14828434 on Build GRCh38
rs145962695 Chr.19: 14828122-14828122 on Build GRCh38
rs761300408 Chr.19: 14828142-14828142 on Build GRCh38
rs746261646 Chr.19: 14828244-14828244 on Build GRCh38
rs760046522 Chr.19: 14828024-14828024 on Build GRCh38
rs776783168 Chr.19: 14828225-14828225 on Build GRCh38
rs200480688 Chr.19: 14828035-14828035 on Build GRCh38
rs995698908 Chr.19: 14828161-14828161 on Build GRCh38
rs751059842 Chr.19: 14828041-14828041 on Build GRCh38
rs753219648 Chr.19: 14828141-14828141 on Build GRCh38
rs756036560 Chr.19: 14828067-14828067 on Build GRCh38
rs547840987 Chr.19: 14828145-14828145 on Build GRCh38
rs183738487 Chr.19: 14828066-14828066 on Build GRCh38
rs780483170 Chr.19: 14828286-14828286 on Build GRCh38
rs989184862 Chr.19: 14828295-14828295 on Build GRCh38
rs763533296 Chr.19: 14828261-14828261 on Build GRCh38
rs771079206 Chr.19: 14828118-14828118 on Build GRCh38
rs374740501 Chr.19: 14828027-14828028 on Build GRCh38
rs777840010 Chr.19: 14828218-14828218 on Build GRCh38
rs765705586 Chr.19: 14828036-14828036 on Build GRCh38
rs118148336 Chr.19: 14828293-14828293 on Build GRCh38
rs936874666 Chr.19: 14828072-14828072 on Build GRCh38
rs199898657 Chr.19: 14828174-14828174 on Build GRCh38
rs372354094 Chr.19: 14828315-14828315 on Build GRCh38
rs760731104 Chr.19: 14828180-14828180 on Build GRCh38
rs748110871 Chr.19: 14828197-14828197 on Build GRCh38
rs576383780 Chr.19: 14828452-14828452 on Build GRCh38
rs754778616 Chr.19: 14828187-14828187 on Build GRCh38
rs747021255 Chr.19: 14828220-14828220 on Build GRCh38
rs780489117 Chr.19: 14828095-14828096 on Build GRCh38
rs752560887 Chr.19: 14828127-14828127 on Build GRCh38
rs376664962 Chr.19: 14828387-14828387 on Build GRCh38
rs766725859 Chr.19: 14828016-14828016 on Build GRCh38
rs747111149 Chr.19: 14828291-14828291 on Build GRCh38
rs138615141 Chr.19: 14828146-14828146 on Build GRCh38
rs770193111 Chr.19: 14828246-14828246 on Build GRCh38
rs749898869 Chr.19: 14828156-14828156 on Build GRCh38
rs773489445 Chr.19: 14828107-14828107 on Build GRCh38
rs750716925 Chr.19: 14828066-14828067 on Build GRCh38
rs942854488 Chr.19: 14828351-14828351 on Build GRCh38
rs773460339 Chr.19: 14828010-14828010 on Build GRCh38
rs760872641 Chr.19: 14828113-14828113 on Build GRCh38
rs780008849 Chr.19: 14828070-14828070 on Build GRCh38
rs777356593 Chr.19: 14828260-14828261 on Build GRCh38
rs368950783 Chr.19: 14828268-14828268 on Build GRCh38

View Full Product Details