Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15914512-15915024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F11
Entrez Gene ID
57834
Gene Name
cytochrome P450 family 4 subfamily F member 11
Gene Aliases
CYPIVF11
UniGene
Hs.187393
Gene Chromosome Location
Chr.19: 15912370-15934866 on Build GRCh38
Transcripts
NM_001128932.1 NM_021187.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3284293 c.1198C>T p.R400* Chr.19: 15914813-15914813 on Build GRCh38
6639360 c.1246A>G p.K416E Chr.19: 15914765-15914765 on Build GRCh38
4676010 c.1235G>A p.R412H Chr.19: 15914776-15914776 on Build GRCh38
4568671 c.1272T>G p.I424M Chr.19: 15914644-15914644 on Build GRCh38
5614613 c.1214A>G p.D405G Chr.19: 15914797-15914797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773559903 Chr.19: 15914884-15914884 on Build GRCh38
rs763829157 Chr.19: 15914897-15914897 on Build GRCh38
rs749399448 Chr.19: 15914788-15914788 on Build GRCh38
rs748057327 Chr.19: 15914628-15914628 on Build GRCh38
rs762240482 Chr.19: 15914805-15914805 on Build GRCh38
rs150118221 Chr.19: 15914699-15914699 on Build GRCh38
rs201779761 Chr.19: 15914552-15914552 on Build GRCh38
rs768672776 Chr.19: 15914797-15914797 on Build GRCh38
rs140718122 Chr.19: 15914775-15914775 on Build GRCh38
rs919545176 Chr.19: 15914990-15914990 on Build GRCh38
rs45574935 Chr.19: 15914841-15914841 on Build GRCh38
rs765697900 Chr.19: 15914575-15914575 on Build GRCh38
rs761126212 Chr.19: 15914891-15914891 on Build GRCh38
rs775698990 Chr.19: 15914742-15914742 on Build GRCh38
rs767864124 Chr.19: 15914754-15914754 on Build GRCh38
rs567758136 Chr.19: 15914872-15914872 on Build GRCh38
rs776736711 Chr.19: 15914713-15914713 on Build GRCh38
rs558822665 Chr.19: 15914959-15914959 on Build GRCh38
rs776875447 Chr.19: 15914647-15914647 on Build GRCh38
rs765306476 Chr.19: 15914655-15914655 on Build GRCh38
rs778045739 Chr.19: 15914586-15914586 on Build GRCh38
rs200031770 Chr.19: 15914629-15914629 on Build GRCh38
rs755824248 Chr.19: 15914608-15914608 on Build GRCh38
rs764780177 Chr.19: 15914774-15914774 on Build GRCh38
rs759559832 Chr.19: 15914648-15914648 on Build GRCh38
rs150098509 Chr.19: 15914781-15914781 on Build GRCh38
rs746834736 Chr.19: 15914701-15914701 on Build GRCh38
rs550369934 Chr.19: 15914906-15914906 on Build GRCh38
rs748298676 Chr.19: 15914947-15914947 on Build GRCh38
rs779656818 Chr.19: 15914612-15914612 on Build GRCh38
rs779083973 Chr.19: 15914943-15914943 on Build GRCh38
rs543811544 Chr.19: 15914907-15914912 on Build GRCh38
rs370263065 Chr.19: 15914681-15914681 on Build GRCh38
rs961358132 Chr.19: 15914752-15914752 on Build GRCh38
rs777895272 Chr.19: 15914910-15914910 on Build GRCh38
rs778738847 Chr.19: 15914625-15914625 on Build GRCh38
rs752351878 Chr.19: 15914825-15914825 on Build GRCh38
rs753481104 Chr.19: 15914682-15914682 on Build GRCh38
rs754723377 Chr.19: 15914765-15914765 on Build GRCh38
rs745638773 Chr.19: 15914782-15914782 on Build GRCh38
rs141887211 Chr.19: 15914824-15914824 on Build GRCh38
rs754470602 Chr.19: 15914623-15914623 on Build GRCh38
rs777878547 Chr.19: 15914776-15914776 on Build GRCh38
rs760483929 Chr.19: 15914952-15914952 on Build GRCh38
rs565985827 Chr.19: 15914969-15914969 on Build GRCh38
rs746785523 Chr.19: 15914641-15914641 on Build GRCh38
rs755947846 Chr.19: 15914917-15914917 on Build GRCh38
rs375685652 Chr.19: 15914680-15914680 on Build GRCh38
rs146262579 Chr.19: 15914892-15914892 on Build GRCh38
rs753851013 Chr.19: 15914901-15914901 on Build GRCh38
rs753203359 Chr.19: 15914580-15914580 on Build GRCh38
rs779030910 Chr.19: 15914871-15914871 on Build GRCh38
rs780110853 Chr.19: 15914787-15914787 on Build GRCh38
rs201533288 Chr.19: 15914700-15914700 on Build GRCh38
rs768723718 Chr.19: 15914864-15914864 on Build GRCh38
rs138369700 Chr.19: 15914823-15914823 on Build GRCh38
rs753621794 Chr.19: 15914923-15914923 on Build GRCh38
rs988550925 Chr.19: 15914802-15914802 on Build GRCh38
rs948989304 Chr.19: 15914548-15914548 on Build GRCh38
rs764572598 Chr.19: 15914671-15914671 on Build GRCh38
rs1042990771 Chr.19: 15914660-15914660 on Build GRCh38
rs754913399 Chr.19: 15914907-15914907 on Build GRCh38
rs763519759 Chr.19: 15914664-15914664 on Build GRCh38
rs771321704 Chr.19: 15914642-15914642 on Build GRCh38
rs372437243 Chr.19: 15914813-15914813 on Build GRCh38
rs928802393 Chr.19: 15914778-15914778 on Build GRCh38
rs766219521 Chr.19: 15914699-15914699 on Build GRCh38
rs770105142 Chr.19: 15914786-15914786 on Build GRCh38
rs375164936 Chr.19: 15914905-15914905 on Build GRCh38
rs564152748 Chr.19: 15914968-15914968 on Build GRCh38
rs764623660 Chr.19: 15914745-15914745 on Build GRCh38
rs200976601 Chr.19: 15914918-15914918 on Build GRCh38
rs145791711 Chr.19: 15914879-15914879 on Build GRCh38
rs570135539 Chr.19: 15914912-15914912 on Build GRCh38
rs920229362 Chr.19: 15914546-15914546 on Build GRCh38
rs200588981 Chr.19: 15914812-15914812 on Build GRCh38
rs757584371 Chr.19: 15914673-15914673 on Build GRCh38
rs755250510 Chr.19: 15914940-15914940 on Build GRCh38
rs374763718 Chr.19: 15914843-15914843 on Build GRCh38
rs758775451 Chr.19: 15914584-15914584 on Build GRCh38
rs891456333 Chr.19: 15914635-15914635 on Build GRCh38
rs751917884 Chr.19: 15914606-15914606 on Build GRCh38
rs753433594 Chr.19: 15914764-15914764 on Build GRCh38
rs545306458 Chr.19: 15914750-15914750 on Build GRCh38
rs767524132 Chr.19: 15914911-15914911 on Build GRCh38
rs774851894 Chr.19: 15914803-15914803 on Build GRCh38
rs762570221 Chr.19: 15914828-15914828 on Build GRCh38
rs73928264 Chr.19: 15914704-15914704 on Build GRCh38
rs763634517 Chr.19: 15914830-15914830 on Build GRCh38
rs893644186 Chr.19: 15914758-15914758 on Build GRCh38
rs773721428 Chr.19: 15914568-15914568 on Build GRCh38
rs147994994 Chr.19: 15914659-15914659 on Build GRCh38
rs755823193 Chr.19: 15914844-15914844 on Build GRCh38
rs751168274 Chr.19: 15914908-15914908 on Build GRCh38
rs748788167 Chr.19: 15914617-15914617 on Build GRCh38
rs192392601 Chr.19: 15914970-15914970 on Build GRCh38
rs749450646 Chr.19: 15914862-15914862 on Build GRCh38
rs771874726 Chr.19: 15914718-15914718 on Build GRCh38
rs761200530 Chr.19: 15914909-15914909 on Build GRCh38
rs763000209 Chr.19: 15914744-15914744 on Build GRCh38
rs751487605 Chr.19: 15914777-15914777 on Build GRCh38
rs746035586 Chr.19: 15914736-15914736 on Build GRCh38
rs1004713453 Chr.19: 15914920-15914920 on Build GRCh38
rs928811297 Chr.19: 15914573-15914573 on Build GRCh38
rs201684723 Chr.19: 15914842-15914842 on Build GRCh38
rs780274514 Chr.19: 15914849-15914849 on Build GRCh38
rs553835355 Chr.19: 15914551-15914551 on Build GRCh38

View Full Product Details