Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19055175-19055682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC6
Entrez Gene ID
93436
Gene Name
armadillo repeat containing 6
Gene Aliases
R30923_1
UniGene
Hs.77876
Gene Chromosome Location
Chr.19: 19033578-19058178 on Build GRCh38
Transcripts
NM_001199196.1 NM_033415.3 XM_005260157.3 XM_017027484.1 XM_017027485.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7263938 c.1100C>T p.A367V Chr.19: 19055341-19055341 on Build GRCh38
4663405 c.1027G>A p.G343S Chr.19: 19055343-19055343 on Build GRCh38
3530951 c.1020C>T p.V340V Chr.19: 19055336-19055336 on Build GRCh38
4075840 c.1061T>C p.I354T Chr.19: 19055302-19055302 on Build GRCh38
3100765 c.1062C>T p.I354I Chr.19: 19055303-19055303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758824206 Chr.19: 19055210-19055210 on Build GRCh38
rs746571727 Chr.19: 19055332-19055332 on Build GRCh38
rs757501938 Chr.19: 19055421-19055421 on Build GRCh38
rs544723649 Chr.19: 19055617-19055617 on Build GRCh38
rs572000202 Chr.19: 19055466-19055466 on Build GRCh38
rs764248400 Chr.19: 19055226-19055226 on Build GRCh38
rs752560255 Chr.19: 19055401-19055401 on Build GRCh38
rs746300385 Chr.19: 19055279-19055279 on Build GRCh38
rs745753966 Chr.19: 19055550-19055550 on Build GRCh38
rs923232356 Chr.19: 19055625-19055625 on Build GRCh38
rs567338730 Chr.19: 19055488-19055488 on Build GRCh38
rs750261442 Chr.19: 19055227-19055227 on Build GRCh38
rs768973848 Chr.19: 19055351-19055351 on Build GRCh38
rs758212812 Chr.19: 19055398-19055398 on Build GRCh38
rs202236249 Chr.19: 19055318-19055318 on Build GRCh38
rs756109914 Chr.19: 19055233-19055233 on Build GRCh38
rs774514859 Chr.19: 19055303-19055303 on Build GRCh38
rs776833065 Chr.19: 19055277-19055277 on Build GRCh38
rs763875700 Chr.19: 19055408-19055408 on Build GRCh38
rs751022551 Chr.19: 19055413-19055413 on Build GRCh38
rs200075145 Chr.19: 19055270-19055270 on Build GRCh38
rs183090986 Chr.19: 19055258-19055258 on Build GRCh38
rs774262935 Chr.19: 19055358-19055358 on Build GRCh38
rs771497905 Chr.19: 19055407-19055408 on Build GRCh38
rs770961820 Chr.19: 19055287-19055287 on Build GRCh38
rs747461803 Chr.19: 19055219-19055219 on Build GRCh38
rs763168211 Chr.19: 19055295-19055295 on Build GRCh38
rs764041903 Chr.19: 19055299-19055299 on Build GRCh38
rs1033353390 Chr.19: 19055201-19055201 on Build GRCh38
rs753679312 Chr.19: 19055311-19055311 on Build GRCh38
rs748728092 Chr.19: 19055521-19055521 on Build GRCh38
rs910606376 Chr.19: 19055612-19055612 on Build GRCh38
rs887905742 Chr.19: 19055427-19055427 on Build GRCh38
rs373374526 Chr.19: 19055310-19055310 on Build GRCh38
rs376608814 Chr.19: 19055312-19055312 on Build GRCh38
rs549061274 Chr.19: 19055316-19055316 on Build GRCh38
rs777095197 Chr.19: 19055326-19055326 on Build GRCh38
rs554420215 Chr.19: 19055480-19055480 on Build GRCh38
rs760509895 Chr.19: 19055308-19055308 on Build GRCh38
rs952831260 Chr.19: 19055291-19055291 on Build GRCh38
rs569700796 Chr.19: 19055206-19055206 on Build GRCh38
rs776652102 Chr.19: 19055368-19055368 on Build GRCh38
rs199554736 Chr.19: 19055292-19055292 on Build GRCh38
rs905451169 Chr.19: 19055492-19055492 on Build GRCh38
rs763081987 Chr.19: 19055221-19055221 on Build GRCh38
rs757919981 Chr.19: 19055266-19055266 on Build GRCh38
rs769995101 Chr.19: 19055288-19055288 on Build GRCh38
rs1032140460 Chr.19: 19055600-19055600 on Build GRCh38
rs749464824 Chr.19: 19055350-19055350 on Build GRCh38
rs966592591 Chr.19: 19055509-19055509 on Build GRCh38
rs200448665 Chr.19: 19055343-19055343 on Build GRCh38
rs372718033 Chr.19: 19055246-19055246 on Build GRCh38
rs1015647200 Chr.19: 19055624-19055624 on Build GRCh38
rs185876668 Chr.19: 19055607-19055607 on Build GRCh38
rs766041005 Chr.19: 19055237-19055237 on Build GRCh38
rs753734141 Chr.19: 19055245-19055245 on Build GRCh38
rs537896032 Chr.19: 19055370-19055370 on Build GRCh38
rs770760571 Chr.19: 19055367-19055367 on Build GRCh38
rs1024930769 Chr.19: 19055494-19055494 on Build GRCh38
rs373323978 Chr.19: 19055260-19055260 on Build GRCh38
rs1033818202 Chr.19: 19055463-19055463 on Build GRCh38
rs3746260 Chr.19: 19055514-19055514 on Build GRCh38
rs28380085 Chr.19: 19055467-19055467 on Build GRCh38
rs371340322 Chr.19: 19055407-19055407 on Build GRCh38
rs976480221 Chr.19: 19055618-19055618 on Build GRCh38
rs267605368 Chr.19: 19055336-19055336 on Build GRCh38
rs201039909 Chr.19: 19055313-19055313 on Build GRCh38
rs376266044 Chr.19: 19055268-19055268 on Build GRCh38
rs374945143 Chr.19: 19055338-19055338 on Build GRCh38
rs374698571 Chr.19: 19055255-19055255 on Build GRCh38
rs957846012 Chr.19: 19055601-19055601 on Build GRCh38
rs975820106 Chr.19: 19055649-19055649 on Build GRCh38
rs917814559 Chr.19: 19055248-19055248 on Build GRCh38
rs555845353 Chr.19: 19055520-19055520 on Build GRCh38
rs1013651788 Chr.19: 19055477-19055477 on Build GRCh38
rs375920271 Chr.19: 19055428-19055428 on Build GRCh38
rs565072017 Chr.19: 19055197-19055197 on Build GRCh38
rs775864031 Chr.19: 19055347-19055347 on Build GRCh38
rs765081788 Chr.19: 19055388-19055388 on Build GRCh38
rs775547313 Chr.19: 19055216-19055216 on Build GRCh38
rs139303339 Chr.19: 19055361-19055361 on Build GRCh38
rs934437839 Chr.19: 19055440-19055440 on Build GRCh38

View Full Product Details