Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50000652-50001053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VRK3
Entrez Gene ID
51231
Gene Name
vaccinia related kinase 3
Gene Aliases
-
UniGene
Hs.443330
Gene Chromosome Location
Chr.19: 49976467-50025954 on Build GRCh38
Transcripts
NM_001025778.1 NM_001308420.1 NM_016440.3 XM_005258971.3 XM_005258972.3 XM_006723237.3 XM_006723238.3 XM_011527023.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4883216 c.580G>C p.G194R Chr.19: 50000822-50000822 on Build GRCh38
4500303 c.561C>T p.S187S Chr.19: 50000841-50000841 on Build GRCh38
6085735 c.596A>T p.K199M Chr.19: 50000806-50000806 on Build GRCh38
6465880 c.592C>T p.Q198* Chr.19: 50000810-50000810 on Build GRCh38
4454820 c.587A>G p.Q196R Chr.19: 50000815-50000815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781151533 Chr.19: 50000755-50000755 on Build GRCh38
rs890634315 Chr.19: 50001018-50001018 on Build GRCh38
rs777230325 Chr.19: 50000787-50000787 on Build GRCh38
rs917094588 Chr.19: 50000945-50000945 on Build GRCh38
rs11882175 Chr.19: 50000715-50000715 on Build GRCh38
rs559500344 Chr.19: 50000873-50000873 on Build GRCh38
rs189775064 Chr.19: 50000895-50000895 on Build GRCh38
rs561783871 Chr.19: 50000834-50000834 on Build GRCh38
rs949854338 Chr.19: 50000953-50000953 on Build GRCh38
rs748901833 Chr.19: 50000773-50000773 on Build GRCh38
rs745557564 Chr.19: 50000887-50000887 on Build GRCh38
rs756928640 Chr.19: 50000882-50000882 on Build GRCh38
rs1019468034 Chr.19: 50000948-50000948 on Build GRCh38
rs765098650 Chr.19: 50000842-50000842 on Build GRCh38
rs754943055 Chr.19: 50000747-50000747 on Build GRCh38
rs549122514 Chr.19: 50000933-50000933 on Build GRCh38
rs765395741 Chr.19: 50000864-50000867 on Build GRCh38
rs949989706 Chr.19: 50000688-50000688 on Build GRCh38
rs925967623 Chr.19: 50000915-50000915 on Build GRCh38
rs777292976 Chr.19: 50000771-50000771 on Build GRCh38
rs759789067 Chr.19: 50000904-50000904 on Build GRCh38
rs1045895341 Chr.19: 50000753-50000753 on Build GRCh38
rs990885048 Chr.19: 50000700-50000700 on Build GRCh38
rs949124089 Chr.19: 50000716-50000716 on Build GRCh38
rs142337369 Chr.19: 50000972-50000972 on Build GRCh38
rs972976144 Chr.19: 50000973-50000973 on Build GRCh38
rs543372025 Chr.19: 50001020-50001020 on Build GRCh38
rs1005593493 Chr.19: 50000735-50000735 on Build GRCh38
rs576270854 Chr.19: 50001009-50001009 on Build GRCh38
rs530045342 Chr.19: 50000954-50000954 on Build GRCh38
rs574459039 Chr.19: 50000989-50000989 on Build GRCh38
rs774383667 Chr.19: 50000898-50000898 on Build GRCh38
rs147702874 Chr.19: 50000892-50000892 on Build GRCh38
rs180780678 Chr.19: 50000990-50000990 on Build GRCh38
rs546061082 Chr.19: 50000690-50000690 on Build GRCh38
rs561369492 Chr.19: 50001027-50001027 on Build GRCh38
rs553437461 Chr.19: 50000686-50000686 on Build GRCh38
rs1038039457 Chr.19: 50000752-50000752 on Build GRCh38
rs537411498 Chr.19: 50000932-50000932 on Build GRCh38
rs368296208 Chr.19: 50000756-50000756 on Build GRCh38
rs535097864 Chr.19: 50000967-50000967 on Build GRCh38
rs775909481 Chr.19: 50000781-50000781 on Build GRCh38
rs557987619 Chr.19: 50000996-50000996 on Build GRCh38
rs375590209 Chr.19: 50000894-50000894 on Build GRCh38
rs766579461 Chr.19: 50000835-50000835 on Build GRCh38
rs867781110 Chr.19: 50000707-50000707 on Build GRCh38
rs776323496 Chr.19: 50000780-50000781 on Build GRCh38
rs575987649 Chr.19: 50000782-50000782 on Build GRCh38
rs761539712 Chr.19: 50000830-50000830 on Build GRCh38
rs567732094 Chr.19: 50000950-50000950 on Build GRCh38
rs756047512 Chr.19: 50000870-50000870 on Build GRCh38
rs376272902 Chr.19: 50000785-50000785 on Build GRCh38
rs2115507 Chr.19: 50000993-50000993 on Build GRCh38
rs996582976 Chr.19: 50000765-50000766 on Build GRCh38
rs773181188 Chr.19: 50000825-50000825 on Build GRCh38
rs759311613 Chr.19: 50000858-50000859 on Build GRCh38
rs11879616 Chr.19: 50000799-50000799 on Build GRCh38
rs755568807 Chr.19: 50000769-50000769 on Build GRCh38
rs770501285 Chr.19: 50000780-50000780 on Build GRCh38
rs571744830 Chr.19: 50000881-50000881 on Build GRCh38
rs752588372 Chr.19: 50000859-50000859 on Build GRCh38
rs113636742 Chr.19: 50000951-50000951 on Build GRCh38
rs367877992 Chr.19: 50000896-50000896 on Build GRCh38
rs11879620 Chr.19: 50000840-50000840 on Build GRCh38
rs767318604 Chr.19: 50000847-50000847 on Build GRCh38
rs1009028507 Chr.19: 50001028-50001028 on Build GRCh38
rs778469388 Chr.19: 50000884-50000884 on Build GRCh38
rs368230198 Chr.19: 50000794-50000794 on Build GRCh38

View Full Product Details