Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38921289-38921783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF33
Entrez Gene ID
100271715
Gene Name
Rho guanine nucleotide exchange factor 33
Gene Aliases
-
UniGene
Hs.99841
Gene Chromosome Location
Chr.2: 38919363-38975449 on Build GRCh38
Transcripts
NM_001145451.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1738712 c.44C>T p.P15L Chr.2: 38921392-38921392 on Build GRCh38
3184029 c.26-8C>T p.? Chr.2: 38921366-38921366 on Build GRCh38
7400195 c.26-7C>G p.? Chr.2: 38921367-38921367 on Build GRCh38
3743799 c.45G>A p.P15P Chr.2: 38921393-38921393 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1052620999 Chr.2: 38921605-38921605 on Build GRCh38
rs950630151 Chr.2: 38921323-38921323 on Build GRCh38
rs565987833 Chr.2: 38921571-38921571 on Build GRCh38
rs900260400 Chr.2: 38921753-38921753 on Build GRCh38
rs1038679672 Chr.2: 38921677-38921677 on Build GRCh38
rs182260142 Chr.2: 38921495-38921495 on Build GRCh38
rs186568961 Chr.2: 38921709-38921709 on Build GRCh38
rs373601820 Chr.2: 38921415-38921415 on Build GRCh38
rs960488224 Chr.2: 38921747-38921747 on Build GRCh38
rs550378225 Chr.2: 38921487-38921487 on Build GRCh38
rs747579614 Chr.2: 38921581-38921581 on Build GRCh38
rs547683401 Chr.2: 38921530-38921530 on Build GRCh38
rs974159058 Chr.2: 38921386-38921386 on Build GRCh38
rs991949788 Chr.2: 38921749-38921749 on Build GRCh38
rs959417075 Chr.2: 38921612-38921612 on Build GRCh38
rs1016584355 Chr.2: 38921579-38921579 on Build GRCh38
rs901578731 Chr.2: 38921567-38921567 on Build GRCh38
rs996244041 Chr.2: 38921450-38921450 on Build GRCh38
rs769081597 Chr.2: 38921671-38921671 on Build GRCh38
rs941352739 Chr.2: 38921377-38921377 on Build GRCh38
rs891269452 Chr.2: 38921612-38921613 on Build GRCh38
rs1056520442 Chr.2: 38921414-38921414 on Build GRCh38
rs368062968 Chr.2: 38921431-38921431 on Build GRCh38
rs796752615 Chr.2: 38921410-38921410 on Build GRCh38
rs926694578 Chr.2: 38921391-38921391 on Build GRCh38
rs77828990 Chr.2: 38921745-38921745 on Build GRCh38
rs926714438 Chr.2: 38921604-38921604 on Build GRCh38
rs527682624 Chr.2: 38921393-38921393 on Build GRCh38
rs916582836 Chr.2: 38921541-38921541 on Build GRCh38
rs754467878 Chr.2: 38921366-38921366 on Build GRCh38
rs536723483 Chr.2: 38921692-38921692 on Build GRCh38
rs1000514432 Chr.2: 38921468-38921468 on Build GRCh38
rs945519300 Chr.2: 38921630-38921630 on Build GRCh38
rs891405765 Chr.2: 38921411-38921411 on Build GRCh38
rs147942922 Chr.2: 38921429-38921429 on Build GRCh38
rs371538488 Chr.2: 38921469-38921469 on Build GRCh38
rs918132968 Chr.2: 38921628-38921628 on Build GRCh38
rs960626425 Chr.2: 38921516-38921516 on Build GRCh38
rs576111221 Chr.2: 38921684-38921684 on Build GRCh38
rs1014719555 Chr.2: 38921733-38921733 on Build GRCh38
rs201910615 Chr.2: 38921409-38921409 on Build GRCh38
rs1004561355 Chr.2: 38921715-38921715 on Build GRCh38
rs370644353 Chr.2: 38921510-38921510 on Build GRCh38
rs950604180 Chr.2: 38921542-38921542 on Build GRCh38
rs749770949 Chr.2: 38921403-38921403 on Build GRCh38
rs1014688468 Chr.2: 38921491-38921491 on Build GRCh38
rs1030676864 Chr.2: 38921315-38921315 on Build GRCh38
rs903546508 Chr.2: 38921445-38921445 on Build GRCh38
rs908581380 Chr.2: 38921372-38921372 on Build GRCh38
rs1042673640 Chr.2: 38921655-38921655 on Build GRCh38
rs925498310 Chr.2: 38921661-38921661 on Build GRCh38
rs1004838120 Chr.2: 38921576-38921576 on Build GRCh38
rs948118517 Chr.2: 38921580-38921580 on Build GRCh38
rs760592816 Chr.2: 38921577-38921577 on Build GRCh38
rs140179942 Chr.2: 38921449-38921449 on Build GRCh38
rs900224485 Chr.2: 38921479-38921479 on Build GRCh38
rs999730310 Chr.2: 38921396-38921396 on Build GRCh38
rs539058206 Chr.2: 38921593-38921593 on Build GRCh38
rs145167412 Chr.2: 38921624-38921624 on Build GRCh38
rs145698222 Chr.2: 38921496-38921496 on Build GRCh38
rs1027716695 Chr.2: 38921668-38921668 on Build GRCh38
rs771247449 Chr.2: 38921408-38921408 on Build GRCh38
rs560510193 Chr.2: 38921392-38921392 on Build GRCh38
rs555568706 Chr.2: 38921700-38921700 on Build GRCh38

View Full Product Details