Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 173909094-173909593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SP3
Entrez Gene ID
6670
Gene Name
Sp3 transcription factor
Gene Aliases
SPR2
UniGene
Hs.531587
Gene Chromosome Location
Chr.2: 173906459-173965702 on Build GRCh38
Transcripts
NM_001017371.4 NM_001172712.1 NM_003111.4

SNP Detail

SNP ID Chromosome Location
rs901590368 Chr.2: 173909291-173909291 on Build GRCh38
rs866959765 Chr.2: 173909365-173909365 on Build GRCh38
rs759387008 Chr.2: 173909164-173909175 on Build GRCh38
rs970052013 Chr.2: 173909363-173909363 on Build GRCh38
rs778030382 Chr.2: 173909439-173909439 on Build GRCh38
rs939151805 Chr.2: 173909241-173909241 on Build GRCh38
rs540847699 Chr.2: 173909312-173909312 on Build GRCh38
rs568870265 Chr.2: 173909453-173909453 on Build GRCh38
rs1038746854 Chr.2: 173909276-173909276 on Build GRCh38
rs960103928 Chr.2: 173909543-173909545 on Build GRCh38
rs762685885 Chr.2: 173909198-173909199 on Build GRCh38
rs528658839 Chr.2: 173909254-173909254 on Build GRCh38
rs772286346 Chr.2: 173909452-173909456 on Build GRCh38
rs565422794 Chr.2: 173909402-173909402 on Build GRCh38
rs997719360 Chr.2: 173909319-173909319 on Build GRCh38
rs757391059 Chr.2: 173909465-173909465 on Build GRCh38
rs945298158 Chr.2: 173909257-173909257 on Build GRCh38
rs561160998 Chr.2: 173909128-173909129 on Build GRCh38
rs903494149 Chr.2: 173909268-173909272 on Build GRCh38
rs1016693423 Chr.2: 173909358-173909362 on Build GRCh38
rs926468771 Chr.2: 173909236-173909236 on Build GRCh38
rs1007734050 Chr.2: 173909270-173909271 on Build GRCh38
rs980184128 Chr.2: 173909206-173909206 on Build GRCh38
rs970567932 Chr.2: 173909154-173909154 on Build GRCh38
rs532853514 Chr.2: 173909436-173909436 on Build GRCh38
rs766271523 Chr.2: 173909128-173909128 on Build GRCh38
rs550276082 Chr.2: 173909442-173909442 on Build GRCh38
rs1026472620 Chr.2: 173909339-173909339 on Build GRCh38
rs534652438 Chr.2: 173909141-173909141 on Build GRCh38
rs756276445 Chr.2: 173909171-173909171 on Build GRCh38
rs1022200391 Chr.2: 173909502-173909502 on Build GRCh38
rs779089346 Chr.2: 173909568-173909568 on Build GRCh38
rs757100528 Chr.2: 173909511-173909511 on Build GRCh38
rs950969223 Chr.2: 173909340-173909340 on Build GRCh38
rs1056221409 Chr.2: 173909250-173909250 on Build GRCh38
rs529776755 Chr.2: 173909535-173909535 on Build GRCh38
rs764391456 Chr.2: 173909262-173909262 on Build GRCh38
rs1006451613 Chr.2: 173909342-173909342 on Build GRCh38

View Full Product Details