Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 174757292-174757794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA1
Entrez Gene ID
1134
Gene Name
cholinergic receptor nicotinic alpha 1 subunit
Gene Aliases
ACHRA, ACHRD, CHRNA, CMS1A, CMS1B, CMS2A, FCCMS, SCCMS
UniGene
Hs.434479
Gene Chromosome Location
Chr.2: 174747592-174764472 on Build GRCh38
Transcripts
NM_000079.3 NM_001039523.2 XM_017003256.1 XM_017003257.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4425485 c.395G>A p.R132H Chr.2: 174757590-174757590 on Build GRCh38
2894142 c.356G>T p.G119V Chr.2: 174757629-174757629 on Build GRCh38
4672255 c.363A>C p.K121N Chr.2: 174757622-174757622 on Build GRCh38
2894144 c.340C>G p.P114A Chr.2: 174757645-174757645 on Build GRCh38
6088302 c.372C>G p.H124Q Chr.2: 174757613-174757613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923766803 Chr.2: 174757359-174757359 on Build GRCh38
rs186465668 Chr.2: 174757344-174757344 on Build GRCh38
rs891174208 Chr.2: 174757499-174757499 on Build GRCh38
rs970536281 Chr.2: 174757683-174757683 on Build GRCh38
rs1036761726 Chr.2: 174757468-174757468 on Build GRCh38
rs912426627 Chr.2: 174757417-174757417 on Build GRCh38
rs754404989 Chr.2: 174757625-174757625 on Build GRCh38
rs11327107 Chr.2: 174757451-174757451 on Build GRCh38
rs912456299 Chr.2: 174757645-174757645 on Build GRCh38
rs569212712 Chr.2: 174757491-174757491 on Build GRCh38
rs199555685 Chr.2: 174757586-174757586 on Build GRCh38
rs757698567 Chr.2: 174757627-174757627 on Build GRCh38
rs564707715 Chr.2: 174757520-174757520 on Build GRCh38
rs397986744 Chr.2: 174757453-174757453 on Build GRCh38
rs775936777 Chr.2: 174757654-174757654 on Build GRCh38
rs752761320 Chr.2: 174757612-174757612 on Build GRCh38
rs749664264 Chr.2: 174757541-174757541 on Build GRCh38
rs77764548 Chr.2: 174757695-174757695 on Build GRCh38
rs773207183 Chr.2: 174757689-174757689 on Build GRCh38
rs764809207 Chr.2: 174757596-174757596 on Build GRCh38
rs778314132 Chr.2: 174757540-174757540 on Build GRCh38
rs781433953 Chr.2: 174757527-174757527 on Build GRCh38
rs774924187 Chr.2: 174757554-174757554 on Build GRCh38
rs376844505 Chr.2: 174757590-174757590 on Build GRCh38
rs199470444 Chr.2: 174757630-174757630 on Build GRCh38
rs1025059009 Chr.2: 174757672-174757672 on Build GRCh38
rs968759827 Chr.2: 174757347-174757347 on Build GRCh38
rs3834175 Chr.2: 174757332-174757333 on Build GRCh38
rs867846997 Chr.2: 174757632-174757632 on Build GRCh38
rs777666730 Chr.2: 174757622-174757622 on Build GRCh38
rs368009217 Chr.2: 174757655-174757655 on Build GRCh38
rs767683848 Chr.2: 174757710-174757710 on Build GRCh38
rs370081129 Chr.2: 174757341-174757341 on Build GRCh38
rs180986978 Chr.2: 174757544-174757544 on Build GRCh38
rs377205515 Chr.2: 174757657-174757657 on Build GRCh38
rs919688742 Chr.2: 174757754-174757754 on Build GRCh38
rs771657663 Chr.2: 174757551-174757551 on Build GRCh38
rs550888688 Chr.2: 174757413-174757413 on Build GRCh38
rs775299837 Chr.2: 174757356-174757356 on Build GRCh38
rs762812233 Chr.2: 174757699-174757699 on Build GRCh38
rs35874191 Chr.2: 174757490-174757490 on Build GRCh38
rs759894314 Chr.2: 174757598-174757598 on Build GRCh38
rs746590795 Chr.2: 174757640-174757640 on Build GRCh38
rs867791759 Chr.2: 174757576-174757576 on Build GRCh38
rs760506557 Chr.2: 174757379-174757379 on Build GRCh38
rs756516966 Chr.2: 174757618-174757618 on Build GRCh38
rs886055150 Chr.2: 174757643-174757643 on Build GRCh38
rs553045974 Chr.2: 174757538-174757538 on Build GRCh38
rs751454407 Chr.2: 174757451-174757452 on Build GRCh38
rs35759748 Chr.2: 174757492-174757492 on Build GRCh38
rs190891827 Chr.2: 174757402-174757402 on Build GRCh38
rs760198514 Chr.2: 174757572-174757572 on Build GRCh38
rs771749641 Chr.2: 174757631-174757631 on Build GRCh38
rs972945082 Chr.2: 174757744-174757744 on Build GRCh38
rs199620699 Chr.2: 174757319-174757320 on Build GRCh38
rs398080830 Chr.2: 174757452-174757452 on Build GRCh38
rs78987947 Chr.2: 174757330-174757330 on Build GRCh38
rs868052890 Chr.2: 174757615-174757615 on Build GRCh38
rs575560034 Chr.2: 174757616-174757616 on Build GRCh38
rs749986265 Chr.2: 174757678-174757678 on Build GRCh38
rs768240064 Chr.2: 174757649-174757649 on Build GRCh38
rs368175496 Chr.2: 174757753-174757753 on Build GRCh38
rs1050964519 Chr.2: 174757405-174757405 on Build GRCh38
rs369497203 Chr.2: 174757737-174757737 on Build GRCh38
rs367964802 Chr.2: 174757441-174757441 on Build GRCh38
rs939840385 Chr.2: 174757432-174757432 on Build GRCh38
rs755183925 Chr.2: 174757525-174757525 on Build GRCh38
rs745528053 Chr.2: 174757550-174757550 on Build GRCh38
rs867954082 Chr.2: 174757616-174757617 on Build GRCh38
rs141744153 Chr.2: 174757493-174757493 on Build GRCh38
rs534792367 Chr.2: 174757348-174757348 on Build GRCh38
rs377079076 Chr.2: 174757512-174757512 on Build GRCh38
rs753250273 Chr.2: 174757617-174757618 on Build GRCh38
rs767773452 Chr.2: 174757609-174757609 on Build GRCh38
rs1053610950 Chr.2: 174757463-174757463 on Build GRCh38
rs763984762 Chr.2: 174757580-174757580 on Build GRCh38
rs374129622 Chr.2: 174757543-174757543 on Build GRCh38
rs760768769 Chr.2: 174757720-174757720 on Build GRCh38
rs769840773 Chr.2: 174757662-174757662 on Build GRCh38
rs140268343 Chr.2: 174757591-174757591 on Build GRCh38
rs202040914 Chr.2: 174757717-174757717 on Build GRCh38
rs144488904 Chr.2: 174757628-174757629 on Build GRCh38
rs905519837 Chr.2: 174757494-174757494 on Build GRCh38

View Full Product Details