Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 222220055-222220552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAX3
Entrez Gene ID
5077
Gene Name
paired box 3
Gene Aliases
CDHS, HUP2, WS1, WS3
UniGene
Hs.42146
Gene Chromosome Location
Chr.2: 222199887-222298996 on Build GRCh38
Transcripts
NM_001127366.2 NM_181457.3 NM_181458.3 NM_181459.3 NM_181460.3 NM_181461.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4782052 c.1121C>T p.S374L Chr.2: 222220192-222220192 on Build GRCh38
5006221 c.982G>A p.V328I Chr.2: 222220331-222220331 on Build GRCh38
4870130 c.1087T>A p.F363I Chr.2: 222220226-222220226 on Build GRCh38
5782071 c.985C>G p.H329D Chr.2: 222220328-222220328 on Build GRCh38
4868426 c.1075A>C p.T359P Chr.2: 222220238-222220238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531422957 Chr.2: 222220289-222220289 on Build GRCh38
rs767521734 Chr.2: 222220333-222220333 on Build GRCh38
rs776176860 Chr.2: 222220198-222220198 on Build GRCh38
rs756229758 Chr.2: 222220168-222220168 on Build GRCh38
rs200701839 Chr.2: 222220195-222220195 on Build GRCh38
rs946285656 Chr.2: 222220149-222220149 on Build GRCh38
rs967535658 Chr.2: 222220495-222220495 on Build GRCh38
rs750849292 Chr.2: 222220155-222220155 on Build GRCh38
rs957106607 Chr.2: 222220459-222220459 on Build GRCh38
rs536403392 Chr.2: 222220115-222220115 on Build GRCh38
rs759065284 Chr.2: 222220203-222220203 on Build GRCh38
rs753902442 Chr.2: 222220262-222220262 on Build GRCh38
rs13022712 Chr.2: 222220114-222220114 on Build GRCh38
rs913226154 Chr.2: 222220314-222220314 on Build GRCh38
rs766714441 Chr.2: 222220432-222220432 on Build GRCh38
rs151199924 Chr.2: 222220310-222220310 on Build GRCh38
rs767572436 Chr.2: 222220257-222220257 on Build GRCh38
rs987480491 Chr.2: 222220153-222220153 on Build GRCh38
rs374429328 Chr.2: 222220332-222220332 on Build GRCh38
rs770424826 Chr.2: 222220197-222220197 on Build GRCh38
rs565109008 Chr.2: 222220117-222220117 on Build GRCh38
rs150637194 Chr.2: 222220100-222220100 on Build GRCh38
rs931413137 Chr.2: 222220206-222220206 on Build GRCh38
rs751571864 Chr.2: 222220166-222220166 on Build GRCh38
rs906829296 Chr.2: 222220363-222220364 on Build GRCh38
rs763965838 Chr.2: 222220330-222220330 on Build GRCh38
rs755061925 Chr.2: 222220363-222220363 on Build GRCh38
rs563030257 Chr.2: 222220094-222220094 on Build GRCh38
rs747502205 Chr.2: 222220284-222220284 on Build GRCh38
rs751534630 Chr.2: 222220239-222220239 on Build GRCh38
rs149273276 Chr.2: 222220194-222220194 on Build GRCh38
rs752483694 Chr.2: 222220131-222220131 on Build GRCh38
rs751858437 Chr.2: 222220379-222220379 on Build GRCh38
rs757653739 Chr.2: 222220380-222220380 on Build GRCh38
rs762097866 Chr.2: 222220095-222220095 on Build GRCh38
rs149804699 Chr.2: 222220266-222220266 on Build GRCh38
rs749503361 Chr.2: 222220174-222220174 on Build GRCh38
rs769339706 Chr.2: 222220400-222220400 on Build GRCh38
rs551614431 Chr.2: 222220315-222220315 on Build GRCh38
rs769211597 Chr.2: 222220309-222220309 on Build GRCh38
rs748503320 Chr.2: 222220191-222220191 on Build GRCh38
rs762763009 Chr.2: 222220217-222220217 on Build GRCh38
rs758760102 Chr.2: 222220275-222220275 on Build GRCh38
rs879255434 Chr.2: 222220345-222220345 on Build GRCh38
rs758607057 Chr.2: 222220377-222220377 on Build GRCh38
rs758136826 Chr.2: 222220147-222220147 on Build GRCh38
rs376460754 Chr.2: 222220116-222220116 on Build GRCh38
rs751880147 Chr.2: 222220433-222220433 on Build GRCh38
rs987029491 Chr.2: 222220502-222220502 on Build GRCh38
rs930186148 Chr.2: 222220524-222220524 on Build GRCh38
rs553899131 Chr.2: 222220382-222220382 on Build GRCh38
rs764880742 Chr.2: 222220212-222220212 on Build GRCh38
rs371292388 Chr.2: 222220396-222220396 on Build GRCh38
rs755170133 Chr.2: 222220175-222220175 on Build GRCh38
rs149799356 Chr.2: 222220331-222220331 on Build GRCh38
rs771371167 Chr.2: 222220285-222220285 on Build GRCh38
rs371919705 Chr.2: 222220096-222220096 on Build GRCh38
rs1034054595 Chr.2: 222220446-222220446 on Build GRCh38
rs775028971 Chr.2: 222220097-222220098 on Build GRCh38
rs763833464 Chr.2: 222220231-222220231 on Build GRCh38
rs967295773 Chr.2: 222220316-222220316 on Build GRCh38
rs369403205 Chr.2: 222220413-222220413 on Build GRCh38
rs547446014 Chr.2: 222220178-222220178 on Build GRCh38
rs748365394 Chr.2: 222220272-222220272 on Build GRCh38
rs923094012 Chr.2: 222220374-222220374 on Build GRCh38
rs535854185 Chr.2: 222220485-222220485 on Build GRCh38
rs368438257 Chr.2: 222220214-222220214 on Build GRCh38
rs573620968 Chr.2: 222220464-222220464 on Build GRCh38
rs368896993 Chr.2: 222220378-222220378 on Build GRCh38
rs139806736 Chr.2: 222220162-222220162 on Build GRCh38
rs1011307192 Chr.2: 222220463-222220463 on Build GRCh38
rs146776741 Chr.2: 222220510-222220510 on Build GRCh38
rs957599534 Chr.2: 222220302-222220302 on Build GRCh38
rs750407449 Chr.2: 222220352-222220352 on Build GRCh38
rs752962005 Chr.2: 222220376-222220376 on Build GRCh38
rs35913673 Chr.2: 222220242-222220242 on Build GRCh38
rs556844210 Chr.2: 222220486-222220486 on Build GRCh38
rs45607236 Chr.2: 222220192-222220192 on Build GRCh38
rs778132947 Chr.2: 222220296-222220296 on Build GRCh38
rs985837785 Chr.2: 222220391-222220391 on Build GRCh38
rs765275049 Chr.2: 222220368-222220368 on Build GRCh38
rs552530067 Chr.2: 222220336-222220336 on Build GRCh38
rs370523775 Chr.2: 222220130-222220130 on Build GRCh38
rs745347593 Chr.2: 222220307-222220307 on Build GRCh38
rs780344754 Chr.2: 222220170-222220170 on Build GRCh38
rs778178524 Chr.2: 222220277-222220277 on Build GRCh38
rs755193173 Chr.2: 222220504-222220504 on Build GRCh38

View Full Product Details