Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3251485-3252001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf194
Entrez Gene ID
25943
Gene Name
chromosome 20 open reading frame 194
Gene Aliases
-
UniGene
Hs.516853
Gene Chromosome Location
Chr.20: 3249302-3407662 on Build GRCh38
Transcripts
NM_001009984.2 XM_005260684.3 XM_005260687.4 XM_011529208.2 XM_017027783.1

SNP Detail

SNP ID Chromosome Location
rs373677290 Chr.20: 3251542-3251542 on Build GRCh38
rs929350026 Chr.20: 3251837-3251837 on Build GRCh38
rs970748980 Chr.20: 3251644-3251644 on Build GRCh38
rs1023593777 Chr.20: 3251910-3251910 on Build GRCh38
rs1050985314 Chr.20: 3251695-3251695 on Build GRCh38
rs896002210 Chr.20: 3251582-3251582 on Build GRCh38
rs62208076 Chr.20: 3251661-3251661 on Build GRCh38
rs570548427 Chr.20: 3251720-3251720 on Build GRCh38
rs962444795 Chr.20: 3251514-3251514 on Build GRCh38
rs1025830599 Chr.20: 3251732-3251732 on Build GRCh38
rs114022546 Chr.20: 3251520-3251520 on Build GRCh38
rs754651390 Chr.20: 3251840-3251840 on Build GRCh38
rs933874145 Chr.20: 3251524-3251524 on Build GRCh38
rs562438486 Chr.20: 3251624-3251624 on Build GRCh38
rs79346697 Chr.20: 3251541-3251541 on Build GRCh38
rs550532356 Chr.20: 3251649-3251649 on Build GRCh38
rs534855622 Chr.20: 3251790-3251790 on Build GRCh38
rs955776195 Chr.20: 3251718-3251718 on Build GRCh38
rs889076487 Chr.20: 3251542-3251542 on Build GRCh38
rs1014415884 Chr.20: 3251901-3251901 on Build GRCh38
rs1037742251 Chr.20: 3251552-3251552 on Build GRCh38
rs562556430 Chr.20: 3251681-3251681 on Build GRCh38
rs533211572 Chr.20: 3251687-3251687 on Build GRCh38
rs568374075 Chr.20: 3251929-3251929 on Build GRCh38
rs911135213 Chr.20: 3251776-3251776 on Build GRCh38
rs899246405 Chr.20: 3251554-3251554 on Build GRCh38
rs985749309 Chr.20: 3251758-3251758 on Build GRCh38
rs924021749 Chr.20: 3251918-3251918 on Build GRCh38
rs941315344 Chr.20: 3251786-3251786 on Build GRCh38
rs1042893678 Chr.20: 3251655-3251655 on Build GRCh38
rs778603091 Chr.20: 3251892-3251892 on Build GRCh38
rs74560547 Chr.20: 3251793-3251793 on Build GRCh38
rs1016773910 Chr.20: 3251719-3251719 on Build GRCh38
rs946783105 Chr.20: 3251630-3251630 on Build GRCh38
rs147956220 Chr.20: 3251972-3251972 on Build GRCh38
rs972935394 Chr.20: 3251517-3251517 on Build GRCh38
rs80317278 Chr.20: 3251971-3251971 on Build GRCh38
rs993606397 Chr.20: 3251572-3251572 on Build GRCh38
rs988674937 Chr.20: 3251907-3251907 on Build GRCh38
rs915397108 Chr.20: 3251871-3251871 on Build GRCh38
rs990956116 Chr.20: 3251625-3251625 on Build GRCh38
rs943393951 Chr.20: 3251551-3251551 on Build GRCh38
rs144825647 Chr.20: 3251691-3251691 on Build GRCh38
rs933970479 Chr.20: 3251957-3251957 on Build GRCh38
rs540567883 Chr.20: 3251558-3251558 on Build GRCh38
rs1023222717 Chr.20: 3251629-3251629 on Build GRCh38
rs906095093 Chr.20: 3251912-3251912 on Build GRCh38
rs1032661022 Chr.20: 3251714-3251714 on Build GRCh38
rs1026454555 Chr.20: 3251577-3251577 on Build GRCh38
rs902482383 Chr.20: 3251685-3251686 on Build GRCh38
rs959608944 Chr.20: 3251836-3251836 on Build GRCh38
rs971066550 Chr.20: 3251757-3251757 on Build GRCh38
rs781577576 Chr.20: 3251723-3251723 on Build GRCh38
rs933842519 Chr.20: 3251724-3251724 on Build GRCh38
rs1035201804 Chr.20: 3251819-3251819 on Build GRCh38
rs529250594 Chr.20: 3251625-3251625 on Build GRCh38

View Full Product Details