Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74567995-74568491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH3
Entrez Gene ID
127254
Gene Name
glutamate rich 3
Gene Aliases
C1orf173
UniGene
Hs.531182
Gene Chromosome Location
Chr.1: 74568111-74674343 on Build GRCh38
Transcripts
NM_001002912.4 XM_017000274.1 XM_017000276.1 XM_017000277.1 XM_017000278.1 XM_017000279.1 XM_017000280.1 XM_017000281.1 XM_017000282.1 XM_017000283.1 XM_017000284.1 XM_017000285.1

SNP Detail

SNP ID Chromosome Location
rs755124817 Chr.1: 74568382-74568382 on Build GRCh38
rs1023001999 Chr.1: 74568176-74568176 on Build GRCh38
rs758592883 Chr.1: 74568426-74568426 on Build GRCh38
rs765493161 Chr.1: 74568392-74568392 on Build GRCh38
rs149146079 Chr.1: 74568297-74568297 on Build GRCh38
rs1056321296 Chr.1: 74568367-74568367 on Build GRCh38
rs970473288 Chr.1: 74568185-74568185 on Build GRCh38
rs960682744 Chr.1: 74568084-74568084 on Build GRCh38
rs903726552 Chr.1: 74568299-74568299 on Build GRCh38
rs375357934 Chr.1: 74568072-74568072 on Build GRCh38
rs1050928827 Chr.1: 74568257-74568257 on Build GRCh38
rs753922801 Chr.1: 74568204-74568204 on Build GRCh38
rs1024533377 Chr.1: 74568314-74568314 on Build GRCh38
rs570725865 Chr.1: 74568353-74568353 on Build GRCh38
rs972926274 Chr.1: 74568234-74568234 on Build GRCh38
rs34054478 Chr.1: 74568236-74568237 on Build GRCh38
rs187778141 Chr.1: 74568341-74568341 on Build GRCh38
rs1012050003 Chr.1: 74568402-74568402 on Build GRCh38
rs894442634 Chr.1: 74568310-74568310 on Build GRCh38
rs1005569966 Chr.1: 74568445-74568445 on Build GRCh38
rs902282391 Chr.1: 74568352-74568352 on Build GRCh38
rs934300218 Chr.1: 74568269-74568269 on Build GRCh38
rs983368673 Chr.1: 74568120-74568120 on Build GRCh38
rs571739938 Chr.1: 74568394-74568394 on Build GRCh38
rs933665043 Chr.1: 74568251-74568251 on Build GRCh38
rs769099906 Chr.1: 74568240-74568241 on Build GRCh38
rs1043349151 Chr.1: 74568343-74568343 on Build GRCh38
rs922743412 Chr.1: 74568259-74568259 on Build GRCh38
rs539684356 Chr.1: 74568408-74568408 on Build GRCh38
rs375985279 Chr.1: 74568208-74568208 on Build GRCh38
rs1000682071 Chr.1: 74568302-74568302 on Build GRCh38
rs549157882 Chr.1: 74568156-74568156 on Build GRCh38
rs766894993 Chr.1: 74568304-74568304 on Build GRCh38
rs945198769 Chr.1: 74568282-74568282 on Build GRCh38
rs1012557348 Chr.1: 74568149-74568149 on Build GRCh38
rs138267967 Chr.1: 74568205-74568205 on Build GRCh38
rs951556804 Chr.1: 74568081-74568081 on Build GRCh38
rs986108402 Chr.1: 74568283-74568283 on Build GRCh38

View Full Product Details