Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3777371-3777912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPEF1
Entrez Gene ID
25876
Gene Name
sperm flagellar 1
Gene Aliases
C20orf28, CLAMP, SPEF1A
UniGene
Hs.72620
Gene Chromosome Location
Chr.20: 3777504-3781466 on Build GRCh38
Transcripts
NM_015417.4 XM_005260683.4

SNP Detail

SNP ID Chromosome Location
rs568840104 Chr.20: 3777615-3777615 on Build GRCh38
rs750731079 Chr.20: 3777758-3777758 on Build GRCh38
rs1009969877 Chr.20: 3777733-3777733 on Build GRCh38
rs984687421 Chr.20: 3777853-3777853 on Build GRCh38
rs965926903 Chr.20: 3777396-3777396 on Build GRCh38
rs957624951 Chr.20: 3777699-3777699 on Build GRCh38
rs948285845 Chr.20: 3777574-3777574 on Build GRCh38
rs377738722 Chr.20: 3777538-3777538 on Build GRCh38
rs769720788 Chr.20: 3777426-3777426 on Build GRCh38
rs552804021 Chr.20: 3777602-3777602 on Build GRCh38
rs761448138 Chr.20: 3777752-3777752 on Build GRCh38
rs945801852 Chr.20: 3777453-3777453 on Build GRCh38
rs984715591 Chr.20: 3777456-3777456 on Build GRCh38
rs912241013 Chr.20: 3777461-3777461 on Build GRCh38
rs572804385 Chr.20: 3777807-3777807 on Build GRCh38
rs112217338 Chr.20: 3777794-3777794 on Build GRCh38
rs780574439 Chr.20: 3777797-3777797 on Build GRCh38
rs760268024 Chr.20: 3777873-3777873 on Build GRCh38
rs910654 Chr.20: 3777779-3777779 on Build GRCh38
rs974567801 Chr.20: 3777772-3777772 on Build GRCh38
rs898318482 Chr.20: 3777533-3777533 on Build GRCh38
rs942598230 Chr.20: 3777468-3777468 on Build GRCh38
rs912194103 Chr.20: 3777765-3777765 on Build GRCh38
rs919808802 Chr.20: 3777815-3777815 on Build GRCh38
rs117635906 Chr.20: 3777476-3777476 on Build GRCh38
rs551625188 Chr.20: 3777754-3777754 on Build GRCh38
rs921905424 Chr.20: 3777432-3777432 on Build GRCh38
rs1046941903 Chr.20: 3777543-3777543 on Build GRCh38
rs73895626 Chr.20: 3777427-3777427 on Build GRCh38
rs1032657072 Chr.20: 3777688-3777688 on Build GRCh38
rs990238876 Chr.20: 3777749-3777749 on Build GRCh38
rs931223524 Chr.20: 3777858-3777858 on Build GRCh38
rs896628821 Chr.20: 3777545-3777552 on Build GRCh38
rs528695451 Chr.20: 3777563-3777563 on Build GRCh38
rs3746642 Chr.20: 3777537-3777537 on Build GRCh38
rs763486516 Chr.20: 3777618-3777618 on Build GRCh38
rs940751551 Chr.20: 3777467-3777467 on Build GRCh38
rs556704569 Chr.20: 3777601-3777601 on Build GRCh38
rs951973137 Chr.20: 3777837-3777837 on Build GRCh38
rs1004991080 Chr.20: 3777588-3777588 on Build GRCh38
rs539716581 Chr.20: 3777680-3777680 on Build GRCh38
rs907869125 Chr.20: 3777890-3777890 on Build GRCh38
rs1040020491 Chr.20: 3777806-3777806 on Build GRCh38
rs886485469 Chr.20: 3777583-3777583 on Build GRCh38
rs967070573 Chr.20: 3777753-3777753 on Build GRCh38
rs535289240 Chr.20: 3777585-3777585 on Build GRCh38
rs1001121997 Chr.20: 3777679-3777679 on Build GRCh38
rs1046908546 Chr.20: 3777865-3777865 on Build GRCh38
rs971451138 Chr.20: 3777611-3777611 on Build GRCh38
rs989850685 Chr.20: 3777425-3777425 on Build GRCh38
rs751124160 Chr.20: 3777644-3777644 on Build GRCh38
rs1050694158 Chr.20: 3777457-3777457 on Build GRCh38

View Full Product Details