Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32791239-32791778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf62
Entrez Gene ID
56245
Gene Name
chromosome 21 open reading frame 62
Gene Aliases
B37, C21orf120, PRED81
UniGene
Hs.517235
Gene Chromosome Location
Chr.21: 32790673-32813808 on Build GRCh38
Transcripts
NM_001162495.2 NM_001162496.2 NM_019596.5 NR_024622.1 NR_024623.1

Gene Symbol
C21orf62-AS1
Entrez Gene ID
54067
Gene Name
C21orf62 antisense RNA 1
Gene Aliases
C21orf49
UniGene
Hs.54725
Gene Chromosome Location
Chr.21: 32772100-32797707 on Build GRCh38
Transcripts
NM_001162495.2 NM_001162496.2 NM_019596.5 NR_024622.1 NR_024623.1

SNP Detail

SNP ID Chromosome Location
rs554783342 Chr.21: 32791364-32791364 on Build GRCh38
rs776529035 Chr.21: 32791720-32791720 on Build GRCh38
rs142317078 Chr.21: 32791296-32791296 on Build GRCh38
rs1013723171 Chr.21: 32791711-32791711 on Build GRCh38
rs1026618062 Chr.21: 32791341-32791341 on Build GRCh38
rs751314457 Chr.21: 32791673-32791673 on Build GRCh38
rs890114458 Chr.21: 32791340-32791340 on Build GRCh38
rs777561521 Chr.21: 32791469-32791469 on Build GRCh38
rs182012591 Chr.21: 32791298-32791298 on Build GRCh38
rs925267059 Chr.21: 32791481-32791481 on Build GRCh38
rs1014745340 Chr.21: 32791559-32791559 on Build GRCh38
rs1004274219 Chr.21: 32791653-32791653 on Build GRCh38
rs771724382 Chr.21: 32791457-32791457 on Build GRCh38
rs186993285 Chr.21: 32791368-32791368 on Build GRCh38
rs1052700029 Chr.21: 32791272-32791272 on Build GRCh38
rs892337370 Chr.21: 32791283-32791283 on Build GRCh38
rs1040876276 Chr.21: 32791582-32791582 on Build GRCh38
rs1048170583 Chr.21: 32791604-32791604 on Build GRCh38
rs1052396151 Chr.21: 32791501-32791501 on Build GRCh38
rs577304220 Chr.21: 32791573-32791573 on Build GRCh38
rs865975562 Chr.21: 32791592-32791592 on Build GRCh38
rs1036591483 Chr.21: 32791382-32791382 on Build GRCh38
rs189506706 Chr.21: 32791463-32791463 on Build GRCh38
rs16989777 Chr.21: 32791337-32791337 on Build GRCh38
rs189858138 Chr.21: 32791292-32791292 on Build GRCh38
rs992270726 Chr.21: 32791450-32791450 on Build GRCh38
rs958199780 Chr.21: 32791678-32791678 on Build GRCh38
rs931863733 Chr.21: 32791734-32791735 on Build GRCh38
rs1015201147 Chr.21: 32791663-32791663 on Build GRCh38
rs565316397 Chr.21: 32791511-32791511 on Build GRCh38
rs746714114 Chr.21: 32791544-32791544 on Build GRCh38
rs952401160 Chr.21: 32791357-32791357 on Build GRCh38
rs879233697 Chr.21: 32791735-32791736 on Build GRCh38
rs770823127 Chr.21: 32791586-32791586 on Build GRCh38
rs544976717 Chr.21: 32791700-32791700 on Build GRCh38
rs888154213 Chr.21: 32791621-32791621 on Build GRCh38
rs936663000 Chr.21: 32791487-32791487 on Build GRCh38
rs962267694 Chr.21: 32791442-32791442 on Build GRCh38
rs2833956 Chr.21: 32791431-32791431 on Build GRCh38
rs989453396 Chr.21: 32791679-32791679 on Build GRCh38
rs768803653 Chr.21: 32791735-32791735 on Build GRCh38
rs899565484 Chr.21: 32791336-32791336 on Build GRCh38
rs996558132 Chr.21: 32791603-32791603 on Build GRCh38

View Full Product Details