Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38313991-38314486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1E
Entrez Gene ID
1454
Gene Name
casein kinase 1 epsilon
Gene Aliases
CKIepsilon, HCKIE
UniGene
Hs.474833
Gene Chromosome Location
Chr.22: 38290691-38318084 on Build GRCh38
Transcripts
NM_001894.4 NM_152221.2 NM_001289912.1

Gene Symbol
LOC400927-CSNK1E
Entrez Gene ID
102800317
Gene Name
LOC400927-CSNK1E readthrough
Gene Aliases
-
UniGene
Hs.720223
Gene Chromosome Location
Chr.22: 38290691-38398522 on Build GRCh38
Transcripts
NM_001894.4 NM_152221.2 NM_001289912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
350317 c.64G>T p.D22Y Chr.22: 38314094-38314094 on Build GRCh38
6636764 c.25T>C p.Y9H Chr.22: 38314133-38314133 on Build GRCh38
350318 c.64G>T p.D22Y Chr.22: 38314094-38314094 on Build GRCh38
4675141 c.37C>T p.R13W Chr.22: 38314121-38314121 on Build GRCh38
350316 c.64G>T p.D22Y Chr.22: 38314094-38314094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs5757034 Chr.22: 38314459-38314459 on Build GRCh38
rs757946999 Chr.22: 38314125-38314125 on Build GRCh38
rs373100204 Chr.22: 38314147-38314147 on Build GRCh38
rs540096910 Chr.22: 38314327-38314327 on Build GRCh38
rs945839259 Chr.22: 38314410-38314410 on Build GRCh38
rs759740908 Chr.22: 38314197-38314197 on Build GRCh38
rs537054296 Chr.22: 38314458-38314458 on Build GRCh38
rs931199459 Chr.22: 38314136-38314136 on Build GRCh38
rs997620105 Chr.22: 38314049-38314049 on Build GRCh38
rs770996047 Chr.22: 38314190-38314190 on Build GRCh38
rs536832784 Chr.22: 38314261-38314261 on Build GRCh38
rs914378333 Chr.22: 38314138-38314138 on Build GRCh38
rs1017627678 Chr.22: 38314016-38314016 on Build GRCh38
rs767961407 Chr.22: 38314200-38314200 on Build GRCh38
rs746723989 Chr.22: 38314131-38314131 on Build GRCh38
rs180982614 Chr.22: 38314069-38314069 on Build GRCh38
rs751568347 Chr.22: 38314162-38314163 on Build GRCh38
rs761139121 Chr.22: 38314230-38314230 on Build GRCh38
rs135764 Chr.22: 38314407-38314407 on Build GRCh38
rs374582268 Chr.22: 38314375-38314375 on Build GRCh38
rs952205823 Chr.22: 38314054-38314054 on Build GRCh38
rs184162238 Chr.22: 38314262-38314262 on Build GRCh38
rs1039030142 Chr.22: 38314185-38314185 on Build GRCh38
rs201363944 Chr.22: 38314168-38314168 on Build GRCh38
rs966309049 Chr.22: 38314302-38314302 on Build GRCh38
rs773953310 Chr.22: 38314166-38314166 on Build GRCh38
rs547567641 Chr.22: 38314277-38314277 on Build GRCh38
rs370688398 Chr.22: 38314195-38314195 on Build GRCh38
rs1024581707 Chr.22: 38314360-38314360 on Build GRCh38
rs770165554 Chr.22: 38314152-38314152 on Build GRCh38
rs55915805 Chr.22: 38314107-38314107 on Build GRCh38
rs370329105 Chr.22: 38314121-38314121 on Build GRCh38
rs768411971 Chr.22: 38314059-38314059 on Build GRCh38
rs753274949 Chr.22: 38314220-38314220 on Build GRCh38
rs761798313 Chr.22: 38314067-38314067 on Build GRCh38
rs766942775 Chr.22: 38314213-38314213 on Build GRCh38
rs370828053 Chr.22: 38314188-38314188 on Build GRCh38
rs565960820 Chr.22: 38314295-38314295 on Build GRCh38
rs549797853 Chr.22: 38314178-38314178 on Build GRCh38
rs753159576 Chr.22: 38314201-38314201 on Build GRCh38
rs761215064 Chr.22: 38314204-38314204 on Build GRCh38
rs567809542 Chr.22: 38314214-38314214 on Build GRCh38
rs989881624 Chr.22: 38314401-38314401 on Build GRCh38
rs1015139509 Chr.22: 38314273-38314273 on Build GRCh38
rs569993722 Chr.22: 38314427-38314427 on Build GRCh38
rs900701029 Chr.22: 38314347-38314357 on Build GRCh38
rs768756857 Chr.22: 38314040-38314040 on Build GRCh38
rs953178303 Chr.22: 38314398-38314398 on Build GRCh38
rs1027751571 Chr.22: 38314228-38314228 on Build GRCh38
rs1021105057 Chr.22: 38314053-38314053 on Build GRCh38
rs1028636401 Chr.22: 38314387-38314387 on Build GRCh38
rs201811069 Chr.22: 38314128-38314128 on Build GRCh38
rs749970868 Chr.22: 38314120-38314120 on Build GRCh38
rs377114359 Chr.22: 38314098-38314098 on Build GRCh38
rs377709817 Chr.22: 38314113-38314113 on Build GRCh38
rs773976199 Chr.22: 38314130-38314130 on Build GRCh38
rs879838958 Chr.22: 38314038-38314038 on Build GRCh38
rs990406832 Chr.22: 38314097-38314097 on Build GRCh38
rs1015190542 Chr.22: 38314078-38314078 on Build GRCh38
rs938630249 Chr.22: 38314203-38314203 on Build GRCh38
rs527571036 Chr.22: 38314104-38314104 on Build GRCh38
rs574640234 Chr.22: 38314062-38314062 on Build GRCh38
rs113683448 Chr.22: 38314295-38314295 on Build GRCh38
rs895222606 Chr.22: 38314020-38314020 on Build GRCh38
rs921624032 Chr.22: 38314119-38314119 on Build GRCh38
rs887909278 Chr.22: 38314244-38314244 on Build GRCh38
rs768049492 Chr.22: 38314106-38314106 on Build GRCh38
rs1012772929 Chr.22: 38314030-38314030 on Build GRCh38
rs976766393 Chr.22: 38314335-38314335 on Build GRCh38
rs748049706 Chr.22: 38314167-38314167 on Build GRCh38

View Full Product Details