Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14124853-14125359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHCHD4
Entrez Gene ID
131474
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 4
Gene Aliases
MIA40, TIMM40
UniGene
Hs.517815
Gene Chromosome Location
Chr.3: 14112077-14124871 on Build GRCh38
Transcripts
NM_001098502.1 NM_144636.2 NM_024334.2 XM_017007176.1

Gene Symbol
TMEM43
Entrez Gene ID
79188
Gene Name
transmembrane protein 43
Gene Aliases
ARVC5, ARVD5, EDMD7, LUMA
UniGene
Hs.517817
Gene Chromosome Location
Chr.3: 14124940-14143680 on Build GRCh38
Transcripts
NM_001098502.1 NM_144636.2 NM_024334.2 XM_017007176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4398650 c.11A>T p.N4I Chr.3: 14125204-14125204 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs535987039 Chr.3: 14125036-14125036 on Build GRCh38
rs111954359 Chr.3: 14125168-14125168 on Build GRCh38
rs908976597 Chr.3: 14125003-14125003 on Build GRCh38
rs1031529543 Chr.3: 14125021-14125021 on Build GRCh38
rs971781776 Chr.3: 14125012-14125012 on Build GRCh38
rs956103773 Chr.3: 14125031-14125031 on Build GRCh38
rs1057037939 Chr.3: 14125316-14125316 on Build GRCh38
rs912834494 Chr.3: 14125252-14125252 on Build GRCh38
rs1036409657 Chr.3: 14124881-14124881 on Build GRCh38
rs753567531 Chr.3: 14125028-14125028 on Build GRCh38
rs1056602621 Chr.3: 14125086-14125086 on Build GRCh38
rs886058026 Chr.3: 14125004-14125004 on Build GRCh38
rs866805269 Chr.3: 14125005-14125005 on Build GRCh38
rs964829809 Chr.3: 14125264-14125264 on Build GRCh38
rs535621654 Chr.3: 14125081-14125081 on Build GRCh38
rs886058028 Chr.3: 14125040-14125040 on Build GRCh38
rs750657252 Chr.3: 14125217-14125217 on Build GRCh38
rs943062299 Chr.3: 14124956-14124956 on Build GRCh38
rs1003696402 Chr.3: 14124994-14124994 on Build GRCh38
rs557505982 Chr.3: 14125042-14125042 on Build GRCh38
rs546844971 Chr.3: 14124949-14124949 on Build GRCh38
rs758354104 Chr.3: 14125257-14125257 on Build GRCh38
rs987049616 Chr.3: 14125024-14125024 on Build GRCh38
rs765619239 Chr.3: 14125195-14125195 on Build GRCh38
rs1016495734 Chr.3: 14125207-14125207 on Build GRCh38
rs780112747 Chr.3: 14125220-14125220 on Build GRCh38
rs188988244 Chr.3: 14124970-14124970 on Build GRCh38
rs144173472 Chr.3: 14124971-14124971 on Build GRCh38
rs551084788 Chr.3: 14125055-14125055 on Build GRCh38
rs368669606 Chr.3: 14125159-14125159 on Build GRCh38
rs942925546 Chr.3: 14125269-14125269 on Build GRCh38
rs916727632 Chr.3: 14125329-14125329 on Build GRCh38
rs930857147 Chr.3: 14124986-14124986 on Build GRCh38
rs370434063 Chr.3: 14125328-14125328 on Build GRCh38
rs930195980 Chr.3: 14124893-14124893 on Build GRCh38
rs202159933 Chr.3: 14125239-14125239 on Build GRCh38
rs1016135408 Chr.3: 14125324-14125324 on Build GRCh38
rs866943820 Chr.3: 14125038-14125038 on Build GRCh38
rs192235215 Chr.3: 14125221-14125221 on Build GRCh38
rs770141102 Chr.3: 14125245-14125245 on Build GRCh38
rs372124334 Chr.3: 14125238-14125238 on Build GRCh38
rs537513930 Chr.3: 14125037-14125037 on Build GRCh38
rs763829810 Chr.3: 14125191-14125191 on Build GRCh38
rs2733579 Chr.3: 14125335-14125335 on Build GRCh38
rs754747623 Chr.3: 14125192-14125192 on Build GRCh38
rs774049247 Chr.3: 14125253-14125253 on Build GRCh38
rs762480165 Chr.3: 14124991-14124991 on Build GRCh38
rs1041785368 Chr.3: 14125307-14125307 on Build GRCh38
rs918915702 Chr.3: 14125015-14125015 on Build GRCh38
rs757083718 Chr.3: 14125194-14125194 on Build GRCh38
rs568648347 Chr.3: 14125002-14125002 on Build GRCh38
rs767001935 Chr.3: 14125183-14125183 on Build GRCh38
rs895225601 Chr.3: 14124998-14124998 on Build GRCh38
rs974422942 Chr.3: 14125038-14125039 on Build GRCh38
rs140833608 Chr.3: 14125313-14125313 on Build GRCh38
rs760598925 Chr.3: 14125190-14125190 on Build GRCh38
rs1039796566 Chr.3: 14124915-14124915 on Build GRCh38
rs975636714 Chr.3: 14125284-14125284 on Build GRCh38
rs563715581 Chr.3: 14125114-14125114 on Build GRCh38
rs540373459 Chr.3: 14124988-14124989 on Build GRCh38
rs908122657 Chr.3: 14125064-14125064 on Build GRCh38
rs577237727 Chr.3: 14125240-14125240 on Build GRCh38
rs974519399 Chr.3: 14125280-14125280 on Build GRCh38
rs1035579595 Chr.3: 14125115-14125115 on Build GRCh38
rs964452765 Chr.3: 14125038-14125038 on Build GRCh38
rs572207514 Chr.3: 14125137-14125137 on Build GRCh38
rs903243400 Chr.3: 14125311-14125311 on Build GRCh38
rs1011593497 Chr.3: 14124940-14124940 on Build GRCh38
rs886058027 Chr.3: 14125019-14125019 on Build GRCh38
rs17038940 Chr.3: 14125052-14125052 on Build GRCh38
rs987647645 Chr.3: 14124929-14124929 on Build GRCh38
rs963434461 Chr.3: 14125010-14125010 on Build GRCh38
rs886058029 Chr.3: 14125064-14125064 on Build GRCh38
rs543050551 Chr.3: 14125301-14125301 on Build GRCh38
rs140252037 Chr.3: 14124933-14124933 on Build GRCh38
rs587780964 Chr.3: 14125201-14125201 on Build GRCh38
rs942857169 Chr.3: 14125034-14125034 on Build GRCh38
rs1013238755 Chr.3: 14125321-14125321 on Build GRCh38
rs891293044 Chr.3: 14124899-14124899 on Build GRCh38
rs111995799 Chr.3: 14125001-14125001 on Build GRCh38
rs886058025 Chr.3: 14124990-14124990 on Build GRCh38
rs967295317 Chr.3: 14124974-14124974 on Build GRCh38
rs914717453 Chr.3: 14125285-14125285 on Build GRCh38
rs951731652 Chr.3: 14125310-14125310 on Build GRCh38
rs1035525527 Chr.3: 14124980-14124980 on Build GRCh38
rs963190652 Chr.3: 14125226-14125226 on Build GRCh38
rs745941923 Chr.3: 14125319-14125319 on Build GRCh38
rs903187822 Chr.3: 14125073-14125073 on Build GRCh38
rs930282644 Chr.3: 14125046-14125047 on Build GRCh38
rs939549729 Chr.3: 14125074-14125074 on Build GRCh38
rs893244209 Chr.3: 14125040-14125040 on Build GRCh38
rs747502348 Chr.3: 14125229-14125229 on Build GRCh38
rs368037403 Chr.3: 14124975-14124975 on Build GRCh38
rs918867085 Chr.3: 14124885-14124885 on Build GRCh38
rs1041753893 Chr.3: 14125068-14125068 on Build GRCh38
rs947534014 Chr.3: 14125292-14125292 on Build GRCh38
rs377412476 Chr.3: 14125127-14125127 on Build GRCh38
rs1056989847 Chr.3: 14124997-14124997 on Build GRCh38
rs1009147604 Chr.3: 14125241-14125241 on Build GRCh38
rs2733578 Chr.3: 14124894-14124894 on Build GRCh38
rs1022216009 Chr.3: 14125004-14125004 on Build GRCh38
rs773961732 Chr.3: 14125150-14125150 on Build GRCh38
rs555579055 Chr.3: 14125102-14125102 on Build GRCh38
rs571615896 Chr.3: 14125020-14125020 on Build GRCh38
rs948213282 Chr.3: 14125332-14125332 on Build GRCh38
rs1043722997 Chr.3: 14125145-14125145 on Build GRCh38
rs1007022971 Chr.3: 14124906-14124906 on Build GRCh38

View Full Product Details