Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 21425394-21425935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF385D
Entrez Gene ID
79750
Gene Name
zinc finger protein 385D
Gene Aliases
ZNF659
UniGene
Hs.21026
Gene Chromosome Location
Chr.3: 21418419-22372641 on Build GRCh38
Transcripts
NM_024697.2 XM_017007191.1 XM_017007192.1 XM_017007193.1 XM_017007194.1 XM_017007195.1 XM_017007196.1 XM_017007197.1 XM_017007198.1 XM_017007199.1 XM_017007200.1 XM_017007201.1 XM_017007202.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1233972 c.710G>A p.G237E Chr.3: 21425634-21425634 on Build GRCh38
4743682 c.726A>T p.K242N Chr.3: 21425618-21425618 on Build GRCh38
6277313 c.690C>A p.T230T Chr.3: 21425654-21425654 on Build GRCh38
1233973 c.839C>T p.T280M Chr.3: 21425505-21425505 on Build GRCh38
4743683 c.682C>T p.H228Y Chr.3: 21425662-21425662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767801307 Chr.3: 21425582-21425582 on Build GRCh38
rs189554171 Chr.3: 21425830-21425830 on Build GRCh38
rs757458233 Chr.3: 21425506-21425506 on Build GRCh38
rs774046883 Chr.3: 21425577-21425577 on Build GRCh38
rs772739527 Chr.3: 21425553-21425553 on Build GRCh38
rs755175151 Chr.3: 21425697-21425697 on Build GRCh38
rs975143883 Chr.3: 21425715-21425715 on Build GRCh38
rs754373020 Chr.3: 21425702-21425702 on Build GRCh38
rs186892 Chr.3: 21425753-21425753 on Build GRCh38
rs200924075 Chr.3: 21425462-21425462 on Build GRCh38
rs199797044 Chr.3: 21425668-21425668 on Build GRCh38
rs544683484 Chr.3: 21425802-21425802 on Build GRCh38
rs1035977777 Chr.3: 21425489-21425489 on Build GRCh38
rs780385176 Chr.3: 21425605-21425605 on Build GRCh38
rs761191649 Chr.3: 21425593-21425593 on Build GRCh38
rs755288391 Chr.3: 21425762-21425762 on Build GRCh38
rs376269511 Chr.3: 21425709-21425709 on Build GRCh38
rs1055133780 Chr.3: 21425660-21425660 on Build GRCh38
rs534530271 Chr.3: 21425733-21425733 on Build GRCh38
rs965098157 Chr.3: 21425461-21425461 on Build GRCh38
rs201734897 Chr.3: 21425753-21425754 on Build GRCh38
rs779277619 Chr.3: 21425641-21425641 on Build GRCh38
rs762619972 Chr.3: 21425469-21425469 on Build GRCh38
rs568626249 Chr.3: 21425854-21425854 on Build GRCh38
rs748815900 Chr.3: 21425675-21425675 on Build GRCh38
rs769155425 Chr.3: 21425757-21425757 on Build GRCh38
rs764537413 Chr.3: 21425607-21425607 on Build GRCh38
rs369607136 Chr.3: 21425576-21425576 on Build GRCh38
rs370078943 Chr.3: 21425505-21425505 on Build GRCh38
rs780375691 Chr.3: 21425695-21425695 on Build GRCh38
rs781378690 Chr.3: 21425805-21425805 on Build GRCh38
rs759889401 Chr.3: 21425581-21425581 on Build GRCh38
rs767997996 Chr.3: 21425486-21425486 on Build GRCh38
rs149202584 Chr.3: 21425658-21425658 on Build GRCh38
rs750554703 Chr.3: 21425720-21425720 on Build GRCh38
rs1055055782 Chr.3: 21425863-21425863 on Build GRCh38
rs757226046 Chr.3: 21425429-21425429 on Build GRCh38
rs764738325 Chr.3: 21425458-21425458 on Build GRCh38
rs140291218 Chr.3: 21425587-21425587 on Build GRCh38
rs770494574 Chr.3: 21425679-21425679 on Build GRCh38
rs9818735 Chr.3: 21425610-21425610 on Build GRCh38
rs760981940 Chr.3: 21425696-21425696 on Build GRCh38
rs766052490 Chr.3: 21425474-21425474 on Build GRCh38
rs181821460 Chr.3: 21425875-21425875 on Build GRCh38
rs1051659313 Chr.3: 21425438-21425438 on Build GRCh38
rs201544208 Chr.3: 21425689-21425689 on Build GRCh38
rs911780781 Chr.3: 21425746-21425747 on Build GRCh38
rs201622878 Chr.3: 21425504-21425504 on Build GRCh38
rs903086560 Chr.3: 21425693-21425693 on Build GRCh38
rs746789731 Chr.3: 21425518-21425518 on Build GRCh38
rs3765027 Chr.3: 21425687-21425687 on Build GRCh38
rs765353996 Chr.3: 21425718-21425718 on Build GRCh38
rs753012936 Chr.3: 21425585-21425585 on Build GRCh38
rs200280562 Chr.3: 21425510-21425510 on Build GRCh38
rs768694954 Chr.3: 21425451-21425451 on Build GRCh38
rs750610082 Chr.3: 21425642-21425642 on Build GRCh38
rs753935865 Chr.3: 21425615-21425615 on Build GRCh38
rs898821431 Chr.3: 21425760-21425760 on Build GRCh38
rs1044767901 Chr.3: 21425799-21425799 on Build GRCh38
rs115693487 Chr.3: 21425701-21425701 on Build GRCh38
rs867503259 Chr.3: 21425735-21425735 on Build GRCh38
rs776772084 Chr.3: 21425453-21425453 on Build GRCh38
rs143903868 Chr.3: 21425519-21425519 on Build GRCh38
rs562851061 Chr.3: 21425809-21425809 on Build GRCh38
rs748061594 Chr.3: 21425536-21425536 on Build GRCh38
rs773855260 Chr.3: 21425682-21425682 on Build GRCh38
rs758708325 Chr.3: 21425645-21425645 on Build GRCh38
rs746844724 Chr.3: 21425449-21425449 on Build GRCh38
rs200229221 Chr.3: 21425547-21425547 on Build GRCh38
rs908759758 Chr.3: 21425602-21425602 on Build GRCh38
rs761229002 Chr.3: 21425454-21425454 on Build GRCh38
rs893895710 Chr.3: 21425673-21425673 on Build GRCh38
rs968680481 Chr.3: 21425867-21425867 on Build GRCh38
rs775373183 Chr.3: 21425447-21425447 on Build GRCh38
rs775881150 Chr.3: 21425690-21425690 on Build GRCh38
rs1001675083 Chr.3: 21425826-21425826 on Build GRCh38
rs1046368520 Chr.3: 21425639-21425639 on Build GRCh38
rs771067417 Chr.3: 21425445-21425445 on Build GRCh38
rs150069278 Chr.3: 21425652-21425652 on Build GRCh38
rs953343145 Chr.3: 21425878-21425878 on Build GRCh38
rs937971094 Chr.3: 21425846-21425846 on Build GRCh38
rs1029040267 Chr.3: 21425877-21425877 on Build GRCh38
rs73820942 Chr.3: 21425480-21425480 on Build GRCh38
rs762412358 Chr.3: 21425566-21425566 on Build GRCh38
rs375068613 Chr.3: 21425501-21425501 on Build GRCh38
rs574303245 Chr.3: 21425788-21425788 on Build GRCh38
rs765097555 Chr.3: 21425640-21425640 on Build GRCh38
rs757473490 Chr.3: 21425617-21425617 on Build GRCh38
rs776576227 Chr.3: 21425525-21425525 on Build GRCh38
rs965407717 Chr.3: 21425710-21425710 on Build GRCh38
rs1020327943 Chr.3: 21425852-21425852 on Build GRCh38
rs896110239 Chr.3: 21425841-21425841 on Build GRCh38

View Full Product Details