Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 45901123-45901629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR9
Entrez Gene ID
10803
Gene Name
C-C motif chemokine receptor 9
Gene Aliases
CC-CKR-9, CDw199, GPR-9-6, GPR28
UniGene
Hs.225946
Gene Chromosome Location
Chr.3: 45886504-45903177 on Build GRCh38
Transcripts
NM_001256369.1 NM_006641.3 NM_031200.2 XM_011533312.2 NM_001276378.1 NM_001276379.1 XM_017006644.1 XM_017006645.1

Gene Symbol
LZTFL1
Entrez Gene ID
54585
Gene Name
leucine zipper transcription factor like 1
Gene Aliases
BBS17
UniGene
Hs.30824
Gene Chromosome Location
Chr.3: 45823316-45916042 on Build GRCh38
Transcripts
NM_001256369.1 NM_006641.3 NM_031200.2 XM_011533312.2 NM_001276378.1 NM_001276379.1 XM_017006644.1 XM_017006645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4552815 c.571G>A p.E191K Chr.3: 45901359-45901359 on Build GRCh38
4543388 c.336G>A p.W112* Chr.3: 45901124-45901124 on Build GRCh38
4517438 c.342_343CC>TT p.Q115* Chr.3: 45901130-45901131 on Build GRCh38
239217 c.436_437insA p.I146fs*59 Chr.3: 45901224-45901225 on Build GRCh38
4754248 c.396C>A p.S132R Chr.3: 45901184-45901184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979032957 Chr.3: 45901432-45901432 on Build GRCh38
rs758349964 Chr.3: 45901506-45901506 on Build GRCh38
rs747802087 Chr.3: 45901273-45901273 on Build GRCh38
rs199541347 Chr.3: 45901212-45901212 on Build GRCh38
rs770003172 Chr.3: 45901409-45901409 on Build GRCh38
rs200638038 Chr.3: 45901550-45901550 on Build GRCh38
rs1040223989 Chr.3: 45901229-45901229 on Build GRCh38
rs770786692 Chr.3: 45901223-45901223 on Build GRCh38
rs778368692 Chr.3: 45901492-45901492 on Build GRCh38
rs61755293 Chr.3: 45901274-45901274 on Build GRCh38
rs374133687 Chr.3: 45901442-45901442 on Build GRCh38
rs145949330 Chr.3: 45901469-45901469 on Build GRCh38
rs763090743 Chr.3: 45901424-45901424 on Build GRCh38
rs372342915 Chr.3: 45901538-45901538 on Build GRCh38
rs757593117 Chr.3: 45901500-45901500 on Build GRCh38
rs200041385 Chr.3: 45901277-45901277 on Build GRCh38
rs754245427 Chr.3: 45901599-45901599 on Build GRCh38
rs775184107 Chr.3: 45901255-45901255 on Build GRCh38
rs1021507742 Chr.3: 45901346-45901346 on Build GRCh38
rs771928543 Chr.3: 45901245-45901245 on Build GRCh38
rs61751653 Chr.3: 45901479-45901479 on Build GRCh38
rs767902559 Chr.3: 45901318-45901318 on Build GRCh38
rs773024857 Chr.3: 45901457-45901460 on Build GRCh38
rs201651405 Chr.3: 45901300-45901300 on Build GRCh38
rs761059125 Chr.3: 45901580-45901580 on Build GRCh38
rs199702895 Chr.3: 45901521-45901521 on Build GRCh38
rs374047706 Chr.3: 45901344-45901344 on Build GRCh38
rs769377797 Chr.3: 45901360-45901360 on Build GRCh38
rs200323402 Chr.3: 45901386-45901386 on Build GRCh38
rs944119215 Chr.3: 45901228-45901228 on Build GRCh38
rs201414744 Chr.3: 45901416-45901416 on Build GRCh38
rs200677145 Chr.3: 45901470-45901470 on Build GRCh38
rs35423527 Chr.3: 45901194-45901194 on Build GRCh38
rs201764174 Chr.3: 45901211-45901211 on Build GRCh38
rs568929897 Chr.3: 45901244-45901244 on Build GRCh38
rs34576487 Chr.3: 45901582-45901583 on Build GRCh38
rs779871093 Chr.3: 45901516-45901516 on Build GRCh38
rs768371173 Chr.3: 45901536-45901536 on Build GRCh38
rs370914315 Chr.3: 45901289-45901289 on Build GRCh38
rs774569331 Chr.3: 45901563-45901563 on Build GRCh38
rs144940552 Chr.3: 45901258-45901258 on Build GRCh38
rs200611238 Chr.3: 45901581-45901581 on Build GRCh38
rs763686968 Chr.3: 45901483-45901483 on Build GRCh38
rs1039558767 Chr.3: 45901569-45901569 on Build GRCh38
rs200806590 Chr.3: 45901208-45901208 on Build GRCh38
rs1035842645 Chr.3: 45901347-45901347 on Build GRCh38
rs113137843 Chr.3: 45901368-45901368 on Build GRCh38
rs753520925 Chr.3: 45901489-45901489 on Build GRCh38
rs201163971 Chr.3: 45901312-45901312 on Build GRCh38
rs377674879 Chr.3: 45901243-45901243 on Build GRCh38
rs749957221 Chr.3: 45901493-45901493 on Build GRCh38
rs202016210 Chr.3: 45901379-45901379 on Build GRCh38
rs149823113 Chr.3: 45901555-45901555 on Build GRCh38
rs202237812 Chr.3: 45901319-45901319 on Build GRCh38
rs759541158 Chr.3: 45901571-45901571 on Build GRCh38
rs201973846 Chr.3: 45901401-45901401 on Build GRCh38
rs200109122 Chr.3: 45901354-45901354 on Build GRCh38
rs766947395 Chr.3: 45901308-45901308 on Build GRCh38
rs141006341 Chr.3: 45901398-45901398 on Build GRCh38
rs201772828 Chr.3: 45901388-45901388 on Build GRCh38
rs201444207 Chr.3: 45901261-45901261 on Build GRCh38
rs765431789 Chr.3: 45901270-45901270 on Build GRCh38
rs202070482 Chr.3: 45901572-45901572 on Build GRCh38
rs771948402 Chr.3: 45901400-45901400 on Build GRCh38
rs200673880 Chr.3: 45901292-45901292 on Build GRCh38
rs748863151 Chr.3: 45901218-45901218 on Build GRCh38
rs559576247 Chr.3: 45901304-45901304 on Build GRCh38
rs887128612 Chr.3: 45901600-45901600 on Build GRCh38
rs147314165 Chr.3: 45901310-45901310 on Build GRCh38
rs201241644 Chr.3: 45901162-45901162 on Build GRCh38
rs774622553 Chr.3: 45901461-45901461 on Build GRCh38
rs756940037 Chr.3: 45901491-45901491 on Build GRCh38
rs367903658 Chr.3: 45901367-45901367 on Build GRCh38
rs148893037 Chr.3: 45901406-45901406 on Build GRCh38
rs200477691 Chr.3: 45901225-45901225 on Build GRCh38

View Full Product Details