Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51541495-51541992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD54L2
Entrez Gene ID
23132
Gene Name
RAD54-like 2 (S. cerevisiae)
Gene Aliases
ARIP4, HSPC325, SRISNF2L
UniGene
Hs.105399
Gene Chromosome Location
Chr.3: 51538683-51668660 on Build GRCh38
Transcripts
NM_001322253.1 NM_001322256.1 NM_001322257.1 NM_015106.3 XM_017005987.1 XM_017005988.1

SNP Detail

SNP ID Chromosome Location
rs782802746 Chr.3: 51541525-51541525 on Build GRCh38
rs536646687 Chr.3: 51541731-51541731 on Build GRCh38
rs886310780 Chr.3: 51541553-51541553 on Build GRCh38
rs966285558 Chr.3: 51541844-51541844 on Build GRCh38
rs577195273 Chr.3: 51541861-51541861 on Build GRCh38
rs558843609 Chr.3: 51541838-51541838 on Build GRCh38
rs185173513 Chr.3: 51541867-51541867 on Build GRCh38
rs190409815 Chr.3: 51541896-51541896 on Build GRCh38
rs989011542 Chr.3: 51541800-51541800 on Build GRCh38
rs1056423047 Chr.3: 51541646-51541646 on Build GRCh38
rs376157371 Chr.3: 51541841-51541841 on Build GRCh38
rs912119043 Chr.3: 51541843-51541843 on Build GRCh38
rs782507945 Chr.3: 51541791-51541792 on Build GRCh38
rs1011894033 Chr.3: 51541716-51541716 on Build GRCh38
rs1004784222 Chr.3: 51541567-51541567 on Build GRCh38
rs1001054516 Chr.3: 51541781-51541781 on Build GRCh38
rs562671428 Chr.3: 51541943-51541943 on Build GRCh38
rs896416331 Chr.3: 51541684-51541684 on Build GRCh38
rs974677114 Chr.3: 51541882-51541882 on Build GRCh38
rs558158534 Chr.3: 51541773-51541773 on Build GRCh38
rs967787091 Chr.3: 51541735-51541735 on Build GRCh38
rs781786864 Chr.3: 51541788-51541788 on Build GRCh38

View Full Product Details