Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52228532-52229042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TWF2
Entrez Gene ID
11344
Gene Name
twinfilin actin binding protein 2
Gene Aliases
A6RP, A6r, MSTP011, PTK9L
UniGene
Hs.436439
Gene Chromosome Location
Chr.3: 52228610-52239167 on Build GRCh38
Transcripts
NM_007284.3

SNP Detail

SNP ID Chromosome Location
rs1046625997 Chr.3: 52228670-52228670 on Build GRCh38
rs760444803 Chr.3: 52228732-52228732 on Build GRCh38
rs893902043 Chr.3: 52228982-52228982 on Build GRCh38
rs11550254 Chr.3: 52229001-52229001 on Build GRCh38
rs538212590 Chr.3: 52228696-52228696 on Build GRCh38
rs938196904 Chr.3: 52228945-52228945 on Build GRCh38
rs554348352 Chr.3: 52228843-52228843 on Build GRCh38
rs891440059 Chr.3: 52228980-52229001 on Build GRCh38
rs377523936 Chr.3: 52228983-52228983 on Build GRCh38
rs771065754 Chr.3: 52229009-52229009 on Build GRCh38
rs766830110 Chr.3: 52228897-52228897 on Build GRCh38
rs368545300 Chr.3: 52229011-52229011 on Build GRCh38
rs889050186 Chr.3: 52228593-52228593 on Build GRCh38
rs919524376 Chr.3: 52228614-52228614 on Build GRCh38
rs574268261 Chr.3: 52228891-52228891 on Build GRCh38
rs903836268 Chr.3: 52228610-52228610 on Build GRCh38
rs1020881956 Chr.3: 52228765-52228765 on Build GRCh38
rs998037893 Chr.3: 52228813-52228813 on Build GRCh38
rs543271454 Chr.3: 52228958-52228958 on Build GRCh38
rs571674987 Chr.3: 52228737-52228737 on Build GRCh38
rs534030369 Chr.3: 52228755-52228755 on Build GRCh38
rs953892818 Chr.3: 52228838-52228838 on Build GRCh38
rs961391107 Chr.3: 52228881-52228881 on Build GRCh38
rs916902622 Chr.3: 52228894-52228894 on Build GRCh38
rs949646386 Chr.3: 52228898-52228898 on Build GRCh38
rs999606493 Chr.3: 52228686-52228686 on Build GRCh38
rs949594528 Chr.3: 52228667-52228667 on Build GRCh38
rs982354285 Chr.3: 52228908-52228908 on Build GRCh38
rs933031886 Chr.3: 52228915-52228916 on Build GRCh38
rs972412208 Chr.3: 52228884-52228884 on Build GRCh38
rs986382097 Chr.3: 52228962-52228964 on Build GRCh38
rs984314921 Chr.3: 52228839-52228839 on Build GRCh38
rs879366651 Chr.3: 52228833-52228833 on Build GRCh38
rs557883075 Chr.3: 52228729-52228729 on Build GRCh38
rs188643496 Chr.3: 52229017-52229017 on Build GRCh38
rs1036667731 Chr.3: 52228599-52228599 on Build GRCh38
rs1009577811 Chr.3: 52228763-52228763 on Build GRCh38
rs1016694176 Chr.3: 52228875-52228875 on Build GRCh38
rs1054328491 Chr.3: 52228948-52228948 on Build GRCh38
rs1028140147 Chr.3: 52228817-52228817 on Build GRCh38
rs746929756 Chr.3: 52228594-52228595 on Build GRCh38
rs910372043 Chr.3: 52228971-52228971 on Build GRCh38
rs568841901 Chr.3: 52228587-52228587 on Build GRCh38
rs773309849 Chr.3: 52228996-52228996 on Build GRCh38
rs376541927 Chr.3: 52229010-52229010 on Build GRCh38
rs909571450 Chr.3: 52228586-52228586 on Build GRCh38
rs926825011 Chr.3: 52228909-52228909 on Build GRCh38
rs771968990 Chr.3: 52228989-52228989 on Build GRCh38
rs1044145372 Chr.3: 52228605-52228605 on Build GRCh38
rs965282952 Chr.3: 52228784-52228784 on Build GRCh38
rs750716032 Chr.3: 52228744-52228744 on Build GRCh38

View Full Product Details