Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114396297-114396799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM33
Entrez Gene ID
51592
Gene Name
tripartite motif containing 33
Gene Aliases
ECTO, PTC7, RFG7, TF1G, TIF1G, TIF1GAMMA, TIFGAMMA
UniGene
Hs.26837
Gene Chromosome Location
Chr.1: 114392777-114511160 on Build GRCh38
Transcripts
NM_015906.3 NM_033020.2 XM_005270936.3 XM_005270937.3 XM_011541568.2 XM_017001452.1 XM_017001453.1

SNP Detail

SNP ID Chromosome Location
rs970894128 Chr.1: 114396332-114396332 on Build GRCh38
rs41296188 Chr.1: 114396457-114396457 on Build GRCh38
rs1027407254 Chr.1: 114396558-114396558 on Build GRCh38
rs926336971 Chr.1: 114396725-114396725 on Build GRCh38
rs951245949 Chr.1: 114396478-114396478 on Build GRCh38
rs546260484 Chr.1: 114396737-114396737 on Build GRCh38
rs146086735 Chr.1: 114396463-114396463 on Build GRCh38
rs915129531 Chr.1: 114396398-114396398 on Build GRCh38
rs1011530515 Chr.1: 114396363-114396363 on Build GRCh38
rs1041140735 Chr.1: 114396501-114396501 on Build GRCh38
rs754778942 Chr.1: 114396581-114396581 on Build GRCh38
rs980840308 Chr.1: 114396577-114396577 on Build GRCh38
rs981814270 Chr.1: 114396346-114396346 on Build GRCh38
rs550701478 Chr.1: 114396554-114396554 on Build GRCh38
rs952963462 Chr.1: 114396560-114396560 on Build GRCh38
rs946891323 Chr.1: 114396757-114396757 on Build GRCh38
rs970862451 Chr.1: 114396675-114396675 on Build GRCh38
rs115227797 Chr.1: 114396455-114396455 on Build GRCh38
rs1015828820 Chr.1: 114396651-114396651 on Build GRCh38
rs750393089 Chr.1: 114396492-114396494 on Build GRCh38
rs1006656463 Chr.1: 114396405-114396405 on Build GRCh38
rs984385450 Chr.1: 114396514-114396514 on Build GRCh38
rs185696017 Chr.1: 114396437-114396437 on Build GRCh38
rs376690148 Chr.1: 114396520-114396520 on Build GRCh38
rs945301571 Chr.1: 114396436-114396436 on Build GRCh38
rs1004854843 Chr.1: 114396605-114396605 on Build GRCh38
rs899956435 Chr.1: 114396509-114396509 on Build GRCh38
rs948110858 Chr.1: 114396567-114396567 on Build GRCh38
rs926408875 Chr.1: 114396357-114396357 on Build GRCh38
rs535550393 Chr.1: 114396395-114396395 on Build GRCh38
rs567344712 Chr.1: 114396682-114396682 on Build GRCh38
rs529704823 Chr.1: 114396689-114396689 on Build GRCh38
rs937678722 Chr.1: 114396362-114396362 on Build GRCh38
rs1052699288 Chr.1: 114396674-114396674 on Build GRCh38
rs982285183 Chr.1: 114396720-114396720 on Build GRCh38

View Full Product Details