Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 187721627-187722145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL6
Entrez Gene ID
604
Gene Name
B-cell CLL/lymphoma 6
Gene Aliases
BCL5, BCL6A, LAZ3, ZBTB27, ZNF51
UniGene
Hs.478588
Gene Chromosome Location
Chr.3: 187721377-187745725 on Build GRCh38
Transcripts
NM_001130845.1 NM_001134738.1 NM_001706.4 XM_005247694.3 XM_011513062.2 NR_034062.1

Gene Symbol
LOC100131635
Entrez Gene ID
100131635
Gene Name
hCG1645011-like
Gene Aliases
-
UniGene
Hs.434448
Gene Chromosome Location
Chr.3: 187702366-187732415 on Build GRCh38
Transcripts
NM_001130845.1 NM_001134738.1 NM_001706.4 XM_005247694.3 XM_011513062.2 NR_034062.1

SNP Detail

SNP ID Chromosome Location
rs111948250 Chr.3: 187722091-187722091 on Build GRCh38
rs1023218253 Chr.3: 187721988-187721989 on Build GRCh38
rs944843474 Chr.3: 187721941-187721941 on Build GRCh38
rs898016585 Chr.3: 187722020-187722020 on Build GRCh38
rs572053706 Chr.3: 187721767-187721767 on Build GRCh38
rs924499146 Chr.3: 187721816-187721816 on Build GRCh38
rs55885092 Chr.3: 187721959-187721960 on Build GRCh38
rs1010505899 Chr.3: 187721670-187721670 on Build GRCh38
rs1036265781 Chr.3: 187721717-187721718 on Build GRCh38
rs140362025 Chr.3: 187721958-187721959 on Build GRCh38
rs887408015 Chr.3: 187722117-187722117 on Build GRCh38
rs915217248 Chr.3: 187721885-187721885 on Build GRCh38
rs946707205 Chr.3: 187721897-187721897 on Build GRCh38
rs971213810 Chr.3: 187721793-187721793 on Build GRCh38
rs981676408 Chr.3: 187721814-187721814 on Build GRCh38
rs1010536854 Chr.3: 187721946-187721946 on Build GRCh38
rs990086872 Chr.3: 187721849-187721849 on Build GRCh38
rs763229831 Chr.3: 187721739-187721739 on Build GRCh38
rs890275462 Chr.3: 187721901-187721901 on Build GRCh38
rs200152004 Chr.3: 187721974-187721974 on Build GRCh38
rs932049557 Chr.3: 187721862-187721862 on Build GRCh38
rs71684257 Chr.3: 187721968-187721969 on Build GRCh38
rs969076611 Chr.3: 187721686-187721686 on Build GRCh38
rs572073977 Chr.3: 187721735-187721735 on Build GRCh38
rs1050513687 Chr.3: 187721646-187721646 on Build GRCh38
rs898063407 Chr.3: 187721725-187721725 on Build GRCh38
rs1049312661 Chr.3: 187722110-187722110 on Build GRCh38
rs980298462 Chr.3: 187721700-187721700 on Build GRCh38
rs762483484 Chr.3: 187721647-187721647 on Build GRCh38
rs1012589265 Chr.3: 187721742-187721742 on Build GRCh38
rs1040929088 Chr.3: 187721958-187721958 on Build GRCh38
rs530131454 Chr.3: 187721873-187721873 on Build GRCh38
rs1040587531 Chr.3: 187721692-187721692 on Build GRCh38
rs994131158 Chr.3: 187722067-187722067 on Build GRCh38
rs993792113 Chr.3: 187721728-187721728 on Build GRCh38
rs1052801689 Chr.3: 187721931-187721931 on Build GRCh38
rs1009483992 Chr.3: 187721679-187721679 on Build GRCh38
rs34066451 Chr.3: 187721666-187721667 on Build GRCh38
rs762837912 Chr.3: 187722098-187722098 on Build GRCh38
rs1023355967 Chr.3: 187721685-187721685 on Build GRCh38
rs893457125 Chr.3: 187721661-187721661 on Build GRCh38
rs397779935 Chr.3: 187721973-187721974 on Build GRCh38
rs1050463175 Chr.3: 187721908-187721908 on Build GRCh38
rs115678349 Chr.3: 187721823-187721823 on Build GRCh38
rs984090742 Chr.3: 187721761-187721761 on Build GRCh38
rs543937019 Chr.3: 187721909-187721909 on Build GRCh38

View Full Product Details