Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119753189-119753694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGCS2
Entrez Gene ID
3158
Gene Name
3-hydroxy-3-methylglutaryl-CoA synthase 2
Gene Aliases
-
UniGene
Hs.59889
Gene Chromosome Location
Chr.1: 119747996-119769125 on Build GRCh38
Transcripts
NM_001166107.1 NM_005518.3 XM_011541313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1333245 c.1270C>T p.R424* Chr.1: 119753304-119753304 on Build GRCh38
7209994 c.1281G>T p.Q427H Chr.1: 119753293-119753293 on Build GRCh38
4950070 c.1235A>C p.Y412S Chr.1: 119753339-119753339 on Build GRCh38
675017 c.1188C>A p.H396Q Chr.1: 119753386-119753386 on Build GRCh38
3472260 c.1281G>A p.Q427Q Chr.1: 119753293-119753293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775967009 Chr.1: 119753390-119753391 on Build GRCh38
rs756936515 Chr.1: 119753258-119753258 on Build GRCh38
rs1017212754 Chr.1: 119753552-119753552 on Build GRCh38
rs760149713 Chr.1: 119753368-119753368 on Build GRCh38
rs141769037 Chr.1: 119753285-119753285 on Build GRCh38
rs1006215214 Chr.1: 119753534-119753534 on Build GRCh38
rs764933977 Chr.1: 119753400-119753400 on Build GRCh38
rs748245814 Chr.1: 119753435-119753435 on Build GRCh38
rs79425709 Chr.1: 119753479-119753479 on Build GRCh38
rs397981515 Chr.1: 119753447-119753448 on Build GRCh38
rs771851167 Chr.1: 119753546-119753546 on Build GRCh38
rs899771896 Chr.1: 119753558-119753558 on Build GRCh38
rs139620933 Chr.1: 119753357-119753357 on Build GRCh38
rs913112346 Chr.1: 119753249-119753249 on Build GRCh38
rs71074435 Chr.1: 119753411-119753412 on Build GRCh38
rs938468535 Chr.1: 119753240-119753240 on Build GRCh38
rs755119914 Chr.1: 119753287-119753287 on Build GRCh38
rs1009556176 Chr.1: 119753493-119753493 on Build GRCh38
rs79911180 Chr.1: 119753216-119753216 on Build GRCh38
rs878960923 Chr.1: 119753412-119753413 on Build GRCh38
rs113318754 Chr.1: 119753387-119753387 on Build GRCh38
rs71723242 Chr.1: 119753411-119753414 on Build GRCh38
rs774949209 Chr.1: 119753365-119753365 on Build GRCh38
rs745658090 Chr.1: 119753269-119753269 on Build GRCh38
rs142963165 Chr.1: 119753233-119753233 on Build GRCh38
rs912619061 Chr.1: 119753505-119753505 on Build GRCh38
rs1003018671 Chr.1: 119753645-119753645 on Build GRCh38
rs758170788 Chr.1: 119753402-119753402 on Build GRCh38
rs879395346 Chr.1: 119753445-119753448 on Build GRCh38
rs111265001 Chr.1: 119753388-119753388 on Build GRCh38
rs1057366453 Chr.1: 119753242-119753242 on Build GRCh38
rs919915380 Chr.1: 119753443-119753448 on Build GRCh38
rs772305902 Chr.1: 119753304-119753308 on Build GRCh38
rs773032536 Chr.1: 119753413-119753414 on Build GRCh38
rs752151669 Chr.1: 119753231-119753231 on Build GRCh38
rs779938993 Chr.1: 119753295-119753295 on Build GRCh38
rs778510329 Chr.1: 119753261-119753261 on Build GRCh38
rs960788052 Chr.1: 119753590-119753590 on Build GRCh38
rs763443637 Chr.1: 119753386-119753386 on Build GRCh38
rs112856146 Chr.1: 119753278-119753278 on Build GRCh38
rs141951474 Chr.1: 119753614-119753614 on Build GRCh38
rs114432088 Chr.1: 119753565-119753565 on Build GRCh38
rs150665087 Chr.1: 119753637-119753637 on Build GRCh38
rs754042619 Chr.1: 119753510-119753510 on Build GRCh38
rs748252959 Chr.1: 119753303-119753303 on Build GRCh38
rs77063433 Chr.1: 119753485-119753485 on Build GRCh38
rs374922447 Chr.1: 119753430-119753430 on Build GRCh38
rs186771480 Chr.1: 119753302-119753302 on Build GRCh38
rs781446924 Chr.1: 119753444-119753444 on Build GRCh38
rs928012050 Chr.1: 119753574-119753574 on Build GRCh38
rs767212029 Chr.1: 119753412-119753412 on Build GRCh38
rs766216120 Chr.1: 119753414-119753415 on Build GRCh38
rs137852637 Chr.1: 119753304-119753304 on Build GRCh38
rs756771993 Chr.1: 119753397-119753397 on Build GRCh38
rs753399373 Chr.1: 119753393-119753393 on Build GRCh38
rs773364048 Chr.1: 119753338-119753338 on Build GRCh38
rs770705048 Chr.1: 119753448-119753448 on Build GRCh38
rs15609 Chr.1: 119753314-119753314 on Build GRCh38
rs200470161 Chr.1: 119753446-119753446 on Build GRCh38
rs1034584179 Chr.1: 119753377-119753377 on Build GRCh38
rs892123999 Chr.1: 119753595-119753595 on Build GRCh38
rs768676776 Chr.1: 119753298-119753298 on Build GRCh38
rs906268092 Chr.1: 119753640-119753640 on Build GRCh38
rs776773438 Chr.1: 119753632-119753632 on Build GRCh38
rs796293163 Chr.1: 119753447-119753448 on Build GRCh38
rs201676260 Chr.1: 119753451-119753451 on Build GRCh38
rs958929372 Chr.1: 119753398-119753398 on Build GRCh38
rs746911457 Chr.1: 119753407-119753407 on Build GRCh38
rs754916281 Chr.1: 119753418-119753418 on Build GRCh38
rs747000856 Chr.1: 119753297-119753297 on Build GRCh38
rs763378721 Chr.1: 119753344-119753344 on Build GRCh38
rs201053412 Chr.1: 119753239-119753239 on Build GRCh38
rs148155079 Chr.1: 119753572-119753572 on Build GRCh38
rs774177329 Chr.1: 119753411-119753416 on Build GRCh38
rs752547595 Chr.1: 119753414-119753418 on Build GRCh38
rs587667207 Chr.1: 119753513-119753513 on Build GRCh38
rs759698550 Chr.1: 119753636-119753636 on Build GRCh38
rs12117456 Chr.1: 119753381-119753381 on Build GRCh38
rs775960446 Chr.1: 119753410-119753411 on Build GRCh38
rs75221205 Chr.1: 119753412-119753412 on Build GRCh38
rs747793090 Chr.1: 119753407-119753408 on Build GRCh38
rs998270882 Chr.1: 119753473-119753473 on Build GRCh38
rs1039398038 Chr.1: 119753426-119753426 on Build GRCh38
rs766898190 Chr.1: 119753354-119753354 on Build GRCh38
rs779849084 Chr.1: 119753403-119753403 on Build GRCh38
rs971540603 Chr.1: 119753283-119753283 on Build GRCh38
rs377290393 Chr.1: 119753256-119753256 on Build GRCh38
rs750087656 Chr.1: 119753401-119753401 on Build GRCh38
rs74885378 Chr.1: 119753254-119753254 on Build GRCh38
rs765596451 Chr.1: 119753666-119753666 on Build GRCh38

View Full Product Details