Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119808868-119809394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REG4
Entrez Gene ID
83998
Gene Name
regenerating family member 4
Gene Aliases
GISP, REG-IV, RELP
UniGene
Hs.660883
Gene Chromosome Location
Chr.1: 119794018-119811580 on Build GRCh38
Transcripts
NM_001159352.1 NM_001159353.1 NM_032044.3

SNP Detail

SNP ID Chromosome Location
rs993777459 Chr.1: 119809094-119809094 on Build GRCh38
rs1045101071 Chr.1: 119809017-119809017 on Build GRCh38
rs747744952 Chr.1: 119809071-119809071 on Build GRCh38
rs905202088 Chr.1: 119809260-119809260 on Build GRCh38
rs1030063499 Chr.1: 119809095-119809095 on Build GRCh38
rs967934398 Chr.1: 119808936-119808936 on Build GRCh38
rs1057057627 Chr.1: 119809200-119809200 on Build GRCh38
rs776670109 Chr.1: 119809347-119809347 on Build GRCh38
rs745487900 Chr.1: 119809038-119809038 on Build GRCh38
rs1021198710 Chr.1: 119808917-119808917 on Build GRCh38
rs896604134 Chr.1: 119808998-119808998 on Build GRCh38
rs929041675 Chr.1: 119809105-119809105 on Build GRCh38
rs897759771 Chr.1: 119809325-119809325 on Build GRCh38
rs925562048 Chr.1: 119809202-119809202 on Build GRCh38
rs949463891 Chr.1: 119809223-119809223 on Build GRCh38
rs925479306 Chr.1: 119808913-119808913 on Build GRCh38
rs935512249 Chr.1: 119809209-119809209 on Build GRCh38
rs1016476447 Chr.1: 119809056-119809056 on Build GRCh38
rs909353734 Chr.1: 119809144-119809144 on Build GRCh38
rs771719074 Chr.1: 119809125-119809125 on Build GRCh38
rs772990268 Chr.1: 119809178-119809178 on Build GRCh38
rs942181312 Chr.1: 119809145-119809145 on Build GRCh38
rs587613378 Chr.1: 119809293-119809293 on Build GRCh38
rs2994809 Chr.1: 119809354-119809354 on Build GRCh38
rs949351082 Chr.1: 119809015-119809015 on Build GRCh38
rs988742345 Chr.1: 119809216-119809216 on Build GRCh38
rs187512546 Chr.1: 119809024-119809024 on Build GRCh38
rs1010722032 Chr.1: 119809254-119809254 on Build GRCh38
rs757167458 Chr.1: 119809283-119809283 on Build GRCh38
rs587721527 Chr.1: 119809332-119809332 on Build GRCh38
rs765358000 Chr.1: 119809369-119809369 on Build GRCh38
rs954491920 Chr.1: 119809103-119809103 on Build GRCh38
rs587745263 Chr.1: 119809261-119809261 on Build GRCh38
rs1000669861 Chr.1: 119809341-119809341 on Build GRCh38
rs979323308 Chr.1: 119808949-119808949 on Build GRCh38
rs769324794 Chr.1: 119809045-119809045 on Build GRCh38
rs1052524784 Chr.1: 119808994-119808994 on Build GRCh38
rs587732581 Chr.1: 119809073-119809073 on Build GRCh38
rs906089388 Chr.1: 119809184-119809184 on Build GRCh38
rs897649856 Chr.1: 119809091-119809091 on Build GRCh38
rs910446327 Chr.1: 119808958-119808958 on Build GRCh38
rs746880371 Chr.1: 119809221-119809221 on Build GRCh38
rs1017243175 Chr.1: 119809141-119809141 on Build GRCh38
rs891828907 Chr.1: 119809222-119809222 on Build GRCh38
rs964633773 Chr.1: 119808975-119808975 on Build GRCh38
rs587606190 Chr.1: 119809065-119809066 on Build GRCh38
rs17838168 Chr.1: 119809188-119809188 on Build GRCh38
rs587608983 Chr.1: 119809126-119809126 on Build GRCh38

View Full Product Details