Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 40811238-40811756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB2
Entrez Gene ID
323
Gene Name
amyloid beta precursor protein binding family B member 2
Gene Aliases
FE65L, FE65L1
UniGene
Hs.479602
Gene Chromosome Location
Chr.4: 40810027-41216714 on Build GRCh38
Transcripts
NM_001166050.1 NM_001166051.1 NM_001166052.1 NM_001166053.1 NM_001166054.1 NM_004307.1 NM_173075.4 XM_005248101.3 XM_006714005.3 XM_006714007.3 XM_006714008.3 XM_006714009.3 XM_006714010.3 XM_006714011.3 XM_006714012.3 XM_011513686.2 XM_011513687.2 XM_017008142.1 XM_017008143.1 XM_017008144.1 XM_017008145.1 XM_017008146.1 XM_017008147.1 XM_017008148.1 XM_017008149.1 XM_017008150.1 XM_017008151.1 XM_017008152.1 XM_017008153.1 XM_017008154.1 XM_017008155.1 XM_017008156.1 XM_017008157.1 XM_017008158.1

SNP Detail

SNP ID Chromosome Location
rs35789166 Chr.4: 40811374-40811374 on Build GRCh38
rs1025920731 Chr.4: 40811591-40811591 on Build GRCh38
rs958209047 Chr.4: 40811558-40811558 on Build GRCh38
rs776813039 Chr.4: 40811668-40811668 on Build GRCh38
rs966927747 Chr.4: 40811503-40811503 on Build GRCh38
rs966957140 Chr.4: 40811343-40811343 on Build GRCh38
rs925491329 Chr.4: 40811352-40811352 on Build GRCh38
rs373308061 Chr.4: 40811524-40811524 on Build GRCh38
rs557149459 Chr.4: 40811328-40811328 on Build GRCh38
rs963018019 Chr.4: 40811305-40811306 on Build GRCh38
rs796558551 Chr.4: 40811546-40811549 on Build GRCh38
rs201921233 Chr.4: 40811545-40811545 on Build GRCh38
rs375872647 Chr.4: 40811455-40811455 on Build GRCh38
rs546358201 Chr.4: 40811380-40811380 on Build GRCh38
rs922019811 Chr.4: 40811570-40811570 on Build GRCh38
rs578147078 Chr.4: 40811650-40811651 on Build GRCh38
rs530984102 Chr.4: 40811648-40811648 on Build GRCh38
rs71276528 Chr.4: 40811524-40811533 on Build GRCh38
rs573277335 Chr.4: 40811447-40811447 on Build GRCh38
rs980235142 Chr.4: 40811637-40811637 on Build GRCh38
rs376987355 Chr.4: 40811525-40811525 on Build GRCh38
rs878943565 Chr.4: 40811487-40811487 on Build GRCh38
rs555640473 Chr.4: 40811305-40811305 on Build GRCh38
rs1033723470 Chr.4: 40811324-40811324 on Build GRCh38
rs377404138 Chr.4: 40811651-40811652 on Build GRCh38
rs372440375 Chr.4: 40811555-40811558 on Build GRCh38
rs1036863549 Chr.4: 40811419-40811419 on Build GRCh38
rs142994993 Chr.4: 40811512-40811513 on Build GRCh38
rs926156583 Chr.4: 40811369-40811369 on Build GRCh38
rs890937922 Chr.4: 40811286-40811286 on Build GRCh38
rs933428078 Chr.4: 40811613-40811613 on Build GRCh38
rs369117672 Chr.4: 40811473-40811473 on Build GRCh38
rs1052220609 Chr.4: 40811375-40811375 on Build GRCh38
rs988787088 Chr.4: 40811388-40811388 on Build GRCh38
rs1025244458 Chr.4: 40811338-40811338 on Build GRCh38
rs1025090389 Chr.4: 40811484-40811484 on Build GRCh38
rs368982838 Chr.4: 40811528-40811528 on Build GRCh38
rs200451977 Chr.4: 40811531-40811533 on Build GRCh38
rs36033290 Chr.4: 40811521-40811522 on Build GRCh38
rs1024434095 Chr.4: 40811616-40811616 on Build GRCh38
rs538063034 Chr.4: 40811326-40811326 on Build GRCh38
rs898333116 Chr.4: 40811423-40811423 on Build GRCh38
rs774467397 Chr.4: 40811557-40811557 on Build GRCh38
rs898239818 Chr.4: 40811299-40811299 on Build GRCh38
rs188650494 Chr.4: 40811711-40811711 on Build GRCh38
rs373450155 Chr.4: 40811675-40811675 on Build GRCh38
rs1036957479 Chr.4: 40811705-40811705 on Build GRCh38
rs796276598 Chr.4: 40811357-40811357 on Build GRCh38
rs1004028084 Chr.4: 40811288-40811288 on Build GRCh38
rs34759176 Chr.4: 40811653-40811654 on Build GRCh38
rs77009651 Chr.4: 40811523-40811524 on Build GRCh38
rs768739819 Chr.4: 40811403-40811403 on Build GRCh38
rs974859373 Chr.4: 40811358-40811358 on Build GRCh38
rs992792045 Chr.4: 40811329-40811329 on Build GRCh38
rs192375253 Chr.4: 40811351-40811351 on Build GRCh38
rs11731472 Chr.4: 40811697-40811697 on Build GRCh38
rs994482762 Chr.4: 40811568-40811568 on Build GRCh38
rs970257865 Chr.4: 40811636-40811636 on Build GRCh38
rs1028866324 Chr.4: 40811331-40811331 on Build GRCh38
rs78199801 Chr.4: 40811562-40811562 on Build GRCh38
rs144663450 Chr.4: 40811414-40811414 on Build GRCh38
rs536798440 Chr.4: 40811467-40811467 on Build GRCh38
rs372515901 Chr.4: 40811634-40811635 on Build GRCh38
rs148066166 Chr.4: 40811520-40811521 on Build GRCh38
rs35774490 Chr.4: 40811546-40811546 on Build GRCh38
rs988839329 Chr.4: 40811662-40811662 on Build GRCh38
rs187702582 Chr.4: 40811307-40811307 on Build GRCh38
rs376394352 Chr.4: 40811329-40811329 on Build GRCh38
rs995628742 Chr.4: 40811431-40811431 on Build GRCh38
rs1033281110 Chr.4: 40811645-40811646 on Build GRCh38
rs142802404 Chr.4: 40811701-40811701 on Build GRCh38
rs894781107 Chr.4: 40811459-40811459 on Build GRCh38
rs553166683 Chr.4: 40811508-40811508 on Build GRCh38
rs6447449 Chr.4: 40811293-40811293 on Build GRCh38
rs921895704 Chr.4: 40811370-40811370 on Build GRCh38

View Full Product Details