Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 48490502-48490990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZAR1
Entrez Gene ID
326340
Gene Name
zygote arrest 1
Gene Aliases
Z3CXXC6
UniGene
Hs.276457
Gene Chromosome Location
Chr.4: 48490252-48497528 on Build GRCh38
Transcripts
NM_175619.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1739059 c.641G>T p.G214V Chr.4: 48490932-48490932 on Build GRCh38
4964386 c.537C>T p.V179V Chr.4: 48490828-48490828 on Build GRCh38
4826461 c.509C>T p.P170L Chr.4: 48490800-48490800 on Build GRCh38
4964972 c.345C>T p.D115D Chr.4: 48490636-48490636 on Build GRCh38
6367782 c.289C>T p.R97C Chr.4: 48490580-48490580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767554974 Chr.4: 48490854-48490854 on Build GRCh38
rs1016453403 Chr.4: 48490936-48490936 on Build GRCh38
rs778371879 Chr.4: 48490890-48490890 on Build GRCh38
rs751380832 Chr.4: 48490521-48490521 on Build GRCh38
rs747315015 Chr.4: 48490834-48490834 on Build GRCh38
rs781073506 Chr.4: 48490735-48490735 on Build GRCh38
rs868427965 Chr.4: 48490937-48490937 on Build GRCh38
rs914079772 Chr.4: 48490870-48490870 on Build GRCh38
rs377660065 Chr.4: 48490846-48490846 on Build GRCh38
rs151152950 Chr.4: 48490644-48490644 on Build GRCh38
rs769255444 Chr.4: 48490561-48490561 on Build GRCh38
rs1032146442 Chr.4: 48490729-48490729 on Build GRCh38
rs1015925871 Chr.4: 48490783-48490783 on Build GRCh38
rs767195551 Chr.4: 48490770-48490770 on Build GRCh38
rs781719259 Chr.4: 48490915-48490915 on Build GRCh38
rs562142882 Chr.4: 48490677-48490677 on Build GRCh38
rs117545505 Chr.4: 48490705-48490705 on Build GRCh38
rs760404099 Chr.4: 48490874-48490874 on Build GRCh38
rs746057544 Chr.4: 48490917-48490917 on Build GRCh38
rs773254829 Chr.4: 48490634-48490634 on Build GRCh38
rs575207390 Chr.4: 48490638-48490638 on Build GRCh38
rs779226242 Chr.4: 48490748-48490748 on Build GRCh38
rs139073114 Chr.4: 48490879-48490879 on Build GRCh38
rs992257572 Chr.4: 48490857-48490857 on Build GRCh38
rs777001580 Chr.4: 48490766-48490766 on Build GRCh38
rs750274501 Chr.4: 48490796-48490796 on Build GRCh38
rs908881418 Chr.4: 48490586-48490586 on Build GRCh38
rs765948384 Chr.4: 48490808-48490808 on Build GRCh38
rs373521806 Chr.4: 48490822-48490822 on Build GRCh38
rs755756974 Chr.4: 48490807-48490807 on Build GRCh38
rs763874668 Chr.4: 48490707-48490707 on Build GRCh38
rs1047558006 Chr.4: 48490905-48490905 on Build GRCh38
rs950468099 Chr.4: 48490565-48490565 on Build GRCh38
rs543756820 Chr.4: 48490939-48490939 on Build GRCh38
rs768349689 Chr.4: 48490774-48490775 on Build GRCh38
rs752137624 Chr.4: 48490906-48490906 on Build GRCh38
rs913963717 Chr.4: 48490656-48490656 on Build GRCh38
rs752322275 Chr.4: 48490886-48490886 on Build GRCh38
rs981467553 Chr.4: 48490604-48490604 on Build GRCh38
rs771440149 Chr.4: 48490841-48490841 on Build GRCh38
rs901659096 Chr.4: 48490736-48490736 on Build GRCh38
rs770113421 Chr.4: 48490931-48490931 on Build GRCh38
rs1005959702 Chr.4: 48490921-48490921 on Build GRCh38
rs878912022 Chr.4: 48490540-48490540 on Build GRCh38
rs754696963 Chr.4: 48490883-48490883 on Build GRCh38
rs745542393 Chr.4: 48490552-48490552 on Build GRCh38
rs746928898 Chr.4: 48490686-48490687 on Build GRCh38
rs776944574 Chr.4: 48490842-48490842 on Build GRCh38
rs778512611 Chr.4: 48490811-48490811 on Build GRCh38
rs775688984 Chr.4: 48490849-48490849 on Build GRCh38
rs74929644 Chr.4: 48490752-48490752 on Build GRCh38
rs73144650 Chr.4: 48490529-48490529 on Build GRCh38
rs529585784 Chr.4: 48490703-48490703 on Build GRCh38
rs773137561 Chr.4: 48490871-48490871 on Build GRCh38
rs905508301 Chr.4: 48490943-48490943 on Build GRCh38
rs926801749 Chr.4: 48490612-48490612 on Build GRCh38
rs747223329 Chr.4: 48490759-48490759 on Build GRCh38
rs570144212 Chr.4: 48490825-48490825 on Build GRCh38
rs774116026 Chr.4: 48490769-48490769 on Build GRCh38
rs1039392591 Chr.4: 48490711-48490711 on Build GRCh38
rs772281714 Chr.4: 48490749-48490749 on Build GRCh38
rs1056453628 Chr.4: 48490637-48490637 on Build GRCh38
rs1035331424 Chr.4: 48490960-48490960 on Build GRCh38
rs754521056 Chr.4: 48490747-48490747 on Build GRCh38
rs945477385 Chr.4: 48490665-48490665 on Build GRCh38
rs960910778 Chr.4: 48490962-48490962 on Build GRCh38
rs778280300 Chr.4: 48490832-48490832 on Build GRCh38
rs769208107 Chr.4: 48490574-48490574 on Build GRCh38
rs1001582578 Chr.4: 48490953-48490953 on Build GRCh38
rs774772725 Chr.4: 48490594-48490594 on Build GRCh38
rs933234525 Chr.4: 48490898-48490898 on Build GRCh38
rs757853076 Chr.4: 48490889-48490889 on Build GRCh38
rs771348437 Chr.4: 48490760-48490760 on Build GRCh38
rs1043697034 Chr.4: 48490691-48490691 on Build GRCh38
rs1044233478 Chr.4: 48490882-48490882 on Build GRCh38
rs549200712 Chr.4: 48490884-48490884 on Build GRCh38
rs748863509 Chr.4: 48490746-48490746 on Build GRCh38
rs992372425 Chr.4: 48490964-48490964 on Build GRCh38
rs751529400 Chr.4: 48490713-48490713 on Build GRCh38
rs756954053 Chr.4: 48490718-48490718 on Build GRCh38
rs559352128 Chr.4: 48490737-48490737 on Build GRCh38
rs755811979 Chr.4: 48490744-48490744 on Build GRCh38
rs759634347 Chr.4: 48490767-48490767 on Build GRCh38
rs757656411 Chr.4: 48490910-48490910 on Build GRCh38
rs779580676 Chr.4: 48490562-48490562 on Build GRCh38
rs772495951 Chr.4: 48490611-48490611 on Build GRCh38
rs370900350 Chr.4: 48490853-48490853 on Build GRCh38
rs753500934 Chr.4: 48490810-48490810 on Build GRCh38
rs901734089 Chr.4: 48490892-48490892 on Build GRCh38
rs563410624 Chr.4: 48490566-48490566 on Build GRCh38
rs567453019 Chr.4: 48490888-48490888 on Build GRCh38

View Full Product Details