Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 147906606-147907110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP10
Entrez Gene ID
79658
Gene Name
Rho GTPase activating protein 10
Gene Aliases
GRAF2, PS-GAP, PSGAP
UniGene
Hs.368631
Gene Chromosome Location
Chr.4: 147732063-148072781 on Build GRCh38
Transcripts
NM_024605.3 XM_005263215.3 XM_017008602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1183350 c.1042G>A p.V348I Chr.4: 147906645-147906645 on Build GRCh38
236022 c.1057C>A p.Q353K Chr.4: 147906660-147906660 on Build GRCh38
4458775 c.1099C>T p.L367L Chr.4: 147906702-147906702 on Build GRCh38
3600781 c.1067C>T p.S356F Chr.4: 147906670-147906670 on Build GRCh38
4122633 c.1092G>A p.L364L Chr.4: 147906695-147906695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938338038 Chr.4: 147906672-147906672 on Build GRCh38
rs200935817 Chr.4: 147906649-147906649 on Build GRCh38
rs57726071 Chr.4: 147906979-147906979 on Build GRCh38
rs144321012 Chr.4: 147906957-147906957 on Build GRCh38
rs745393301 Chr.4: 147906720-147906720 on Build GRCh38
rs768996160 Chr.4: 147906631-147906631 on Build GRCh38
rs551810354 Chr.4: 147906898-147906898 on Build GRCh38
rs146090111 Chr.4: 147907078-147907078 on Build GRCh38
rs762949964 Chr.4: 147906761-147906761 on Build GRCh38
rs1006159115 Chr.4: 147906882-147906882 on Build GRCh38
rs980227720 Chr.4: 147906897-147906897 on Build GRCh38
rs971909653 Chr.4: 147906946-147906946 on Build GRCh38
rs757230994 Chr.4: 147906707-147906707 on Build GRCh38
rs866084772 Chr.4: 147906815-147906815 on Build GRCh38
rs571754730 Chr.4: 147907068-147907068 on Build GRCh38
rs757865628 Chr.4: 147906726-147906726 on Build GRCh38
rs11099672 Chr.4: 147906822-147906822 on Build GRCh38
rs483352831 Chr.4: 147906740-147906740 on Build GRCh38
rs912931329 Chr.4: 147906929-147906929 on Build GRCh38
rs766526062 Chr.4: 147906658-147906658 on Build GRCh38
rs547390854 Chr.4: 147906827-147906827 on Build GRCh38
rs914791622 Chr.4: 147906997-147906997 on Build GRCh38
rs748691472 Chr.4: 147906746-147906746 on Build GRCh38
rs563860915 Chr.4: 147906657-147906657 on Build GRCh38
rs969929135 Chr.4: 147906892-147906892 on Build GRCh38
rs997468913 Chr.4: 147906826-147906826 on Build GRCh38
rs1026306686 Chr.4: 147906843-147906843 on Build GRCh38
rs542295536 Chr.4: 147906710-147906710 on Build GRCh38
rs558333599 Chr.4: 147906988-147906988 on Build GRCh38
rs61748165 Chr.4: 147906644-147906644 on Build GRCh38
rs756616736 Chr.4: 147906645-147906645 on Build GRCh38
rs910271514 Chr.4: 147906816-147906816 on Build GRCh38
rs199714695 Chr.4: 147906762-147906762 on Build GRCh38
rs763835743 Chr.4: 147906770-147906770 on Build GRCh38
rs763938835 Chr.4: 147906702-147906702 on Build GRCh38
rs77276930 Chr.4: 147906908-147906908 on Build GRCh38
rs747931937 Chr.4: 147906952-147906952 on Build GRCh38
rs1044272220 Chr.4: 147906739-147906739 on Build GRCh38
rs1023065781 Chr.4: 147906944-147906944 on Build GRCh38
rs752148813 Chr.4: 147907033-147907033 on Build GRCh38
rs943551357 Chr.4: 147907011-147907011 on Build GRCh38
rs10049932 Chr.4: 147906782-147906782 on Build GRCh38
rs374709295 Chr.4: 147906643-147906643 on Build GRCh38
rs267600033 Chr.4: 147906650-147906650 on Build GRCh38
rs752989314 Chr.4: 147906683-147906685 on Build GRCh38
rs531394218 Chr.4: 147906736-147906736 on Build GRCh38
rs774497035 Chr.4: 147906768-147906768 on Build GRCh38
rs776953685 Chr.4: 147906742-147906742 on Build GRCh38
rs770312389 Chr.4: 147906755-147906755 on Build GRCh38
rs759783652 Chr.4: 147906769-147906769 on Build GRCh38
rs148015211 Chr.4: 147906699-147906699 on Build GRCh38
rs540570241 Chr.4: 147906642-147906642 on Build GRCh38
rs11734553 Chr.4: 147906883-147906883 on Build GRCh38
rs139674812 Chr.4: 147906733-147906733 on Build GRCh38
rs777068765 Chr.4: 147906639-147906639 on Build GRCh38
rs570106376 Chr.4: 147906913-147906913 on Build GRCh38
rs371799366 Chr.4: 147906671-147906671 on Build GRCh38
rs946325277 Chr.4: 147906812-147906812 on Build GRCh38
rs772974691 Chr.4: 147907037-147907037 on Build GRCh38
rs377129201 Chr.4: 147906757-147906757 on Build GRCh38
rs779541539 Chr.4: 147906846-147906846 on Build GRCh38
rs924783372 Chr.4: 147906763-147906763 on Build GRCh38
rs1039216629 Chr.4: 147907031-147907031 on Build GRCh38
rs1002943411 Chr.4: 147906950-147906950 on Build GRCh38
rs967725545 Chr.4: 147907082-147907082 on Build GRCh38
rs925408228 Chr.4: 147907002-147907002 on Build GRCh38
rs149405506 Chr.4: 147906640-147906640 on Build GRCh38

View Full Product Details