Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 144207055-144207563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD16
Entrez Gene ID
57528
Gene Name
potassium channel tetramerization domain containing 16
Gene Aliases
-
UniGene
Hs.661870
Gene Chromosome Location
Chr.5: 144170723-144485686 on Build GRCh38
Transcripts
NM_020768.3 XM_005268492.2 XM_005268493.2 XM_006714789.2 XM_011537671.2 XM_011537672.2 XM_011537673.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4841200 c.446G>T p.R149I Chr.5: 144207160-144207160 on Build GRCh38
6489559 c.502G>T p.V168L Chr.5: 144207216-144207216 on Build GRCh38
3852347 c.597C>A p.S199S Chr.5: 144207311-144207311 on Build GRCh38
334624 c.607G>C p.E203Q Chr.5: 144207321-144207321 on Build GRCh38
3852349 c.832+1G>A p.? Chr.5: 144207547-144207547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148158462 Chr.5: 144207192-144207192 on Build GRCh38
rs748673825 Chr.5: 144207498-144207498 on Build GRCh38
rs147117374 Chr.5: 144207152-144207152 on Build GRCh38
rs753433220 Chr.5: 144207195-144207195 on Build GRCh38
rs750638083 Chr.5: 144207164-144207164 on Build GRCh38
rs900201583 Chr.5: 144207312-144207312 on Build GRCh38
rs773482773 Chr.5: 144207441-144207441 on Build GRCh38
rs150713689 Chr.5: 144207459-144207459 on Build GRCh38
rs898632679 Chr.5: 144207488-144207488 on Build GRCh38
rs141163654 Chr.5: 144207425-144207425 on Build GRCh38
rs754826888 Chr.5: 144207187-144207187 on Build GRCh38
rs371601238 Chr.5: 144207504-144207504 on Build GRCh38
rs747584875 Chr.5: 144207237-144207237 on Build GRCh38
rs117464471 Chr.5: 144207119-144207119 on Build GRCh38
rs746212035 Chr.5: 144207220-144207220 on Build GRCh38
rs932967948 Chr.5: 144207335-144207335 on Build GRCh38
rs947127517 Chr.5: 144207283-144207283 on Build GRCh38
rs761889328 Chr.5: 144207263-144207263 on Build GRCh38
rs779107130 Chr.5: 144207206-144207206 on Build GRCh38
rs750023878 Chr.5: 144207472-144207472 on Build GRCh38
rs774793246 Chr.5: 144207161-144207161 on Build GRCh38
rs776987268 Chr.5: 144207174-144207174 on Build GRCh38
rs139113029 Chr.5: 144207489-144207489 on Build GRCh38
rs751314984 Chr.5: 144207437-144207437 on Build GRCh38
rs760940703 Chr.5: 144207089-144207089 on Build GRCh38
rs374426613 Chr.5: 144207138-144207138 on Build GRCh38
rs766798809 Chr.5: 144207476-144207476 on Build GRCh38
rs769305903 Chr.5: 144207414-144207414 on Build GRCh38
rs770082687 Chr.5: 144207266-144207266 on Build GRCh38
rs757998131 Chr.5: 144207473-144207473 on Build GRCh38
rs200806853 Chr.5: 144207191-144207191 on Build GRCh38
rs780514964 Chr.5: 144207135-144207135 on Build GRCh38
rs373669684 Chr.5: 144207296-144207296 on Build GRCh38
rs761373442 Chr.5: 144207469-144207469 on Build GRCh38
rs367855290 Chr.5: 144207482-144207482 on Build GRCh38
rs747458169 Chr.5: 144207412-144207412 on Build GRCh38
rs751384415 Chr.5: 144207127-144207127 on Build GRCh38
rs759366932 Chr.5: 144207293-144207293 on Build GRCh38
rs767471123 Chr.5: 144207190-144207190 on Build GRCh38
rs774598298 Chr.5: 144207533-144207533 on Build GRCh38
rs771175116 Chr.5: 144207517-144207517 on Build GRCh38
rs749481448 Chr.5: 144207509-144207509 on Build GRCh38
rs758252977 Chr.5: 144207116-144207116 on Build GRCh38
rs865986167 Chr.5: 144207111-144207111 on Build GRCh38
rs754953427 Chr.5: 144207128-144207128 on Build GRCh38
rs142447175 Chr.5: 144207500-144207500 on Build GRCh38
rs771798745 Chr.5: 144207446-144207446 on Build GRCh38
rs963138337 Chr.5: 144207125-144207125 on Build GRCh38
rs752567922 Chr.5: 144207297-144207297 on Build GRCh38
rs35597558 Chr.5: 144207285-144207286 on Build GRCh38
rs764720574 Chr.5: 144207470-144207470 on Build GRCh38
rs780567617 Chr.5: 144207408-144207408 on Build GRCh38
rs769825888 Chr.5: 144207440-144207440 on Build GRCh38
rs201451163 Chr.5: 144207086-144207086 on Build GRCh38
rs756957270 Chr.5: 144207196-144207196 on Build GRCh38
rs772665965 Chr.5: 144207167-144207167 on Build GRCh38
rs776180589 Chr.5: 144207171-144207171 on Build GRCh38
rs778606553 Chr.5: 144207348-144207348 on Build GRCh38
rs763548397 Chr.5: 144207313-144207313 on Build GRCh38
rs776862453 Chr.5: 144207255-144207255 on Build GRCh38
rs773343153 Chr.5: 144207273-144207273 on Build GRCh38
rs764325824 Chr.5: 144207172-144207172 on Build GRCh38
rs760621216 Chr.5: 144207305-144207305 on Build GRCh38
rs544361716 Chr.5: 144207369-144207369 on Build GRCh38
rs753768585 Chr.5: 144207466-144207466 on Build GRCh38
rs753379870 Chr.5: 144207318-144207318 on Build GRCh38
rs199689235 Chr.5: 144207175-144207175 on Build GRCh38
rs768786960 Chr.5: 144207242-144207242 on Build GRCh38
rs748223261 Chr.5: 144207430-144207430 on Build GRCh38
rs750101809 Chr.5: 144207364-144207364 on Build GRCh38
rs766026967 Chr.5: 144207178-144207178 on Build GRCh38
rs143360365 Chr.5: 144207140-144207140 on Build GRCh38
rs763092682 Chr.5: 144207445-144207445 on Build GRCh38
rs752000601 Chr.5: 144207480-144207480 on Build GRCh38
rs754904967 Chr.5: 144207458-144207458 on Build GRCh38
rs751388398 Chr.5: 144207183-144207183 on Build GRCh38
rs1051368863 Chr.5: 144207371-144207371 on Build GRCh38
rs777394482 Chr.5: 144207142-144207142 on Build GRCh38
rs775287854 Chr.5: 144207453-144207453 on Build GRCh38
rs763107933 Chr.5: 144207166-144207166 on Build GRCh38
rs761877984 Chr.5: 144207088-144207088 on Build GRCh38
rs780692300 Chr.5: 144207229-144207229 on Build GRCh38
rs765341890 Chr.5: 144207110-144207110 on Build GRCh38
rs756758031 Chr.5: 144207320-144207320 on Build GRCh38
rs371296124 Chr.5: 144207225-144207225 on Build GRCh38
rs143356794 Chr.5: 144207153-144207153 on Build GRCh38
rs988316029 Chr.5: 144207203-144207203 on Build GRCh38

View Full Product Details