Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 144473571-144474067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD16
Entrez Gene ID
57528
Gene Name
potassium channel tetramerization domain containing 16
Gene Aliases
-
UniGene
Hs.661870
Gene Chromosome Location
Chr.5: 144170723-144485686 on Build GRCh38
Transcripts
NM_020768.3 XM_005268492.2 XM_005268493.2 XM_006714789.2 XM_011537671.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3209818 c.1141A>C p.K381Q Chr.5: 144473968-144473968 on Build GRCh38
5633379 c.872G>T p.C291F Chr.5: 144473699-144473699 on Build GRCh38
1211682 c.888A>C p.K296N Chr.5: 144473715-144473715 on Build GRCh38
4920634 c.870C>G p.C290W Chr.5: 144473697-144473697 on Build GRCh38
6513434 c.885C>A p.G295G Chr.5: 144473712-144473712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867978847 Chr.5: 144474001-144474001 on Build GRCh38
rs764587697 Chr.5: 144473831-144473831 on Build GRCh38
rs745350480 Chr.5: 144473926-144473926 on Build GRCh38
rs770198057 Chr.5: 144474042-144474042 on Build GRCh38
rs147489800 Chr.5: 144473955-144473955 on Build GRCh38
rs1023797281 Chr.5: 144473706-144473706 on Build GRCh38
rs748487086 Chr.5: 144474039-144474039 on Build GRCh38
rs372522368 Chr.5: 144473741-144473741 on Build GRCh38
rs373514581 Chr.5: 144473965-144473965 on Build GRCh38
rs753193056 Chr.5: 144473613-144473613 on Build GRCh38
rs113240704 Chr.5: 144473745-144473745 on Build GRCh38
rs773239066 Chr.5: 144473728-144473728 on Build GRCh38
rs368969663 Chr.5: 144473959-144473959 on Build GRCh38
rs778983148 Chr.5: 144473985-144473985 on Build GRCh38
rs780115075 Chr.5: 144473882-144473882 on Build GRCh38
rs1008551266 Chr.5: 144473595-144473595 on Build GRCh38
rs753037349 Chr.5: 144473747-144473747 on Build GRCh38
rs775001984 Chr.5: 144473927-144473927 on Build GRCh38
rs905285392 Chr.5: 144473945-144473945 on Build GRCh38
rs777554566 Chr.5: 144473795-144473795 on Build GRCh38
rs762923781 Chr.5: 144473953-144473953 on Build GRCh38
rs201201619 Chr.5: 144473997-144473997 on Build GRCh38
rs775259127 Chr.5: 144473824-144473824 on Build GRCh38
rs754275862 Chr.5: 144473841-144473841 on Build GRCh38
rs763261967 Chr.5: 144473784-144473784 on Build GRCh38
rs568179987 Chr.5: 144473738-144473738 on Build GRCh38
rs879003392 Chr.5: 144473915-144473915 on Build GRCh38
rs374565442 Chr.5: 144473916-144473916 on Build GRCh38
rs755853865 Chr.5: 144473901-144473901 on Build GRCh38
rs370037481 Chr.5: 144473867-144473867 on Build GRCh38
rs1027204536 Chr.5: 144473739-144473739 on Build GRCh38
rs777002557 Chr.5: 144473941-144473941 on Build GRCh38
rs141010798 Chr.5: 144473992-144473992 on Build GRCh38
rs776301270 Chr.5: 144473692-144473692 on Build GRCh38
rs201108801 Chr.5: 144474007-144474007 on Build GRCh38
rs570566784 Chr.5: 144473904-144473904 on Build GRCh38
rs142965087 Chr.5: 144473742-144473742 on Build GRCh38
rs746593076 Chr.5: 144474004-144474004 on Build GRCh38
rs774084184 Chr.5: 144473740-144473740 on Build GRCh38
rs1013701218 Chr.5: 144473663-144473663 on Build GRCh38
rs778967528 Chr.5: 144473772-144473772 on Build GRCh38
rs766232634 Chr.5: 144473844-144473844 on Build GRCh38
rs760934349 Chr.5: 144473830-144473830 on Build GRCh38
rs749299224 Chr.5: 144473635-144473635 on Build GRCh38
rs148474129 Chr.5: 144473807-144473807 on Build GRCh38
rs757167871 Chr.5: 144473977-144473977 on Build GRCh38
rs762320309 Chr.5: 144473856-144473856 on Build GRCh38
rs931777861 Chr.5: 144473903-144473903 on Build GRCh38
rs771025690 Chr.5: 144473909-144473909 on Build GRCh38
rs758569494 Chr.5: 144473774-144473774 on Build GRCh38
rs750531391 Chr.5: 144473873-144473873 on Build GRCh38
rs758542754 Chr.5: 144473877-144473877 on Build GRCh38
rs113526376 Chr.5: 144473962-144473962 on Build GRCh38
rs568390179 Chr.5: 144473621-144473621 on Build GRCh38
rs754856759 Chr.5: 144473967-144473968 on Build GRCh38
rs763943596 Chr.5: 144473964-144473964 on Build GRCh38
rs371767006 Chr.5: 144474002-144474002 on Build GRCh38
rs918707256 Chr.5: 144473908-144473908 on Build GRCh38
rs867136933 Chr.5: 144473949-144473949 on Build GRCh38
rs765700501 Chr.5: 144473952-144473952 on Build GRCh38
rs746155133 Chr.5: 144473685-144473685 on Build GRCh38
rs751862752 Chr.5: 144473884-144473884 on Build GRCh38
rs766543181 Chr.5: 144473954-144473954 on Build GRCh38
rs753765917 Chr.5: 144473968-144473968 on Build GRCh38
rs935283310 Chr.5: 144473911-144473911 on Build GRCh38
rs941350605 Chr.5: 144473859-144473859 on Build GRCh38
rs762266741 Chr.5: 144473947-144473947 on Build GRCh38
rs146541436 Chr.5: 144473998-144473998 on Build GRCh38
rs748067897 Chr.5: 144473717-144473717 on Build GRCh38
rs757377226 Chr.5: 144473661-144473661 on Build GRCh38
rs777871647 Chr.5: 144473625-144473625 on Build GRCh38
rs139920765 Chr.5: 144473737-144473737 on Build GRCh38
rs759227443 Chr.5: 144473814-144473814 on Build GRCh38
rs377389682 Chr.5: 144473930-144473930 on Build GRCh38
rs750586344 Chr.5: 144473773-144473773 on Build GRCh38
rs867663751 Chr.5: 144473876-144473876 on Build GRCh38
rs142821752 Chr.5: 144473934-144473934 on Build GRCh38
rs779094393 Chr.5: 144473671-144473671 on Build GRCh38
rs756604950 Chr.5: 144473616-144473616 on Build GRCh38
rs769674983 Chr.5: 144473792-144473792 on Build GRCh38
rs868542964 Chr.5: 144473857-144473857 on Build GRCh38
rs146014269 Chr.5: 144473736-144473736 on Build GRCh38
rs768373714 Chr.5: 144473691-144473691 on Build GRCh38
rs771594591 Chr.5: 144473823-144473823 on Build GRCh38
rs375766879 Chr.5: 144473787-144473787 on Build GRCh38
rs749245269 Chr.5: 144473805-144473805 on Build GRCh38
rs202165401 Chr.5: 144473906-144473906 on Build GRCh38
rs780997593 Chr.5: 144473782-144473782 on Build GRCh38
rs757325891 Chr.5: 144473769-144473769 on Build GRCh38
rs902294491 Chr.5: 144473999-144473999 on Build GRCh38
rs146760710 Chr.5: 144473764-144473764 on Build GRCh38
rs776304481 Chr.5: 144474030-144474030 on Build GRCh38
rs774023034 Chr.5: 144473808-144473808 on Build GRCh38
rs761142772 Chr.5: 144473760-144473760 on Build GRCh38

View Full Product Details