Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149618832-149619325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF37
Entrez Gene ID
389337
Gene Name
Rho guanine nucleotide exchange factor 37
Gene Aliases
-
UniGene
Hs.256206
Gene Chromosome Location
Chr.5: 149552049-149634968 on Build GRCh38
Transcripts
NM_001001669.2 XM_005268447.4 XM_005268448.1 XM_006714784.3 XM_011537642.2 XM_017009460.1 XM_017009461.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1064184 c.839C>T p.S280F Chr.5: 149618987-149618987 on Build GRCh38
5615488 c.811G>A p.D271N Chr.5: 149618959-149618959 on Build GRCh38
4417445 c.871G>A p.A291T Chr.5: 149619019-149619019 on Build GRCh38
1435091 c.842_849delTGTGTGTG p.L281fs*2 Chr.5: 149618990-149618997 on Build GRCh38
1245428 c.891G>T p.L297L Chr.5: 149619039-149619039 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs111494355 Chr.5: 149619229-149619229 on Build GRCh38
rs768482044 Chr.5: 149618913-149618913 on Build GRCh38
rs778432032 Chr.5: 149619123-149619123 on Build GRCh38
rs753545664 Chr.5: 149618947-149618947 on Build GRCh38
rs938716168 Chr.5: 149619127-149619127 on Build GRCh38
rs879108219 Chr.5: 149619137-149619137 on Build GRCh38
rs767015920 Chr.5: 149618940-149618941 on Build GRCh38
rs775915621 Chr.5: 149619294-149619294 on Build GRCh38
rs776522884 Chr.5: 149618918-149618918 on Build GRCh38
rs374402620 Chr.5: 149619071-149619071 on Build GRCh38
rs766377123 Chr.5: 149619075-149619075 on Build GRCh38
rs929923548 Chr.5: 149619051-149619051 on Build GRCh38
rs556542147 Chr.5: 149619228-149619228 on Build GRCh38
rs752226394 Chr.5: 149619153-149619153 on Build GRCh38
rs749393957 Chr.5: 149618896-149618896 on Build GRCh38
rs374895852 Chr.5: 149618860-149618860 on Build GRCh38
rs138480167 Chr.5: 149619015-149619015 on Build GRCh38
rs766033761 Chr.5: 149618883-149618883 on Build GRCh38
rs773017951 Chr.5: 149618991-149618991 on Build GRCh38
rs551886267 Chr.5: 149619122-149619122 on Build GRCh38
rs759060909 Chr.5: 149619093-149619093 on Build GRCh38
rs199665632 Chr.5: 149619043-149619043 on Build GRCh38
rs761455984 Chr.5: 149618941-149618941 on Build GRCh38
rs762952527 Chr.5: 149619072-149619072 on Build GRCh38
rs377074496 Chr.5: 149619039-149619039 on Build GRCh38
rs371470773 Chr.5: 149619056-149619056 on Build GRCh38
rs762099156 Chr.5: 149618861-149618861 on Build GRCh38
rs1000143839 Chr.5: 149619154-149619154 on Build GRCh38
rs1039406603 Chr.5: 149619074-149619074 on Build GRCh38
rs1034979909 Chr.5: 149619258-149619258 on Build GRCh38
rs748039067 Chr.5: 149619059-149619059 on Build GRCh38
rs750016059 Chr.5: 149618951-149618951 on Build GRCh38
rs964636382 Chr.5: 149619269-149619269 on Build GRCh38
rs767205206 Chr.5: 149619094-149619094 on Build GRCh38
rs542328445 Chr.5: 149619246-149619246 on Build GRCh38
rs78412086 Chr.5: 149619161-149619161 on Build GRCh38
rs774614644 Chr.5: 149618905-149618905 on Build GRCh38
rs927859419 Chr.5: 149619047-149619047 on Build GRCh38
rs770997602 Chr.5: 149618897-149618897 on Build GRCh38
rs746068826 Chr.5: 149618911-149618911 on Build GRCh38
rs993213026 Chr.5: 149619215-149619215 on Build GRCh38
rs774018882 Chr.5: 149619081-149619081 on Build GRCh38
rs78618835 Chr.5: 149619019-149619019 on Build GRCh38
rs534284876 Chr.5: 149618870-149618870 on Build GRCh38
rs983078283 Chr.5: 149619265-149619265 on Build GRCh38
rs745910491 Chr.5: 149619057-149619057 on Build GRCh38
rs756370096 Chr.5: 149619022-149619022 on Build GRCh38
rs374852028 Chr.5: 149619139-149619139 on Build GRCh38
rs751293426 Chr.5: 149619009-149619009 on Build GRCh38
rs1037913401 Chr.5: 149619163-149619163 on Build GRCh38
rs767231512 Chr.5: 149619016-149619016 on Build GRCh38
rs1017918926 Chr.5: 149619264-149619264 on Build GRCh38
rs765224432 Chr.5: 149618959-149618959 on Build GRCh38
rs762610623 Chr.5: 149618995-149618995 on Build GRCh38
rs369712407 Chr.5: 149619069-149619069 on Build GRCh38
rs921003381 Chr.5: 149619293-149619293 on Build GRCh38
rs778934598 Chr.5: 149619044-149619044 on Build GRCh38
rs551082835 Chr.5: 149618884-149618884 on Build GRCh38
rs187760278 Chr.5: 149619136-149619136 on Build GRCh38
rs1011091846 Chr.5: 149619227-149619227 on Build GRCh38
rs538294147 Chr.5: 149619197-149619197 on Build GRCh38
rs776102180 Chr.5: 149619066-149619066 on Build GRCh38
rs567476859 Chr.5: 149619058-149619058 on Build GRCh38
rs7728256 Chr.5: 149619284-149619284 on Build GRCh38
rs1050750184 Chr.5: 149619142-149619142 on Build GRCh38
rs755743670 Chr.5: 149618986-149618987 on Build GRCh38
rs765961852 Chr.5: 149619004-149619004 on Build GRCh38
rs779858044 Chr.5: 149619057-149619057 on Build GRCh38
rs954496174 Chr.5: 149619238-149619238 on Build GRCh38
rs914654473 Chr.5: 149619052-149619052 on Build GRCh38
rs975247039 Chr.5: 149619290-149619290 on Build GRCh38

View Full Product Details