Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179765638-179766143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAML1
Entrez Gene ID
9794
Gene Name
mastermind like transcriptional coactivator 1
Gene Aliases
Mam-1, Mam1
UniGene
Hs.631951
Gene Chromosome Location
Chr.5: 179732850-179777286 on Build GRCh38
Transcripts
NM_014757.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4883327 c.1108G>T p.A370S Chr.5: 179766118-179766118 on Build GRCh38
3994238 c.987G>A p.L329L Chr.5: 179765886-179765886 on Build GRCh38
397304 c.1224G>A p.Q408Q Chr.5: 179766123-179766123 on Build GRCh38
1066695 c.793G>T p.E265* Chr.5: 179765803-179765803 on Build GRCh38
5014554 c.717C>G p.N239K Chr.5: 179765727-179765727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762246688 Chr.5: 179766116-179766116 on Build GRCh38
rs772531851 Chr.5: 179766088-179766088 on Build GRCh38
rs140999569 Chr.5: 179765832-179765832 on Build GRCh38
rs759419227 Chr.5: 179765702-179765702 on Build GRCh38
rs959261236 Chr.5: 179765830-179765830 on Build GRCh38
rs913231984 Chr.5: 179765864-179765864 on Build GRCh38
rs747105579 Chr.5: 179765968-179765968 on Build GRCh38
rs764633954 Chr.5: 179766033-179766033 on Build GRCh38
rs759216464 Chr.5: 179766105-179766105 on Build GRCh38
rs954739173 Chr.5: 179765802-179765802 on Build GRCh38
rs776692289 Chr.5: 179765987-179765987 on Build GRCh38
rs777774533 Chr.5: 179765876-179765876 on Build GRCh38
rs753262731 Chr.5: 179765935-179765935 on Build GRCh38
rs869199048 Chr.5: 179765870-179765870 on Build GRCh38
rs199770098 Chr.5: 179766057-179766057 on Build GRCh38
rs199827612 Chr.5: 179765951-179765951 on Build GRCh38
rs748600395 Chr.5: 179765873-179765873 on Build GRCh38
rs148455396 Chr.5: 179765718-179765718 on Build GRCh38
rs557255881 Chr.5: 179765751-179765751 on Build GRCh38
rs35681801 Chr.5: 179765799-179765800 on Build GRCh38
rs770441866 Chr.5: 179766094-179766094 on Build GRCh38
rs772641668 Chr.5: 179765799-179765799 on Build GRCh38
rs890601085 Chr.5: 179766058-179766058 on Build GRCh38
rs775086037 Chr.5: 179765970-179765970 on Build GRCh38
rs996292105 Chr.5: 179765744-179765744 on Build GRCh38
rs994472392 Chr.5: 179766119-179766119 on Build GRCh38
rs371564064 Chr.5: 179766104-179766104 on Build GRCh38
rs886091042 Chr.5: 179766054-179766054 on Build GRCh38
rs757304194 Chr.5: 179765967-179765967 on Build GRCh38
rs200781660 Chr.5: 179766048-179766048 on Build GRCh38
rs764838678 Chr.5: 179765999-179765999 on Build GRCh38
rs751546910 Chr.5: 179765700-179765700 on Build GRCh38
rs752965564 Chr.5: 179766023-179766023 on Build GRCh38
rs762697925 Chr.5: 179766089-179766089 on Build GRCh38
rs756666515 Chr.5: 179766032-179766032 on Build GRCh38
rs752553028 Chr.5: 179765835-179765835 on Build GRCh38
rs766966774 Chr.5: 179765704-179765704 on Build GRCh38
rs151052902 Chr.5: 179765827-179765827 on Build GRCh38
rs752185395 Chr.5: 179765706-179765706 on Build GRCh38
rs763137822 Chr.5: 179765788-179765788 on Build GRCh38
rs755439959 Chr.5: 179765923-179765923 on Build GRCh38
rs967627602 Chr.5: 179765877-179765877 on Build GRCh38
rs772977594 Chr.5: 179766020-179766020 on Build GRCh38
rs755279459 Chr.5: 179765866-179765866 on Build GRCh38
rs575822811 Chr.5: 179765775-179765775 on Build GRCh38
rs761066690 Chr.5: 179766103-179766103 on Build GRCh38
rs756277424 Chr.5: 179765874-179765874 on Build GRCh38
rs774092069 Chr.5: 179765820-179765820 on Build GRCh38
rs775562183 Chr.5: 179765888-179765888 on Build GRCh38
rs916283309 Chr.5: 179765994-179765994 on Build GRCh38
rs926136518 Chr.5: 179765899-179765899 on Build GRCh38
rs749517746 Chr.5: 179765879-179765879 on Build GRCh38
rs373506544 Chr.5: 179766077-179766077 on Build GRCh38
rs747545832 Chr.5: 179765734-179765734 on Build GRCh38
rs773290253 Chr.5: 179765668-179765668 on Build GRCh38
rs771731691 Chr.5: 179765885-179765885 on Build GRCh38
rs751927067 Chr.5: 179765865-179765865 on Build GRCh38
rs763707827 Chr.5: 179765898-179765898 on Build GRCh38
rs199996110 Chr.5: 179765910-179765910 on Build GRCh38
rs763505113 Chr.5: 179765839-179765839 on Build GRCh38
rs745723439 Chr.5: 179765883-179765883 on Build GRCh38
rs978970827 Chr.5: 179765880-179765880 on Build GRCh38
rs780565545 Chr.5: 179765863-179765863 on Build GRCh38
rs781348203 Chr.5: 179766079-179766079 on Build GRCh38
rs748314960 Chr.5: 179765772-179765772 on Build GRCh38
rs879206269 Chr.5: 179765730-179765730 on Build GRCh38
rs769444821 Chr.5: 179766084-179766084 on Build GRCh38
rs760514083 Chr.5: 179765895-179765895 on Build GRCh38
rs142619738 Chr.5: 179765726-179765726 on Build GRCh38
rs768670271 Chr.5: 179765978-179765978 on Build GRCh38
rs369643072 Chr.5: 179766076-179766076 on Build GRCh38
rs781744208 Chr.5: 179765867-179765867 on Build GRCh38
rs368036232 Chr.5: 179766021-179766021 on Build GRCh38
rs758663524 Chr.5: 179765724-179765724 on Build GRCh38
rs753600218 Chr.5: 179765844-179765844 on Build GRCh38
rs776209806 Chr.5: 179765903-179765903 on Build GRCh38
rs750668310 Chr.5: 179766074-179766074 on Build GRCh38
rs1004814335 Chr.5: 179766056-179766056 on Build GRCh38
rs146285187 Chr.5: 179765673-179765673 on Build GRCh38
rs774031818 Chr.5: 179766101-179766101 on Build GRCh38
rs202236629 Chr.5: 179765921-179765921 on Build GRCh38
rs778196575 Chr.5: 179765735-179765735 on Build GRCh38
rs767761852 Chr.5: 179765932-179765932 on Build GRCh38
rs749317962 Chr.5: 179765950-179765950 on Build GRCh38
rs866033737 Chr.5: 179766086-179766086 on Build GRCh38
rs777862608 Chr.5: 179765948-179765948 on Build GRCh38
rs766129941 Chr.5: 179765674-179765674 on Build GRCh38
rs777540931 Chr.5: 179765717-179765717 on Build GRCh38
rs144589897 Chr.5: 179765850-179765850 on Build GRCh38
rs760270059 Chr.5: 179765836-179765836 on Build GRCh38
rs770519131 Chr.5: 179765915-179765915 on Build GRCh38
rs748087606 Chr.5: 179766080-179766080 on Build GRCh38
rs11549693 Chr.5: 179765783-179765783 on Build GRCh38
rs774153392 Chr.5: 179765882-179765882 on Build GRCh38
rs761452849 Chr.5: 179765993-179765993 on Build GRCh38
rs746966618 Chr.5: 179765974-179765974 on Build GRCh38
rs755737129 Chr.5: 179765707-179765707 on Build GRCh38
rs756512969 Chr.5: 179765941-179765941 on Build GRCh38

View Full Product Details