Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180609813-180610312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT4
Entrez Gene ID
2324
Gene Name
fms related tyrosine kinase 4
Gene Aliases
FLT-4, FLT41, LMPH1A, PCL, VEGFR-3, VEGFR3
UniGene
Hs.646917
Gene Chromosome Location
Chr.5: 180601506-180650298 on Build GRCh38
Transcripts
NM_002020.4 NM_182925.4 XM_011534478.2 XM_011534484.2 XM_017009263.1 XM_017009264.1 XM_017009265.1 XM_017009266.1 XM_017009267.1 XM_017009268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5403230 c.3729G>A p.G1243G Chr.5: 180609983-180609983 on Build GRCh38
5608888 c.3720G>A p.L1240L Chr.5: 180609992-180609992 on Build GRCh38
7116465 c.3696C>T p.N1232N Chr.5: 180610016-180610016 on Build GRCh38
482649 c.3731C>T p.A1244V Chr.5: 180609981-180609981 on Build GRCh38
6915064 c.3766G>C p.E1256Q Chr.5: 180609946-180609946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562421112 Chr.5: 180610190-180610190 on Build GRCh38
rs775755471 Chr.5: 180609856-180609856 on Build GRCh38
rs769978743 Chr.5: 180609962-180609962 on Build GRCh38
rs971065703 Chr.5: 180609969-180609969 on Build GRCh38
rs376136714 Chr.5: 180610059-180610059 on Build GRCh38
rs890067233 Chr.5: 180609850-180609850 on Build GRCh38
rs2242219 Chr.5: 180609871-180609871 on Build GRCh38
rs948259444 Chr.5: 180610122-180610122 on Build GRCh38
rs780696184 Chr.5: 180609937-180609937 on Build GRCh38
rs931449105 Chr.5: 180610053-180610053 on Build GRCh38
rs911384461 Chr.5: 180610282-180610282 on Build GRCh38
rs759443068 Chr.5: 180610077-180610077 on Build GRCh38
rs1014875495 Chr.5: 180610200-180610200 on Build GRCh38
rs766960409 Chr.5: 180610000-180610000 on Build GRCh38
rs762732020 Chr.5: 180609973-180609973 on Build GRCh38
rs758849473 Chr.5: 180610040-180610040 on Build GRCh38
rs970630938 Chr.5: 180610184-180610184 on Build GRCh38
rs373893251 Chr.5: 180610281-180610281 on Build GRCh38
rs746207295 Chr.5: 180609934-180609934 on Build GRCh38
rs201542981 Chr.5: 180609951-180609951 on Build GRCh38
rs902175250 Chr.5: 180610120-180610120 on Build GRCh38
rs754430379 Chr.5: 180609895-180609895 on Build GRCh38
rs781175230 Chr.5: 180610054-180610054 on Build GRCh38
rs370452038 Chr.5: 180609998-180609998 on Build GRCh38
rs1044248124 Chr.5: 180610124-180610124 on Build GRCh38
rs541987226 Chr.5: 180610272-180610272 on Build GRCh38
rs370764266 Chr.5: 180610130-180610130 on Build GRCh38
rs534953690 Chr.5: 180610286-180610286 on Build GRCh38
rs774148239 Chr.5: 180610070-180610070 on Build GRCh38
rs770715511 Chr.5: 180610063-180610063 on Build GRCh38
rs961661100 Chr.5: 180610203-180610203 on Build GRCh38
rs758619712 Chr.5: 180610082-180610082 on Build GRCh38
rs545823626 Chr.5: 180610185-180610185 on Build GRCh38
rs752230258 Chr.5: 180610029-180610029 on Build GRCh38
rs376656307 Chr.5: 180610021-180610021 on Build GRCh38
rs752325680 Chr.5: 180609920-180609920 on Build GRCh38
rs1007872322 Chr.5: 180609854-180609854 on Build GRCh38
rs750704776 Chr.5: 180609977-180609977 on Build GRCh38
rs1041686765 Chr.5: 180610291-180610291 on Build GRCh38
rs770911540 Chr.5: 180609996-180609996 on Build GRCh38
rs1037109406 Chr.5: 180610001-180610001 on Build GRCh38
rs751147187 Chr.5: 180609877-180609877 on Build GRCh38
rs555884475 Chr.5: 180610131-180610131 on Build GRCh38
rs747495466 Chr.5: 180609950-180609950 on Build GRCh38
rs759044742 Chr.5: 180609935-180609935 on Build GRCh38
rs1003993593 Chr.5: 180610284-180610284 on Build GRCh38
rs371214562 Chr.5: 180609907-180609907 on Build GRCh38
rs755743097 Chr.5: 180609923-180609923 on Build GRCh38
rs773030985 Chr.5: 180609970-180609970 on Build GRCh38
rs535200757 Chr.5: 180610084-180610084 on Build GRCh38
rs931401662 Chr.5: 180610261-180610261 on Build GRCh38
rs774986638 Chr.5: 180609882-180609882 on Build GRCh38
rs755462273 Chr.5: 180610051-180610051 on Build GRCh38
rs1012005475 Chr.5: 180610148-180610148 on Build GRCh38
rs768983796 Chr.5: 180609862-180609862 on Build GRCh38
rs772685630 Chr.5: 180609855-180609855 on Build GRCh38
rs766271754 Chr.5: 180609874-180609874 on Build GRCh38
rs763837459 Chr.5: 180610031-180610031 on Build GRCh38
rs147580317 Chr.5: 180610105-180610105 on Build GRCh38
rs555429852 Chr.5: 180610042-180610042 on Build GRCh38
rs367771647 Chr.5: 180610045-180610045 on Build GRCh38
rs919647363 Chr.5: 180609848-180609849 on Build GRCh38
rs992445647 Chr.5: 180610080-180610080 on Build GRCh38
rs200178139 Chr.5: 180609972-180609972 on Build GRCh38
rs893581541 Chr.5: 180610147-180610147 on Build GRCh38
rs112990569 Chr.5: 180610057-180610057 on Build GRCh38
rs760444440 Chr.5: 180610030-180610030 on Build GRCh38
rs995354891 Chr.5: 180610206-180610206 on Build GRCh38
rs190270414 Chr.5: 180610252-180610252 on Build GRCh38
rs1023432854 Chr.5: 180610159-180610159 on Build GRCh38
rs375662657 Chr.5: 180609924-180609924 on Build GRCh38
rs944297073 Chr.5: 180610060-180610060 on Build GRCh38
rs1008162348 Chr.5: 180610178-180610180 on Build GRCh38
rs747059125 Chr.5: 180609857-180609857 on Build GRCh38
rs903937918 Chr.5: 180610128-180610128 on Build GRCh38
rs935454750 Chr.5: 180610129-180610129 on Build GRCh38
rs142649193 Chr.5: 180609959-180609959 on Build GRCh38
rs972750223 Chr.5: 180610208-180610208 on Build GRCh38
rs572585085 Chr.5: 180610058-180610058 on Build GRCh38
rs201290417 Chr.5: 180609872-180609872 on Build GRCh38
rs750879918 Chr.5: 180610035-180610035 on Build GRCh38
rs375640346 Chr.5: 180609870-180609870 on Build GRCh38
rs999488864 Chr.5: 180609933-180609933 on Build GRCh38
rs748651988 Chr.5: 180609837-180609837 on Build GRCh38

View Full Product Details