Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 17763601-17764105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF13A
Entrez Gene ID
63971
Gene Name
kinesin family member 13A
Gene Aliases
RBKIN, bA500C11.2
UniGene
Hs.94499
Gene Chromosome Location
Chr.6: 17759183-17987623 on Build GRCh38
Transcripts
NM_001105566.2 NM_001105567.2 NM_001105568.2 NM_022113.5 XM_017011188.1 XM_017011189.1 XM_017011190.1 XM_017011191.1 XM_017011192.1 XM_017011193.1

SNP Detail

SNP ID Chromosome Location
rs868410905 Chr.6: 17763621-17763621 on Build GRCh38
rs928922573 Chr.6: 17763874-17763874 on Build GRCh38
rs183885434 Chr.6: 17763945-17763945 on Build GRCh38
rs1020921006 Chr.6: 17763635-17763635 on Build GRCh38
rs753815632 Chr.6: 17764053-17764053 on Build GRCh38
rs534793822 Chr.6: 17763975-17763975 on Build GRCh38
rs1006281091 Chr.6: 17763918-17763918 on Build GRCh38
rs764420027 Chr.6: 17763974-17763974 on Build GRCh38
rs984249175 Chr.6: 17763666-17763666 on Build GRCh38
rs974702135 Chr.6: 17763665-17763666 on Build GRCh38
rs1005598882 Chr.6: 17763966-17763966 on Build GRCh38
rs972979986 Chr.6: 17763994-17763994 on Build GRCh38
rs1003942590 Chr.6: 17763940-17763940 on Build GRCh38
rs80074989 Chr.6: 17763765-17763765 on Build GRCh38
rs576278202 Chr.6: 17764014-17764014 on Build GRCh38
rs369532909 Chr.6: 17764070-17764070 on Build GRCh38
rs910084791 Chr.6: 17763680-17763680 on Build GRCh38
rs1030841143 Chr.6: 17763999-17763999 on Build GRCh38
rs959438426 Chr.6: 17763950-17763950 on Build GRCh38
rs891714659 Chr.6: 17763628-17763628 on Build GRCh38
rs75469261 Chr.6: 17763925-17763925 on Build GRCh38
rs1044654 Chr.6: 17764048-17764048 on Build GRCh38
rs776223427 Chr.6: 17764080-17764080 on Build GRCh38
rs192244902 Chr.6: 17763871-17763871 on Build GRCh38
rs773010262 Chr.6: 17763963-17763974 on Build GRCh38
rs182845252 Chr.6: 17763739-17763739 on Build GRCh38
rs766252941 Chr.6: 17764074-17764074 on Build GRCh38
rs574003667 Chr.6: 17763786-17763786 on Build GRCh38
rs771685620 Chr.6: 17764062-17764062 on Build GRCh38
rs1010605721 Chr.6: 17763632-17763632 on Build GRCh38
rs149671299 Chr.6: 17763681-17763681 on Build GRCh38
rs535025987 Chr.6: 17763814-17763814 on Build GRCh38
rs80258593 Chr.6: 17763980-17763980 on Build GRCh38
rs188317575 Chr.6: 17763774-17763774 on Build GRCh38
rs977958474 Chr.6: 17763691-17763691 on Build GRCh38
rs1046050761 Chr.6: 17763873-17763873 on Build GRCh38
rs1030328757 Chr.6: 17763977-17763977 on Build GRCh38
rs760397336 Chr.6: 17764071-17764071 on Build GRCh38
rs543661912 Chr.6: 17764047-17764047 on Build GRCh38
rs982196393 Chr.6: 17763641-17763641 on Build GRCh38
rs74913055 Chr.6: 17763916-17763916 on Build GRCh38
rs752833874 Chr.6: 17763903-17763903 on Build GRCh38
rs959479398 Chr.6: 17763662-17763662 on Build GRCh38
rs1010117369 Chr.6: 17763897-17763897 on Build GRCh38
rs570195315 Chr.6: 17763745-17763745 on Build GRCh38
rs193156590 Chr.6: 17763731-17763731 on Build GRCh38
rs922267697 Chr.6: 17763713-17763713 on Build GRCh38
rs906507777 Chr.6: 17763911-17763911 on Build GRCh38
rs973396263 Chr.6: 17763707-17763707 on Build GRCh38
rs1033740353 Chr.6: 17763649-17763649 on Build GRCh38
rs777227022 Chr.6: 17763757-17763757 on Build GRCh38
rs961861319 Chr.6: 17763988-17763988 on Build GRCh38
rs916265293 Chr.6: 17763772-17763772 on Build GRCh38
rs971141010 Chr.6: 17763640-17763640 on Build GRCh38
rs1047894136 Chr.6: 17763910-17763910 on Build GRCh38

View Full Product Details