Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41905532-41906038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED20
Entrez Gene ID
9477
Gene Name
mediator complex subunit 20
Gene Aliases
PRO0213, TRFP
UniGene
Hs.278434
Gene Chromosome Location
Chr.6: 41905353-41921147 on Build GRCh38
Transcripts
NM_001305455.1 NM_001305456.1 NM_001305457.1 NM_004275.4 NR_131160.1

SNP Detail

SNP ID Chromosome Location
rs776299321 Chr.6: 41905661-41905661 on Build GRCh38
rs201104795 Chr.6: 41905998-41905998 on Build GRCh38
rs972756699 Chr.6: 41905596-41905596 on Build GRCh38
rs561081262 Chr.6: 41905927-41905927 on Build GRCh38
rs1039590370 Chr.6: 41905860-41905860 on Build GRCh38
rs571203934 Chr.6: 41905881-41905881 on Build GRCh38
rs190277806 Chr.6: 41905816-41905816 on Build GRCh38
rs963962462 Chr.6: 41905602-41905602 on Build GRCh38
rs1000527072 Chr.6: 41905637-41905637 on Build GRCh38
rs142076626 Chr.6: 41905903-41905903 on Build GRCh38
rs574262392 Chr.6: 41905720-41905720 on Build GRCh38
rs917685419 Chr.6: 41905686-41905686 on Build GRCh38
rs182046694 Chr.6: 41905835-41905835 on Build GRCh38
rs151319098 Chr.6: 41906017-41906017 on Build GRCh38
rs772220965 Chr.6: 41905656-41905656 on Build GRCh38
rs769871429 Chr.6: 41905692-41905692 on Build GRCh38
rs762622416 Chr.6: 41905738-41905738 on Build GRCh38
rs895957182 Chr.6: 41905843-41905843 on Build GRCh38
rs981584883 Chr.6: 41905714-41905714 on Build GRCh38
rs768556669 Chr.6: 41905785-41905785 on Build GRCh38
rs1032461573 Chr.6: 41905644-41905644 on Build GRCh38
rs749958887 Chr.6: 41905956-41905956 on Build GRCh38
rs541106196 Chr.6: 41905908-41905908 on Build GRCh38
rs952829372 Chr.6: 41905928-41905928 on Build GRCh38
rs1043232581 Chr.6: 41905807-41905807 on Build GRCh38
rs994438343 Chr.6: 41905883-41905883 on Build GRCh38
rs1018162978 Chr.6: 41905593-41905593 on Build GRCh38
rs1008081038 Chr.6: 41905557-41905557 on Build GRCh38
rs148607776 Chr.6: 41905793-41905793 on Build GRCh38
rs527754433 Chr.6: 41905904-41905904 on Build GRCh38
rs1031993422 Chr.6: 41905966-41905966 on Build GRCh38
rs761462742 Chr.6: 41905792-41905792 on Build GRCh38
rs1001213880 Chr.6: 41905964-41905964 on Build GRCh38
rs984613161 Chr.6: 41905684-41905684 on Build GRCh38
rs373865535 Chr.6: 41905713-41905713 on Build GRCh38
rs892096264 Chr.6: 41905977-41905977 on Build GRCh38
rs529891732 Chr.6: 41906008-41906008 on Build GRCh38
rs778004990 Chr.6: 41905659-41905659 on Build GRCh38
rs988324729 Chr.6: 41905668-41905668 on Build GRCh38
rs551193749 Chr.6: 41905875-41905875 on Build GRCh38
rs956464168 Chr.6: 41905660-41905660 on Build GRCh38
rs773819826 Chr.6: 41905791-41905791 on Build GRCh38
rs1026077625 Chr.6: 41905638-41905643 on Build GRCh38
rs562274823 Chr.6: 41905728-41905728 on Build GRCh38
rs952717979 Chr.6: 41905677-41905677 on Build GRCh38
rs937630860 Chr.6: 41905723-41905723 on Build GRCh38
rs919953087 Chr.6: 41905775-41905775 on Build GRCh38
rs554377283 Chr.6: 41905719-41905719 on Build GRCh38

View Full Product Details