Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 151536306-151536850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC170
Entrez Gene ID
80129
Gene Name
coiled-coil domain containing 170
Gene Aliases
C6orf97, bA282P11.1
UniGene
Hs.660044
Gene Chromosome Location
Chr.6: 151494040-151621193 on Build GRCh38
Transcripts
NM_025059.3 XM_011536147.2 XM_011536148.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198974 c.137G>A p.R46Q Chr.6: 151536397-151536397 on Build GRCh38
259849 c.167T>G p.F56C Chr.6: 151536427-151536427 on Build GRCh38
239089 c.85C>A p.P29T Chr.6: 151536345-151536345 on Build GRCh38
740595 c.82G>T p.V28F Chr.6: 151536342-151536342 on Build GRCh38
3715429 c.169G>T p.E57* Chr.6: 151536429-151536429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765819188 Chr.6: 151536779-151536780 on Build GRCh38
rs764981257 Chr.6: 151536355-151536355 on Build GRCh38
rs1022977985 Chr.6: 151536353-151536353 on Build GRCh38
rs775903848 Chr.6: 151536775-151536779 on Build GRCh38
rs929690343 Chr.6: 151536517-151536519 on Build GRCh38
rs776261278 Chr.6: 151536346-151536346 on Build GRCh38
rs763893224 Chr.6: 151536481-151536481 on Build GRCh38
rs910890599 Chr.6: 151536509-151536509 on Build GRCh38
rs140337376 Chr.6: 151536707-151536707 on Build GRCh38
rs947206587 Chr.6: 151536647-151536647 on Build GRCh38
rs752264786 Chr.6: 151536356-151536356 on Build GRCh38
rs550538688 Chr.6: 151536577-151536577 on Build GRCh38
rs762372180 Chr.6: 151536339-151536339 on Build GRCh38
rs559723721 Chr.6: 151536774-151536776 on Build GRCh38
rs374285090 Chr.6: 151536805-151536805 on Build GRCh38
rs6905111 Chr.6: 151536344-151536344 on Build GRCh38
rs11967028 Chr.6: 151536740-151536740 on Build GRCh38
rs147822104 Chr.6: 151536579-151536579 on Build GRCh38
rs938636175 Chr.6: 151536602-151536602 on Build GRCh38
rs182979274 Chr.6: 151536590-151536590 on Build GRCh38
rs199810277 Chr.6: 151536337-151536337 on Build GRCh38
rs935100616 Chr.6: 151536786-151536786 on Build GRCh38
rs768646774 Chr.6: 151536774-151536778 on Build GRCh38
rs994737113 Chr.6: 151536702-151536702 on Build GRCh38
rs750586909 Chr.6: 151536418-151536418 on Build GRCh38
rs375681294 Chr.6: 151536333-151536333 on Build GRCh38
rs78462879 Chr.6: 151536817-151536817 on Build GRCh38
rs780550074 Chr.6: 151536354-151536354 on Build GRCh38
rs201797826 Chr.6: 151536374-151536374 on Build GRCh38
rs974176926 Chr.6: 151536506-151536506 on Build GRCh38
rs905875986 Chr.6: 151536663-151536663 on Build GRCh38
rs780372313 Chr.6: 151536440-151536440 on Build GRCh38
rs200830022 Chr.6: 151536475-151536475 on Build GRCh38
rs1043881838 Chr.6: 151536531-151536531 on Build GRCh38
rs557748866 Chr.6: 151536808-151536808 on Build GRCh38
rs755048144 Chr.6: 151536514-151536514 on Build GRCh38
rs763833974 Chr.6: 151536376-151536376 on Build GRCh38
rs910860955 Chr.6: 151536727-151536727 on Build GRCh38
rs915797707 Chr.6: 151536639-151536639 on Build GRCh38
rs775236606 Chr.6: 151536477-151536477 on Build GRCh38
rs1012171451 Chr.6: 151536658-151536658 on Build GRCh38
rs896368816 Chr.6: 151536617-151536617 on Build GRCh38
rs1027536660 Chr.6: 151536747-151536747 on Build GRCh38
rs1056269899 Chr.6: 151536789-151536789 on Build GRCh38
rs146031527 Chr.6: 151536793-151536793 on Build GRCh38
rs946465769 Chr.6: 151536522-151536522 on Build GRCh38
rs375627254 Chr.6: 151536396-151536396 on Build GRCh38
rs756120638 Chr.6: 151536419-151536419 on Build GRCh38
rs1005580044 Chr.6: 151536670-151536670 on Build GRCh38
rs1044392913 Chr.6: 151536662-151536662 on Build GRCh38
rs555579253 Chr.6: 151536685-151536685 on Build GRCh38
rs979147188 Chr.6: 151536739-151536739 on Build GRCh38
rs779164339 Chr.6: 151536515-151536515 on Build GRCh38
rs770779659 Chr.6: 151536726-151536726 on Build GRCh38
rs55663799 Chr.6: 151536794-151536798 on Build GRCh38
rs759135370 Chr.6: 151536463-151536463 on Build GRCh38
rs1018486986 Chr.6: 151536737-151536737 on Build GRCh38
rs1008464949 Chr.6: 151536712-151536713 on Build GRCh38
rs1031392692 Chr.6: 151536383-151536383 on Build GRCh38
rs968106402 Chr.6: 151536735-151536735 on Build GRCh38
rs951190487 Chr.6: 151536788-151536788 on Build GRCh38
rs917773041 Chr.6: 151536791-151536791 on Build GRCh38
rs985695675 Chr.6: 151536724-151536724 on Build GRCh38
rs535301530 Chr.6: 151536648-151536648 on Build GRCh38
rs1832727 Chr.6: 151536578-151536578 on Build GRCh38
rs1017073506 Chr.6: 151536362-151536362 on Build GRCh38
rs961830058 Chr.6: 151536370-151536370 on Build GRCh38
rs955582483 Chr.6: 151536723-151536723 on Build GRCh38
rs749422970 Chr.6: 151536441-151536441 on Build GRCh38
rs759965922 Chr.6: 151536471-151536471 on Build GRCh38
rs982919670 Chr.6: 151536790-151536790 on Build GRCh38
rs987299422 Chr.6: 151536799-151536799 on Build GRCh38
rs762524224 Chr.6: 151536478-151536478 on Build GRCh38
rs777077287 Chr.6: 151536466-151536466 on Build GRCh38
rs528679889 Chr.6: 151536388-151536388 on Build GRCh38
rs961522714 Chr.6: 151536700-151536700 on Build GRCh38
rs1055711864 Chr.6: 151536633-151536633 on Build GRCh38
rs748508645 Chr.6: 151536458-151536458 on Build GRCh38
rs781096492 Chr.6: 151536397-151536397 on Build GRCh38
rs371345564 Chr.6: 151536799-151536799 on Build GRCh38
rs869071804 Chr.6: 151536790-151536794 on Build GRCh38
rs6931664 Chr.6: 151536459-151536459 on Build GRCh38
rs566615865 Chr.6: 151536621-151536621 on Build GRCh38
rs779174342 Chr.6: 151536774-151536779 on Build GRCh38
rs530957586 Chr.6: 151536539-151536539 on Build GRCh38
rs779355411 Chr.6: 151536455-151536455 on Build GRCh38
rs201381723 Chr.6: 151536375-151536375 on Build GRCh38
rs903951121 Chr.6: 151536676-151536676 on Build GRCh38
rs947921957 Chr.6: 151536792-151536792 on Build GRCh38
rs769121829 Chr.6: 151536338-151536338 on Build GRCh38
rs71014593 Chr.6: 151536796-151536798 on Build GRCh38

View Full Product Details