Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73735753-73736242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD11
Entrez Gene ID
83451
Gene Name
abhydrolase domain containing 11
Gene Aliases
PP1226, WBSCR21
UniGene
Hs.647045
Gene Chromosome Location
Chr.7: 73736094-73738867 on Build GRCh38
Transcripts
NM_001145364.2 NM_001301058.1 NM_001321382.1 NM_001321383.1 NM_148912.3 NM_148913.3 NR_026912.2 NR_135627.1 XM_006716144.2 XM_011516590.2 XM_011516591.1 NR_026690.1

Gene Symbol
ABHD11-AS1
Entrez Gene ID
171022
Gene Name
ABHD11 antisense RNA 1 (tail to tail)
Gene Aliases
LINC00035, NCRNA00035, WBSCR26
UniGene
Hs.650513
Gene Chromosome Location
Chr.7: 73735069-73736000 on Build GRCh38
Transcripts
NM_001145364.2 NM_001301058.1 NM_001321382.1 NM_001321383.1 NM_148912.3 NM_148913.3 NR_026912.2 NR_135627.1 XM_006716144.2 XM_011516590.2 XM_011516591.1 NR_026690.1

SNP Detail

SNP ID Chromosome Location
rs782676002 Chr.7: 73736030-73736030 on Build GRCh38
rs117303091 Chr.7: 73735829-73735829 on Build GRCh38
rs782506956 Chr.7: 73735937-73735937 on Build GRCh38
rs782436764 Chr.7: 73736173-73736173 on Build GRCh38
rs1043283300 Chr.7: 73735809-73735809 on Build GRCh38
rs916160744 Chr.7: 73736055-73736055 on Build GRCh38
rs782547525 Chr.7: 73736067-73736067 on Build GRCh38
rs200403289 Chr.7: 73735777-73735777 on Build GRCh38
rs1041421581 Chr.7: 73735835-73735835 on Build GRCh38
rs782605222 Chr.7: 73735828-73735828 on Build GRCh38
rs934713729 Chr.7: 73736104-73736104 on Build GRCh38
rs996684938 Chr.7: 73735904-73735904 on Build GRCh38
rs961398964 Chr.7: 73735974-73735974 on Build GRCh38
rs782335923 Chr.7: 73736207-73736207 on Build GRCh38
rs1035067905 Chr.7: 73735997-73735997 on Build GRCh38
rs1005332474 Chr.7: 73735965-73735965 on Build GRCh38
rs35181612 Chr.7: 73735791-73735791 on Build GRCh38
rs1059779 Chr.7: 73736181-73736181 on Build GRCh38
rs924824250 Chr.7: 73736086-73736086 on Build GRCh38
rs782210618 Chr.7: 73736213-73736213 on Build GRCh38
rs978908722 Chr.7: 73736068-73736068 on Build GRCh38
rs370893374 Chr.7: 73736057-73736057 on Build GRCh38
rs540181318 Chr.7: 73735776-73735776 on Build GRCh38
rs754915211 Chr.7: 73735935-73735935 on Build GRCh38
rs888402645 Chr.7: 73735946-73735946 on Build GRCh38
rs1041143207 Chr.7: 73736128-73736128 on Build GRCh38
rs371076218 Chr.7: 73736008-73736008 on Build GRCh38
rs562705747 Chr.7: 73736033-73736033 on Build GRCh38
rs148268213 Chr.7: 73736152-73736152 on Build GRCh38
rs947536417 Chr.7: 73735795-73735795 on Build GRCh38
rs549126223 Chr.7: 73735851-73735851 on Build GRCh38
rs782661767 Chr.7: 73736111-73736111 on Build GRCh38
rs544099611 Chr.7: 73735989-73735989 on Build GRCh38
rs1015456073 Chr.7: 73735973-73735973 on Build GRCh38
rs189308070 Chr.7: 73735878-73735878 on Build GRCh38

View Full Product Details